Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 63184219-63184715 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
JMJD1C
Entrez Gene ID
221037
Gene Name
jumonji domain containing 1C
Gene Aliases
TRIP-8, TRIP8
UniGene
Hs.413416
Gene Chromosome Location
Chr.10: 63167221-63522075 on Build GRCh38
Transcripts
NM_001282948.1 NM_001318153.1 NM_001318154.1 NM_001322252.1 NM_001322254.1 NM_001322258.1 NM_032776.2 NR_134512.1 XM_011539503.1 XM_017015897.1 XM_017015898.1 XM_017015899.1 XM_017015900.1 XM_017015901.1 XM_017015902.1 XM_017015903.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1211269 c.6214C>T p.R2072C Chr.10: 63184644-63184644 on Build GRCh38
458810 c.6904C>T p.H2302Y Chr.10: 63184665-63184665 on Build GRCh38
5616475 c.6900C>T p.A2300A Chr.10: 63184669-63184669 on Build GRCh38
5732519 c.6210delT p.F2070fs*6 Chr.10: 63184648-63184648 on Build GRCh38
1211268 c.6925C>T p.R2309C Chr.10: 63184644-63184644 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs925641366 Chr.10: 63184644-63184644 on Build GRCh38
rs537558036 Chr.10: 63184414-63184414 on Build GRCh38
rs879890823 Chr.10: 63184299-63184299 on Build GRCh38
rs892819387 Chr.10: 63184241-63184241 on Build GRCh38
rs79059103 Chr.10: 63184621-63184621 on Build GRCh38
rs943065146 Chr.10: 63184319-63184319 on Build GRCh38
rs548565400 Chr.10: 63184386-63184386 on Build GRCh38
rs942992139 Chr.10: 63184592-63184592 on Build GRCh38
rs986247284 Chr.10: 63184454-63184454 on Build GRCh38
rs373251590 Chr.10: 63184334-63184334 on Build GRCh38
rs570337138 Chr.10: 63184526-63184526 on Build GRCh38
rs776458468 Chr.10: 63184570-63184571 on Build GRCh38
rs949667058 Chr.10: 63184512-63184512 on Build GRCh38
rs762991366 Chr.10: 63184494-63184494 on Build GRCh38
rs551611026 Chr.10: 63184445-63184445 on Build GRCh38
rs559637074 Chr.10: 63184321-63184321 on Build GRCh38
rs528564569 Chr.10: 63184516-63184517 on Build GRCh38
rs962925724 Chr.10: 63184489-63184489 on Build GRCh38
rs769510538 Chr.10: 63184663-63184663 on Build GRCh38
rs145027090 Chr.10: 63184584-63184584 on Build GRCh38
rs879540249 Chr.10: 63184298-63184299 on Build GRCh38
rs113662753 Chr.10: 63184383-63184383 on Build GRCh38
rs1019942314 Chr.10: 63184289-63184289 on Build GRCh38
rs879640954 Chr.10: 63184303-63184304 on Build GRCh38
rs532205674 Chr.10: 63184306-63184306 on Build GRCh38
rs970434019 Chr.10: 63184465-63184465 on Build GRCh38
rs750248198 Chr.10: 63184491-63184491 on Build GRCh38
rs746895316 Chr.10: 63184643-63184643 on Build GRCh38
rs10740107 Chr.10: 63184557-63184557 on Build GRCh38
rs777873564 Chr.10: 63184612-63184612 on Build GRCh38
rs61853340 Chr.10: 63184415-63184415 on Build GRCh38
rs770245136 Chr.10: 63184337-63184337 on Build GRCh38
rs904444783 Chr.10: 63184387-63184387 on Build GRCh38
rs918737984 Chr.10: 63184516-63184516 on Build GRCh38
rs762780254 Chr.10: 63184536-63184536 on Build GRCh38
rs765785670 Chr.10: 63184681-63184681 on Build GRCh38
rs1030166921 Chr.10: 63184393-63184393 on Build GRCh38
rs181369726 Chr.10: 63184317-63184317 on Build GRCh38
rs935888184 Chr.10: 63184666-63184666 on Build GRCh38
rs528471416 Chr.10: 63184323-63184323 on Build GRCh38
rs75974662 Chr.10: 63184259-63184259 on Build GRCh38
rs187156794 Chr.10: 63184540-63184540 on Build GRCh38
rs1039330816 Chr.10: 63184345-63184345 on Build GRCh38
rs369060332 Chr.10: 63184657-63184657 on Build GRCh38
rs757627271 Chr.10: 63184241-63184241 on Build GRCh38
rs568420484 Chr.10: 63184392-63184392 on Build GRCh38
rs1000155920 Chr.10: 63184388-63184388 on Build GRCh38
rs751980419 Chr.10: 63184570-63184570 on Build GRCh38
rs141197246 Chr.10: 63184488-63184488 on Build GRCh38
rs563166657 Chr.10: 63184260-63184260 on Build GRCh38
rs762708090 Chr.10: 63184674-63184674 on Build GRCh38
rs56308427 Chr.10: 63184258-63184259 on Build GRCh38
rs756266028 Chr.10: 63184595-63184595 on Build GRCh38
rs199608318 Chr.10: 63184647-63184647 on Build GRCh38
rs762083028 Chr.10: 63184391-63184391 on Build GRCh38
rs938149103 Chr.10: 63184546-63184546 on Build GRCh38
rs776562309 Chr.10: 63184654-63184654 on Build GRCh38
rs919250452 Chr.10: 63184509-63184509 on Build GRCh38
rs749337724 Chr.10: 63184617-63184617 on Build GRCh38
rs1024410893 Chr.10: 63184464-63184464 on Build GRCh38
rs79393353 Chr.10: 63184257-63184257 on Build GRCh38
rs781528820 Chr.10: 63184580-63184580 on Build GRCh38
rs573485288 Chr.10: 63184547-63184547 on Build GRCh38
rs779086923 Chr.10: 63184638-63184638 on Build GRCh38
rs772773150 Chr.10: 63184667-63184667 on Build GRCh38
rs1014735449 Chr.10: 63184463-63184463 on Build GRCh38
rs980133532 Chr.10: 63184466-63184466 on Build GRCh38
rs755527983 Chr.10: 63184571-63184571 on Build GRCh38

View Full Product Details