Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 79899526-79900068 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNG2
Entrez Gene ID
26251
Gene Name
potassium voltage-gated channel modifier subfamily G member 2
Gene Aliases
KCNF2, KV6.2
UniGene
Hs.247905
Gene Chromosome Location
Chr.18: 79797963-79901894 on Build GRCh38
Transcripts
NM_012283.1 XM_011525918.2 XM_011525920.2 XM_017025704.1 XM_017025705.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3796538 c.1276G>A p.E426K Chr.18: 79899691-79899691 on Build GRCh38
6989068 c.1301C>T p.T434M Chr.18: 79899716-79899716 on Build GRCh38
7221246 c.1146G>T p.G382G Chr.18: 79899561-79899561 on Build GRCh38
3147566 c.1157C>T p.A386V Chr.18: 79899572-79899572 on Build GRCh38
7263518 c.1303C>T p.H435Y Chr.18: 79899718-79899718 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760752482 Chr.18: 79899730-79899730 on Build GRCh38
rs962820081 Chr.18: 79899874-79899874 on Build GRCh38
rs1032453831 Chr.18: 79899774-79899776 on Build GRCh38
rs778343980 Chr.18: 79899652-79899652 on Build GRCh38
rs777956598 Chr.18: 79899826-79899826 on Build GRCh38
rs746226223 Chr.18: 79899662-79899662 on Build GRCh38
rs927327083 Chr.18: 79899873-79899873 on Build GRCh38
rs377484468 Chr.18: 79899716-79899716 on Build GRCh38
rs890154326 Chr.18: 79899807-79899807 on Build GRCh38
rs761985274 Chr.18: 79899678-79899678 on Build GRCh38
rs954428789 Chr.18: 79899573-79899573 on Build GRCh38
rs755933671 Chr.18: 79899681-79899681 on Build GRCh38
rs751005765 Chr.18: 79899792-79899792 on Build GRCh38
rs749845712 Chr.18: 79899708-79899708 on Build GRCh38
rs538997842 Chr.18: 79899554-79899554 on Build GRCh38
rs774536374 Chr.18: 79899677-79899677 on Build GRCh38
rs143864015 Chr.18: 79899680-79899680 on Build GRCh38
rs891559004 Chr.18: 79900040-79900040 on Build GRCh38
rs753788445 Chr.18: 79899689-79899689 on Build GRCh38
rs764200959 Chr.18: 79899615-79899615 on Build GRCh38
rs374320136 Chr.18: 79899751-79899751 on Build GRCh38
rs890112167 Chr.18: 79899754-79899754 on Build GRCh38
rs139869249 Chr.18: 79899644-79899644 on Build GRCh38
rs918176713 Chr.18: 79899787-79899787 on Build GRCh38
rs971016178 Chr.18: 79899881-79899881 on Build GRCh38
rs764940621 Chr.18: 79899774-79899774 on Build GRCh38
rs776467192 Chr.18: 79899767-79899767 on Build GRCh38
rs555649072 Chr.18: 79899623-79899623 on Build GRCh38
rs557508303 Chr.18: 79899558-79899558 on Build GRCh38
rs1026159820 Chr.18: 79899991-79899991 on Build GRCh38
rs373621656 Chr.18: 79899679-79899679 on Build GRCh38
rs1038685512 Chr.18: 79899927-79899927 on Build GRCh38
rs773973336 Chr.18: 79899867-79899867 on Build GRCh38
rs758799034 Chr.18: 79899824-79899824 on Build GRCh38
rs193071442 Chr.18: 79899917-79899917 on Build GRCh38
rs146567075 Chr.18: 79899995-79899995 on Build GRCh38
rs541001173 Chr.18: 79899675-79899675 on Build GRCh38
rs548663326 Chr.18: 79899804-79899804 on Build GRCh38
rs973406574 Chr.18: 79900019-79900020 on Build GRCh38
rs912823082 Chr.18: 79899915-79899915 on Build GRCh38
rs779473882 Chr.18: 79899546-79899546 on Build GRCh38
rs553007399 Chr.18: 79900036-79900036 on Build GRCh38
rs528666051 Chr.18: 79899842-79899842 on Build GRCh38
rs747076633 Chr.18: 79899827-79899827 on Build GRCh38
rs187309710 Chr.18: 79899848-79899848 on Build GRCh38
rs759517480 Chr.18: 79899745-79899745 on Build GRCh38
rs7233469 Chr.18: 79899846-79899846 on Build GRCh38
rs751841591 Chr.18: 79899616-79899616 on Build GRCh38
rs773489126 Chr.18: 79899726-79899726 on Build GRCh38
rs75339189 Chr.18: 79899553-79899553 on Build GRCh38
rs762990430 Chr.18: 79899861-79899861 on Build GRCh38
rs759321246 Chr.18: 79899768-79899768 on Build GRCh38
rs949371428 Chr.18: 79899802-79899802 on Build GRCh38
rs202065839 Chr.18: 79899581-79899581 on Build GRCh38
rs780982352 Chr.18: 79899803-79899803 on Build GRCh38
rs962768811 Chr.18: 79899983-79899983 on Build GRCh38
rs779558647 Chr.18: 79899762-79899762 on Build GRCh38
rs370405014 Chr.18: 79899958-79899958 on Build GRCh38
rs974180907 Chr.18: 79899989-79899989 on Build GRCh38
rs760995612 Chr.18: 79899765-79899765 on Build GRCh38
rs755400361 Chr.18: 79899806-79899806 on Build GRCh38
rs1051649138 Chr.18: 79899831-79899831 on Build GRCh38
rs759501570 Chr.18: 79900010-79900011 on Build GRCh38
rs370084139 Chr.18: 79899641-79899641 on Build GRCh38
rs766097351 Chr.18: 79899682-79899682 on Build GRCh38
rs780608443 Chr.18: 79899755-79899755 on Build GRCh38
rs533564133 Chr.18: 79899892-79899893 on Build GRCh38
rs999943392 Chr.18: 79899942-79899942 on Build GRCh38
rs1034390292 Chr.18: 79899981-79899981 on Build GRCh38
rs748865441 Chr.18: 79899643-79899643 on Build GRCh38
rs569272864 Chr.18: 79899972-79899972 on Build GRCh38
rs999899003 Chr.18: 79899852-79899852 on Build GRCh38
rs748676278 Chr.18: 79899823-79899823 on Build GRCh38
rs770268069 Chr.18: 79899667-79899667 on Build GRCh38
rs984136550 Chr.18: 79899886-79899886 on Build GRCh38
rs904179976 Chr.18: 79899941-79899941 on Build GRCh38
rs561182267 Chr.18: 79899805-79899805 on Build GRCh38
rs760001156 Chr.18: 79899594-79899594 on Build GRCh38
rs766191697 Chr.18: 79899735-79899735 on Build GRCh38
rs770205282 Chr.18: 79899700-79899700 on Build GRCh38
rs1012339095 Chr.18: 79899979-79899979 on Build GRCh38
rs752211575 Chr.18: 79899747-79899747 on Build GRCh38
rs771118162 Chr.18: 79899766-79899766 on Build GRCh38
rs577587970 Chr.18: 79899696-79899696 on Build GRCh38
rs973480059 Chr.18: 79899896-79899896 on Build GRCh38
rs374210814 Chr.18: 79899702-79899702 on Build GRCh38
rs72982245 Chr.18: 79900031-79900031 on Build GRCh38
rs751828610 Chr.18: 79899971-79899972 on Build GRCh38
rs530473860 Chr.18: 79899764-79899764 on Build GRCh38
rs771723963 Chr.18: 79899654-79899654 on Build GRCh38
rs867148066 Chr.18: 79899593-79899593 on Build GRCh38
rs747828572 Chr.18: 79899653-79899653 on Build GRCh38
rs746572573 Chr.18: 79899699-79899699 on Build GRCh38
rs921629104 Chr.18: 79900026-79900026 on Build GRCh38
rs775767437 Chr.18: 79899606-79899606 on Build GRCh38
rs762747319 Chr.18: 79899784-79899784 on Build GRCh38
rs761182454 Chr.18: 79899704-79899706 on Build GRCh38
rs737504 Chr.18: 79900015-79900015 on Build GRCh38
rs912781782 Chr.18: 79899999-79899999 on Build GRCh38
rs200115509 Chr.18: 79899655-79899655 on Build GRCh38
rs756933104 Chr.18: 79899753-79899753 on Build GRCh38
rs1010498095 Chr.18: 79899963-79899963 on Build GRCh38
rs201158480 Chr.18: 79899763-79899763 on Build GRCh38
rs767250806 Chr.18: 79899625-79899625 on Build GRCh38
rs907118216 Chr.18: 79899971-79899971 on Build GRCh38
rs1051616217 Chr.18: 79899918-79899918 on Build GRCh38
rs767641357 Chr.18: 79899560-79899561 on Build GRCh38
rs538664631 Chr.18: 79899883-79899883 on Build GRCh38
rs959498044 Chr.18: 79899870-79899870 on Build GRCh38
rs78587287 Chr.18: 79899591-79899591 on Build GRCh38
rs753597483 Chr.18: 79899739-79899739 on Build GRCh38
rs769348837 Chr.18: 79899760-79899760 on Build GRCh38
rs747671179 Chr.18: 79899692-79899692 on Build GRCh38
rs750205598 Chr.18: 79899636-79899636 on Build GRCh38
rs769595833 Chr.18: 79899849-79899849 on Build GRCh38
rs1015628276 Chr.18: 79900000-79900000 on Build GRCh38
rs761939934 Chr.18: 79899717-79899717 on Build GRCh38
rs769258719 Chr.18: 79899709-79899709 on Build GRCh38
rs34106166 Chr.18: 79900024-79900024 on Build GRCh38
rs763117510 Chr.18: 79899614-79899614 on Build GRCh38
rs376255323 Chr.18: 79899785-79899785 on Build GRCh38
rs900255170 Chr.18: 79899931-79899931 on Build GRCh38
rs1032844018 Chr.18: 79899840-79899840 on Build GRCh38
rs907853627 Chr.18: 79899890-79899890 on Build GRCh38
rs769735266 Chr.18: 79899545-79899545 on Build GRCh38
rs989213177 Chr.18: 79899710-79899710 on Build GRCh38
rs1038734472 Chr.18: 79899818-79899818 on Build GRCh38
rs780027833 Chr.18: 79899642-79899642 on Build GRCh38
rs1042065663 Chr.18: 79899939-79899939 on Build GRCh38
rs982488403 Chr.18: 79899702-79899716 on Build GRCh38
rs747734392 Chr.18: 79899572-79899572 on Build GRCh38
rs991024505 Chr.18: 79899786-79899786 on Build GRCh38
rs764294896 Chr.18: 79899676-79899676 on Build GRCh38
rs775104508 Chr.18: 79899850-79899850 on Build GRCh38
rs891655399 Chr.18: 79899962-79899962 on Build GRCh38
rs922020802 Chr.18: 79899910-79899910 on Build GRCh38
rs544952689 Chr.18: 79899757-79899757 on Build GRCh38
rs776141768 Chr.18: 79899674-79899674 on Build GRCh38
rs951553744 Chr.18: 79899992-79899992 on Build GRCh38
rs745818640 Chr.18: 79899543-79899543 on Build GRCh38
rs974234887 Chr.18: 79899875-79899875 on Build GRCh38
rs758043184 Chr.18: 79899694-79899694 on Build GRCh38
rs752690222 Chr.18: 79899782-79899782 on Build GRCh38
rs571878476 Chr.18: 79899879-79899879 on Build GRCh38
rs911770615 Chr.18: 79899926-79899926 on Build GRCh38
rs763308297 Chr.18: 79899864-79899864 on Build GRCh38
rs147321722 Chr.18: 79899723-79899723 on Build GRCh38
rs370780046 Chr.18: 79899746-79899746 on Build GRCh38
rs941833468 Chr.18: 79899718-79899718 on Build GRCh38
rs918785869 Chr.18: 79899876-79899876 on Build GRCh38
rs1054679095 Chr.18: 79899957-79899957 on Build GRCh38
rs748713635 Chr.18: 79899812-79899812 on Build GRCh38
rs3744886 Chr.18: 79899561-79899561 on Build GRCh38
rs763919367 Chr.18: 79899863-79899863 on Build GRCh38
rs765752285 Chr.18: 79899601-79899601 on Build GRCh38
rs1024714761 Chr.18: 79899855-79899855 on Build GRCh38
rs754867366 Chr.18: 79899691-79899691 on Build GRCh38
rs757022948 Chr.18: 79899800-79899800 on Build GRCh38
rs935739487 Chr.18: 79900028-79900028 on Build GRCh38
rs777086927 Chr.18: 79899748-79899748 on Build GRCh38
rs1024203085 Chr.18: 79899969-79899969 on Build GRCh38
rs779090577 Chr.18: 79899811-79899811 on Build GRCh38
rs750816350 Chr.18: 79899574-79899585 on Build GRCh38

View Full Product Details