Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 45094905-45095402 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNS1
Entrez Gene ID
3787
Gene Name
potassium voltage-gated channel modifier subfamily S member 1
Gene Aliases
Kv9.1
UniGene
Hs.117780
Gene Chromosome Location
Chr.20: 45091214-45101112 on Build GRCh38
Transcripts
NM_001322799.1 NM_002251.4 XM_017027846.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5857962 c.1345G>A p.V449I Chr.20: 45095106-45095106 on Build GRCh38
365775 c.1273G>T p.G425W Chr.20: 45095178-45095178 on Build GRCh38
1179636 c.1212C>T p.G404G Chr.20: 45095239-45095239 on Build GRCh38
6691806 c.1420G>A p.V474M Chr.20: 45095031-45095031 on Build GRCh38
1027168 c.1419C>T p.A473A Chr.20: 45095032-45095032 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754548784 Chr.20: 45094987-45094987 on Build GRCh38
rs762163059 Chr.20: 45095046-45095046 on Build GRCh38
rs752341457 Chr.20: 45095054-45095054 on Build GRCh38
rs868030773 Chr.20: 45095321-45095321 on Build GRCh38
rs761292608 Chr.20: 45095298-45095298 on Build GRCh38
rs778788520 Chr.20: 45095065-45095065 on Build GRCh38
rs138845297 Chr.20: 45095240-45095240 on Build GRCh38
rs1020796428 Chr.20: 45095269-45095269 on Build GRCh38
rs547343787 Chr.20: 45095187-45095187 on Build GRCh38
rs774657523 Chr.20: 45095247-45095247 on Build GRCh38
rs569244976 Chr.20: 45094949-45094949 on Build GRCh38
rs770626039 Chr.20: 45094931-45094931 on Build GRCh38
rs755204252 Chr.20: 45095203-45095203 on Build GRCh38
rs777841036 Chr.20: 45095377-45095377 on Build GRCh38
rs768864485 Chr.20: 45095098-45095098 on Build GRCh38
rs775135836 Chr.20: 45095145-45095145 on Build GRCh38
rs142180542 Chr.20: 45094932-45094932 on Build GRCh38
rs144534326 Chr.20: 45095055-45095055 on Build GRCh38
rs146346276 Chr.20: 45095331-45095331 on Build GRCh38
rs756161489 Chr.20: 45095360-45095360 on Build GRCh38
rs760339067 Chr.20: 45095248-45095248 on Build GRCh38
rs958169371 Chr.20: 45095256-45095256 on Build GRCh38
rs141241786 Chr.20: 45094973-45094973 on Build GRCh38
rs752951820 Chr.20: 45095353-45095353 on Build GRCh38
rs776312206 Chr.20: 45095276-45095276 on Build GRCh38
rs753817305 Chr.20: 45095057-45095057 on Build GRCh38
rs1011371794 Chr.20: 45095266-45095266 on Build GRCh38
rs770254063 Chr.20: 45095114-45095114 on Build GRCh38
rs576896465 Chr.20: 45094990-45094990 on Build GRCh38
rs757086443 Chr.20: 45095023-45095023 on Build GRCh38
rs734784 Chr.20: 45094986-45094986 on Build GRCh38
rs756189229 Chr.20: 45095204-45095204 on Build GRCh38
rs768240334 Chr.20: 45095273-45095273 on Build GRCh38
rs766303133 Chr.20: 45095333-45095333 on Build GRCh38
rs748217785 Chr.20: 45095211-45095211 on Build GRCh38
rs7264544 Chr.20: 45094928-45094928 on Build GRCh38
rs760252990 Chr.20: 45095155-45095155 on Build GRCh38
rs752243589 Chr.20: 45095159-45095159 on Build GRCh38
rs765465660 Chr.20: 45094979-45094979 on Build GRCh38
rs551483956 Chr.20: 45094966-45094966 on Build GRCh38
rs753539756 Chr.20: 45095195-45095196 on Build GRCh38
rs763375088 Chr.20: 45095126-45095126 on Build GRCh38
rs777754203 Chr.20: 45095013-45095013 on Build GRCh38
rs557819979 Chr.20: 45094968-45094968 on Build GRCh38
rs930365392 Chr.20: 45094991-45094991 on Build GRCh38
rs573545376 Chr.20: 45095027-45095027 on Build GRCh38
rs751865429 Chr.20: 45095194-45095194 on Build GRCh38
rs754772738 Chr.20: 45095348-45095348 on Build GRCh38
rs748713721 Chr.20: 45095112-45095112 on Build GRCh38
rs34613760 Chr.20: 45095032-45095032 on Build GRCh38
rs1038272919 Chr.20: 45094940-45094940 on Build GRCh38
rs147562294 Chr.20: 45095028-45095028 on Build GRCh38
rs752253447 Chr.20: 45095009-45095009 on Build GRCh38
rs745696064 Chr.20: 45095071-45095071 on Build GRCh38
rs779931108 Chr.20: 45095188-45095188 on Build GRCh38
rs150479277 Chr.20: 45095037-45095037 on Build GRCh38
rs749168228 Chr.20: 45095021-45095021 on Build GRCh38
rs148463252 Chr.20: 45095244-45095244 on Build GRCh38
rs777210810 Chr.20: 45095042-45095042 on Build GRCh38
rs1023321676 Chr.20: 45095113-45095113 on Build GRCh38
rs753668270 Chr.20: 45095364-45095364 on Build GRCh38
rs367800622 Chr.20: 45095047-45095047 on Build GRCh38
rs918530458 Chr.20: 45094961-45094961 on Build GRCh38
rs765845606 Chr.20: 45095158-45095159 on Build GRCh38
rs757099393 Chr.20: 45095058-45095058 on Build GRCh38
rs901413257 Chr.20: 45095108-45095108 on Build GRCh38
rs758232581 Chr.20: 45095085-45095085 on Build GRCh38
rs750395240 Chr.20: 45095329-45095329 on Build GRCh38
rs749825373 Chr.20: 45095379-45095379 on Build GRCh38
rs754396124 Chr.20: 45094972-45094972 on Build GRCh38
rs753235667 Chr.20: 45095168-45095168 on Build GRCh38
rs577538394 Chr.20: 45094969-45094969 on Build GRCh38
rs564831482 Chr.20: 45095109-45095109 on Build GRCh38
rs764750248 Chr.20: 45095311-45095311 on Build GRCh38
rs147790004 Chr.20: 45094971-45094971 on Build GRCh38
rs778503757 Chr.20: 45095025-45095025 on Build GRCh38
rs756765029 Chr.20: 45095173-45095173 on Build GRCh38
rs138344643 Chr.20: 45095053-45095053 on Build GRCh38
rs751450539 Chr.20: 45095347-45095347 on Build GRCh38
rs780485677 Chr.20: 45095090-45095090 on Build GRCh38
rs375436325 Chr.20: 45095212-45095212 on Build GRCh38
rs186549683 Chr.20: 45095095-45095095 on Build GRCh38
rs201155637 Chr.20: 45095011-45095011 on Build GRCh38
rs781335617 Chr.20: 45095351-45095351 on Build GRCh38
rs771287659 Chr.20: 45095245-45095245 on Build GRCh38
rs145118094 Chr.20: 45095171-45095171 on Build GRCh38
rs766715667 Chr.20: 45095138-45095138 on Build GRCh38
rs765123903 Chr.20: 45095179-45095179 on Build GRCh38
rs142045218 Chr.20: 45095330-45095330 on Build GRCh38
rs200995411 Chr.20: 45095117-45095117 on Build GRCh38
rs768846266 Chr.20: 45095041-45095041 on Build GRCh38

View Full Product Details