Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 83842267-83842813 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIF27
Entrez Gene ID
55582
Gene Name
kinesin family member 27
Gene Aliases
-
UniGene
Hs.697514
Gene Chromosome Location
Chr.9: 83832795-83921539 on Build GRCh38
Transcripts
NM_001271927.1 NM_001271928.1 NM_017576.2 XM_011518848.2 XM_011518849.2 XM_011518850.2 XM_011518854.2 XM_011518857.2 XM_011518858.2 XM_017014900.1 XM_017014901.1 XM_017014902.1 XM_017014903.1 XM_017014904.1 XM_017014905.1 XM_017014906.1 XM_017014907.1 XM_017014908.1 XM_017014909.1 XM_017014910.1 XM_017014911.1 XM_017014912.1 XM_017014913.1 XM_017014915.1 XM_017014916.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1736435 c.3689A>G p.E1230G Chr.9: 83842269-83842269 on Build GRCh38
3908530 c.3652C>T p.R1218W Chr.9: 83842306-83842306 on Build GRCh38
3848973 c.3640A>C p.K1214Q Chr.9: 83842318-83842318 on Build GRCh38
4767036 c.3644A>T p.K1215I Chr.9: 83842314-83842314 on Build GRCh38
7175853 c.3663G>A p.K1221K Chr.9: 83842295-83842295 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs532028447 Chr.9: 83842515-83842515 on Build GRCh38
rs769780309 Chr.9: 83842364-83842364 on Build GRCh38
rs1032192649 Chr.9: 83842721-83842721 on Build GRCh38
rs376993923 Chr.9: 83842306-83842306 on Build GRCh38
rs913351494 Chr.9: 83842748-83842748 on Build GRCh38
rs147388642 Chr.9: 83842486-83842486 on Build GRCh38
rs371418764 Chr.9: 83842717-83842717 on Build GRCh38
rs887496628 Chr.9: 83842654-83842654 on Build GRCh38
rs774382074 Chr.9: 83842289-83842289 on Build GRCh38
rs71365387 Chr.9: 83842472-83842473 on Build GRCh38
rs571657802 Chr.9: 83842726-83842726 on Build GRCh38
rs1040594828 Chr.9: 83842596-83842596 on Build GRCh38
rs554621908 Chr.9: 83842365-83842365 on Build GRCh38
rs188940238 Chr.9: 83842623-83842623 on Build GRCh38
rs749396761 Chr.9: 83842343-83842343 on Build GRCh38
rs567718731 Chr.9: 83842702-83842702 on Build GRCh38
rs376395111 Chr.9: 83842409-83842409 on Build GRCh38
rs547160735 Chr.9: 83842712-83842712 on Build GRCh38
rs897504418 Chr.9: 83842695-83842695 on Build GRCh38
rs765929828 Chr.9: 83842383-83842383 on Build GRCh38
rs773830704 Chr.9: 83842394-83842394 on Build GRCh38
rs757343170 Chr.9: 83842344-83842344 on Build GRCh38
rs767037994 Chr.9: 83842412-83842412 on Build GRCh38
rs980624069 Chr.9: 83842504-83842504 on Build GRCh38
rs543755730 Chr.9: 83842627-83842627 on Build GRCh38
rs1005990595 Chr.9: 83842666-83842666 on Build GRCh38
rs186264782 Chr.9: 83842762-83842762 on Build GRCh38
rs568353520 Chr.9: 83842583-83842583 on Build GRCh38
rs902026570 Chr.9: 83842322-83842322 on Build GRCh38
rs957644260 Chr.9: 83842725-83842726 on Build GRCh38
rs762001430 Chr.9: 83842298-83842299 on Build GRCh38
rs542484683 Chr.9: 83842311-83842311 on Build GRCh38
rs969322908 Chr.9: 83842706-83842706 on Build GRCh38
rs772150791 Chr.9: 83842357-83842357 on Build GRCh38
rs765389731 Chr.9: 83842530-83842530 on Build GRCh38
rs1014965882 Chr.9: 83842667-83842667 on Build GRCh38
rs756676470 Chr.9: 83842327-83842327 on Build GRCh38
rs1013292861 Chr.9: 83842696-83842696 on Build GRCh38
rs946046338 Chr.9: 83842751-83842751 on Build GRCh38
rs1054839933 Chr.9: 83842537-83842537 on Build GRCh38
rs762976966 Chr.9: 83842614-83842614 on Build GRCh38
rs1050646773 Chr.9: 83842647-83842647 on Build GRCh38
rs764671105 Chr.9: 83842446-83842446 on Build GRCh38
rs752975744 Chr.9: 83842320-83842320 on Build GRCh38
rs776518666 Chr.9: 83842359-83842359 on Build GRCh38
rs747845313 Chr.9: 83842363-83842363 on Build GRCh38
rs969203248 Chr.9: 83842502-83842502 on Build GRCh38
rs539803116 Chr.9: 83842414-83842414 on Build GRCh38
rs951854745 Chr.9: 83842467-83842467 on Build GRCh38
rs1029155211 Chr.9: 83842461-83842464 on Build GRCh38
rs563864759 Chr.9: 83842574-83842574 on Build GRCh38
rs1014606684 Chr.9: 83842475-83842476 on Build GRCh38
rs927357182 Chr.9: 83842523-83842523 on Build GRCh38
rs746153634 Chr.9: 83842355-83842355 on Build GRCh38
rs549324795 Chr.9: 83842699-83842699 on Build GRCh38
rs917724038 Chr.9: 83842496-83842496 on Build GRCh38
rs996284430 Chr.9: 83842628-83842628 on Build GRCh38
rs1006290413 Chr.9: 83842470-83842472 on Build GRCh38
rs762873465 Chr.9: 83842374-83842374 on Build GRCh38
rs961934400 Chr.9: 83842476-83842476 on Build GRCh38
rs754467403 Chr.9: 83842448-83842448 on Build GRCh38
rs867211348 Chr.9: 83842764-83842764 on Build GRCh38
rs779132941 Chr.9: 83842352-83842352 on Build GRCh38
rs200646527 Chr.9: 83842291-83842291 on Build GRCh38
rs1024895440 Chr.9: 83842705-83842705 on Build GRCh38
rs184624623 Chr.9: 83842559-83842559 on Build GRCh38
rs756380011 Chr.9: 83842428-83842428 on Build GRCh38
rs778094907 Chr.9: 83842338-83842338 on Build GRCh38
rs767862319 Chr.9: 83842319-83842319 on Build GRCh38
rs398011242 Chr.9: 83842480-83842481 on Build GRCh38
rs539068167 Chr.9: 83842756-83842756 on Build GRCh38
rs181382813 Chr.9: 83842709-83842709 on Build GRCh38
rs770767937 Chr.9: 83842552-83842552 on Build GRCh38
rs181774128 Chr.9: 83842555-83842555 on Build GRCh38

View Full Product Details