Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 44934161-44934561 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KLHL28
Entrez Gene ID
54813
Gene Name
kelch like family member 28
Gene Aliases
BTBD5
UniGene
Hs.550906
Gene Chromosome Location
Chr.14: 44924310-44961978 on Build GRCh38
Transcripts
NM_001308112.1 NM_017658.4 XM_005267770.3 XM_011536847.2 XM_011536849.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2197213 c.1268G>A p.R423K Chr.14: 44934190-44934190 on Build GRCh38
6696666 c.1018G>A p.A340T Chr.14: 44934440-44934440 on Build GRCh38
5480101 c.956T>A p.I319N Chr.14: 44934502-44934502 on Build GRCh38
5631141 c.1001A>G p.Y334C Chr.14: 44934457-44934457 on Build GRCh38
698590 c.1037G>C p.G346A Chr.14: 44934421-44934421 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs142116819 Chr.14: 44934476-44934476 on Build GRCh38
rs748603133 Chr.14: 44934185-44934195 on Build GRCh38
rs925408963 Chr.14: 44934201-44934201 on Build GRCh38
rs750402666 Chr.14: 44934267-44934267 on Build GRCh38
rs373999644 Chr.14: 44934198-44934198 on Build GRCh38
rs758862979 Chr.14: 44934192-44934192 on Build GRCh38
rs780054236 Chr.14: 44934477-44934477 on Build GRCh38
rs761893347 Chr.14: 44934260-44934260 on Build GRCh38
rs144860260 Chr.14: 44934419-44934419 on Build GRCh38
rs771160780 Chr.14: 44934533-44934533 on Build GRCh38
rs772544177 Chr.14: 44934235-44934235 on Build GRCh38
rs747721007 Chr.14: 44934226-44934226 on Build GRCh38
rs763226531 Chr.14: 44934414-44934414 on Build GRCh38
rs751647943 Chr.14: 44934469-44934469 on Build GRCh38
rs775406625 Chr.14: 44934258-44934258 on Build GRCh38
rs745780927 Chr.14: 44934364-44934364 on Build GRCh38
rs772157986 Chr.14: 44934371-44934371 on Build GRCh38
rs35728857 Chr.14: 44934413-44934413 on Build GRCh38
rs546980465 Chr.14: 44934427-44934427 on Build GRCh38
rs139388902 Chr.14: 44934213-44934213 on Build GRCh38
rs866679355 Chr.14: 44934494-44934494 on Build GRCh38
rs756562080 Chr.14: 44934267-44934267 on Build GRCh38
rs377577087 Chr.14: 44934230-44934230 on Build GRCh38
rs749009458 Chr.14: 44934323-44934323 on Build GRCh38
rs368953046 Chr.14: 44934270-44934270 on Build GRCh38
rs765318218 Chr.14: 44934398-44934398 on Build GRCh38
rs756492113 Chr.14: 44934433-44934433 on Build GRCh38
rs985300208 Chr.14: 44934223-44934223 on Build GRCh38
rs748838457 Chr.14: 44934249-44934249 on Build GRCh38
rs751684670 Chr.14: 44934420-44934420 on Build GRCh38
rs199867476 Chr.14: 44934312-44934312 on Build GRCh38
rs775108364 Chr.14: 44934374-44934374 on Build GRCh38
rs929674858 Chr.14: 44934351-44934351 on Build GRCh38
rs755517468 Chr.14: 44934303-44934303 on Build GRCh38
rs778068577 Chr.14: 44934442-44934442 on Build GRCh38
rs921732070 Chr.14: 44934195-44934195 on Build GRCh38
rs774419924 Chr.14: 44934253-44934253 on Build GRCh38
rs753512662 Chr.14: 44934451-44934451 on Build GRCh38
rs376857549 Chr.14: 44934529-44934529 on Build GRCh38
rs745403290 Chr.14: 44934466-44934466 on Build GRCh38
rs918478697 Chr.14: 44934287-44934287 on Build GRCh38
rs776448491 Chr.14: 44934507-44934507 on Build GRCh38
rs759616103 Chr.14: 44934255-44934255 on Build GRCh38
rs755659647 Chr.14: 44934317-44934317 on Build GRCh38
rs923880408 Chr.14: 44934228-44934228 on Build GRCh38
rs974862447 Chr.14: 44934194-44934194 on Build GRCh38
rs781600798 Chr.14: 44934225-44934225 on Build GRCh38
rs147583402 Chr.14: 44934459-44934459 on Build GRCh38
rs35352691 Chr.14: 44934256-44934256 on Build GRCh38
rs568042374 Chr.14: 44934393-44934393 on Build GRCh38
rs768096920 Chr.14: 44934422-44934422 on Build GRCh38
rs117295933 Chr.14: 44934496-44934496 on Build GRCh38
rs150835238 Chr.14: 44934199-44934199 on Build GRCh38
rs774045666 Chr.14: 44934363-44934363 on Build GRCh38
rs544393993 Chr.14: 44934515-44934515 on Build GRCh38
rs752054910 Chr.14: 44934292-44934292 on Build GRCh38
rs765527097 Chr.14: 44934264-44934264 on Build GRCh38
rs374089650 Chr.14: 44934359-44934359 on Build GRCh38
rs769197019 Chr.14: 44934290-44934290 on Build GRCh38
rs778519316 Chr.14: 44934463-44934463 on Build GRCh38
rs150062543 Chr.14: 44934251-44934251 on Build GRCh38

View Full Product Details