Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 205599019-205599516 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELK4
Entrez Gene ID
2005
Gene Name
ELK4, ETS transcription factor
Gene Aliases
SAP1
UniGene
Hs.497520
Gene Chromosome Location
Chr.1: 205597774-205634077 on Build GRCh38
Transcripts
NM_181644.4 XM_011509229.2

Gene Symbol
MFSD4A
Entrez Gene ID
148808
Gene Name
major facilitator superfamily domain containing 4A
Gene Aliases
MFSD4, UNQ3064
UniGene
Hs.567714
Gene Chromosome Location
Chr.1: 205568984-205602918 on Build GRCh38
Transcripts
NM_181644.4 XM_011509229.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4928544 c.1392T>C p.Y464Y Chr.1: 205599154-205599154 on Build GRCh38
425118 c.1372T>C p.F458L Chr.1: 205599134-205599134 on Build GRCh38
7176402 c.1446C>T p.I482I Chr.1: 205599208-205599208 on Build GRCh38
4701953 c.1403T>C p.V468A Chr.1: 205599165-205599165 on Build GRCh38
6712018 c.1410C>T p.G470G Chr.1: 205599172-205599172 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368097405 Chr.1: 205599276-205599276 on Build GRCh38
rs565644305 Chr.1: 205599155-205599155 on Build GRCh38
rs144932899 Chr.1: 205599225-205599225 on Build GRCh38
rs774284140 Chr.1: 205599245-205599245 on Build GRCh38
rs981781448 Chr.1: 205599429-205599429 on Build GRCh38
rs755895547 Chr.1: 205599183-205599183 on Build GRCh38
rs376885748 Chr.1: 205599099-205599099 on Build GRCh38
rs138733261 Chr.1: 205599302-205599302 on Build GRCh38
rs1009237194 Chr.1: 205599134-205599134 on Build GRCh38
rs763292031 Chr.1: 205599170-205599170 on Build GRCh38
rs866347043 Chr.1: 205599240-205599240 on Build GRCh38
rs932470295 Chr.1: 205599092-205599092 on Build GRCh38
rs749088167 Chr.1: 205599140-205599140 on Build GRCh38
rs770842139 Chr.1: 205599105-205599108 on Build GRCh38
rs769972889 Chr.1: 205599159-205599159 on Build GRCh38
rs574735551 Chr.1: 205599430-205599430 on Build GRCh38
rs775692447 Chr.1: 205599234-205599234 on Build GRCh38
rs749470752 Chr.1: 205599235-205599235 on Build GRCh38
rs199748975 Chr.1: 205599071-205599071 on Build GRCh38
rs2282449 Chr.1: 205599085-205599085 on Build GRCh38
rs541368655 Chr.1: 205599341-205599341 on Build GRCh38
rs2282452 Chr.1: 205599314-205599314 on Build GRCh38
rs1028931492 Chr.1: 205599308-205599308 on Build GRCh38
rs930462249 Chr.1: 205599444-205599444 on Build GRCh38
rs745945183 Chr.1: 205599232-205599232 on Build GRCh38
rs754827345 Chr.1: 205599213-205599213 on Build GRCh38
rs371101261 Chr.1: 205599148-205599148 on Build GRCh38
rs1000482396 Chr.1: 205599393-205599393 on Build GRCh38
rs963355555 Chr.1: 205599250-205599250 on Build GRCh38
rs555195408 Chr.1: 205599403-205599403 on Build GRCh38
rs774556544 Chr.1: 205599252-205599252 on Build GRCh38
rs1005402084 Chr.1: 205599482-205599482 on Build GRCh38
rs765923298 Chr.1: 205599297-205599297 on Build GRCh38
rs1041094627 Chr.1: 205599477-205599477 on Build GRCh38
rs370691567 Chr.1: 205599192-205599192 on Build GRCh38
rs1048515010 Chr.1: 205599459-205599459 on Build GRCh38
rs544589364 Chr.1: 205599472-205599472 on Build GRCh38
rs549465212 Chr.1: 205599086-205599086 on Build GRCh38
rs568265775 Chr.1: 205599226-205599226 on Build GRCh38
rs570149192 Chr.1: 205599306-205599306 on Build GRCh38
rs150621373 Chr.1: 205599190-205599190 on Build GRCh38
rs937936331 Chr.1: 205599196-205599196 on Build GRCh38
rs755803062 Chr.1: 205599114-205599114 on Build GRCh38
rs775186010 Chr.1: 205599078-205599078 on Build GRCh38
rs912332561 Chr.1: 205599313-205599313 on Build GRCh38
rs745342397 Chr.1: 205599291-205599292 on Build GRCh38
rs760272468 Chr.1: 205599296-205599296 on Build GRCh38
rs769088044 Chr.1: 205599246-205599246 on Build GRCh38
rs778925681 Chr.1: 205599147-205599147 on Build GRCh38
rs41308411 Chr.1: 205599088-205599088 on Build GRCh38
rs937950000 Chr.1: 205599434-205599434 on Build GRCh38
rs989226911 Chr.1: 205599163-205599163 on Build GRCh38
rs778536592 Chr.1: 205599218-205599218 on Build GRCh38
rs2282451 Chr.1: 205599310-205599310 on Build GRCh38
rs770061092 Chr.1: 205599233-205599233 on Build GRCh38
rs945045367 Chr.1: 205599415-205599415 on Build GRCh38
rs2282450 Chr.1: 205599262-205599262 on Build GRCh38
rs539062231 Chr.1: 205599309-205599309 on Build GRCh38
rs561337844 Chr.1: 205599083-205599083 on Build GRCh38
rs71582041 Chr.1: 205599310-205599314 on Build GRCh38
rs776528325 Chr.1: 205599156-205599156 on Build GRCh38
rs148530507 Chr.1: 205599173-205599173 on Build GRCh38
rs780031193 Chr.1: 205599139-205599139 on Build GRCh38
rs759803572 Chr.1: 205599157-205599157 on Build GRCh38
rs748143265 Chr.1: 205599221-205599221 on Build GRCh38
rs932509775 Chr.1: 205599300-205599300 on Build GRCh38
rs1050898652 Chr.1: 205599095-205599095 on Build GRCh38
rs543597531 Chr.1: 205599440-205599440 on Build GRCh38
rs749865508 Chr.1: 205599182-205599182 on Build GRCh38
rs975676170 Chr.1: 205599305-205599305 on Build GRCh38
rs778871499 Chr.1: 205599243-205599244 on Build GRCh38
rs1042107735 Chr.1: 205599328-205599328 on Build GRCh38
rs766435629 Chr.1: 205599303-205599303 on Build GRCh38
rs965578399 Chr.1: 205599112-205599112 on Build GRCh38
rs887231359 Chr.1: 205599471-205599471 on Build GRCh38
rs34506739 Chr.1: 205599081-205599082 on Build GRCh38
rs1021015296 Chr.1: 205599145-205599145 on Build GRCh38
rs764297998 Chr.1: 205599172-205599172 on Build GRCh38
rs1055027927 Chr.1: 205599439-205599439 on Build GRCh38
rs146674823 Chr.1: 205599146-205599146 on Build GRCh38
rs775902709 Chr.1: 205599166-205599166 on Build GRCh38
rs767901443 Chr.1: 205599274-205599274 on Build GRCh38

View Full Product Details