Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 40134384-40134889 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MPLKIP
Entrez Gene ID
136647
Gene Name
M-phase specific PLK1 interacting protein
Gene Aliases
ABHS, C7orf11, ORF20, TTD4
UniGene
Hs.654989
Gene Chromosome Location
Chr.7: 40132743-40134652 on Build GRCh38
Transcripts
NM_138701.3 XM_006715775.3

Gene Symbol
SUGCT
Entrez Gene ID
79783
Gene Name
succinyl-CoA:glutarate-CoA transferase
Gene Aliases
C7orf10, DERP13, GA3, ORF19
UniGene
Hs.586313
Gene Chromosome Location
Chr.7: 40134887-40860767 on Build GRCh38
Transcripts
NM_138701.3 XM_006715775.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4164328 c.146A>C p.H49P Chr.7: 40134422-40134422 on Build GRCh38
4703030 c.96C>T p.G32G Chr.7: 40134472-40134472 on Build GRCh38
6561421 c.173C>G p.S58C Chr.7: 40134395-40134395 on Build GRCh38
32235 c.86G>A p.G29E Chr.7: 40134482-40134482 on Build GRCh38
4595722 c.4C>G p.Q2E Chr.7: 40134564-40134564 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs866153347 Chr.7: 40134721-40134721 on Build GRCh38
rs147383072 Chr.7: 40134722-40134722 on Build GRCh38
rs772187994 Chr.7: 40134688-40134688 on Build GRCh38
rs980300429 Chr.7: 40134822-40134822 on Build GRCh38
rs958307323 Chr.7: 40134805-40134805 on Build GRCh38
rs989403973 Chr.7: 40134828-40134828 on Build GRCh38
rs113464225 Chr.7: 40134403-40134403 on Build GRCh38
rs913849575 Chr.7: 40134833-40134833 on Build GRCh38
rs1044188102 Chr.7: 40134429-40134429 on Build GRCh38
rs932273833 Chr.7: 40134725-40134725 on Build GRCh38
rs746398211 Chr.7: 40134559-40134559 on Build GRCh38
rs750387406 Chr.7: 40134505-40134505 on Build GRCh38
rs764263319 Chr.7: 40134488-40134488 on Build GRCh38
rs573905757 Chr.7: 40134659-40134659 on Build GRCh38
rs770161710 Chr.7: 40134564-40134564 on Build GRCh38
rs761977352 Chr.7: 40134611-40134611 on Build GRCh38
rs891834226 Chr.7: 40134704-40134704 on Build GRCh38
rs747470385 Chr.7: 40134475-40134476 on Build GRCh38
rs927023495 Chr.7: 40134430-40134430 on Build GRCh38
rs771561256 Chr.7: 40134468-40134468 on Build GRCh38
rs587776531 Chr.7: 40134430-40134431 on Build GRCh38
rs763275349 Chr.7: 40134567-40134567 on Build GRCh38
rs980652263 Chr.7: 40134442-40134442 on Build GRCh38
rs1040309554 Chr.7: 40134458-40134458 on Build GRCh38
rs565833937 Chr.7: 40134482-40134482 on Build GRCh38
rs1040814804 Chr.7: 40134649-40134649 on Build GRCh38
rs936467344 Chr.7: 40134452-40134452 on Build GRCh38
rs913615620 Chr.7: 40134461-40134461 on Build GRCh38
rs779917455 Chr.7: 40134518-40134518 on Build GRCh38
rs768140543 Chr.7: 40134455-40134455 on Build GRCh38
rs1024981046 Chr.7: 40134761-40134761 on Build GRCh38
rs900833107 Chr.7: 40134501-40134501 on Build GRCh38
rs1050655777 Chr.7: 40134630-40134630 on Build GRCh38
rs1048729514 Chr.7: 40134443-40134443 on Build GRCh38
rs897990160 Chr.7: 40134758-40134758 on Build GRCh38
rs889886256 Chr.7: 40134635-40134635 on Build GRCh38
rs1024102684 Chr.7: 40134502-40134502 on Build GRCh38
rs966495844 Chr.7: 40134861-40134861 on Build GRCh38
rs993627245 Chr.7: 40134760-40134760 on Build GRCh38
rs537469520 Chr.7: 40134546-40134546 on Build GRCh38
rs764948501 Chr.7: 40134599-40134599 on Build GRCh38
rs913720684 Chr.7: 40134604-40134604 on Build GRCh38
rs747797874 Chr.7: 40134544-40134544 on Build GRCh38
rs754784607 Chr.7: 40134535-40134535 on Build GRCh38
rs1000198489 Chr.7: 40134673-40134673 on Build GRCh38
rs371245257 Chr.7: 40134484-40134484 on Build GRCh38
rs766280337 Chr.7: 40134593-40134593 on Build GRCh38
rs775211403 Chr.7: 40134471-40134471 on Build GRCh38
rs143182074 Chr.7: 40134685-40134685 on Build GRCh38
rs950261801 Chr.7: 40134427-40134427 on Build GRCh38
rs773220405 Chr.7: 40134740-40134740 on Build GRCh38
rs751808736 Chr.7: 40134489-40134489 on Build GRCh38
rs945238445 Chr.7: 40134627-40134627 on Build GRCh38
rs534831071 Chr.7: 40134498-40134498 on Build GRCh38
rs781356195 Chr.7: 40134496-40134497 on Build GRCh38
rs1034889628 Chr.7: 40134840-40134840 on Build GRCh38
rs917376489 Chr.7: 40134421-40134421 on Build GRCh38
rs746094400 Chr.7: 40134542-40134542 on Build GRCh38
rs949420400 Chr.7: 40134414-40134414 on Build GRCh38
rs767715729 Chr.7: 40134584-40134584 on Build GRCh38
rs536445664 Chr.7: 40134684-40134684 on Build GRCh38
rs750477350 Chr.7: 40134585-40134585 on Build GRCh38
rs774530760 Chr.7: 40134596-40134596 on Build GRCh38
rs545074042 Chr.7: 40134869-40134869 on Build GRCh38
rs780610824 Chr.7: 40134405-40134405 on Build GRCh38
rs1049436728 Chr.7: 40134734-40134734 on Build GRCh38
rs760689021 Chr.7: 40134588-40134588 on Build GRCh38
rs917899841 Chr.7: 40134404-40134404 on Build GRCh38
rs777297067 Chr.7: 40134613-40134613 on Build GRCh38
rs778600723 Chr.7: 40134459-40134459 on Build GRCh38
rs201269559 Chr.7: 40134519-40134519 on Build GRCh38
rs901738307 Chr.7: 40134601-40134601 on Build GRCh38
rs761185509 Chr.7: 40134777-40134777 on Build GRCh38
rs878854339 Chr.7: 40134416-40134420 on Build GRCh38
rs556959601 Chr.7: 40134640-40134640 on Build GRCh38
rs1040843591 Chr.7: 40134491-40134491 on Build GRCh38
rs993530962 Chr.7: 40134557-40134557 on Build GRCh38
rs757445367 Chr.7: 40134488-40134488 on Build GRCh38
rs762040983 Chr.7: 40134490-40134490 on Build GRCh38
rs1033664311 Chr.7: 40134791-40134791 on Build GRCh38
rs1016450721 Chr.7: 40134537-40134537 on Build GRCh38
rs1006773863 Chr.7: 40134527-40134527 on Build GRCh38
rs747709922 Chr.7: 40134464-40134464 on Build GRCh38
rs113147043 Chr.7: 40134499-40134499 on Build GRCh38
rs756038629 Chr.7: 40134438-40134438 on Build GRCh38
rs762057501 Chr.7: 40134583-40134583 on Build GRCh38
rs768815647 Chr.7: 40134536-40134536 on Build GRCh38
rs745817430 Chr.7: 40134433-40134433 on Build GRCh38
rs930353802 Chr.7: 40134576-40134576 on Build GRCh38
rs779644594 Chr.7: 40134444-40134444 on Build GRCh38
rs777209092 Chr.7: 40134548-40134548 on Build GRCh38
rs769783319 Chr.7: 40134448-40134448 on Build GRCh38
rs756055084 Chr.7: 40134510-40134510 on Build GRCh38
rs775669059 Chr.7: 40134476-40134476 on Build GRCh38
rs776034351 Chr.7: 40134566-40134566 on Build GRCh38
rs753805134 Chr.7: 40134594-40134594 on Build GRCh38
rs746561954 Chr.7: 40134617-40134617 on Build GRCh38
rs778474274 Chr.7: 40134403-40134429 on Build GRCh38
rs1002135081 Chr.7: 40134788-40134788 on Build GRCh38
rs752361823 Chr.7: 40134606-40134606 on Build GRCh38
rs764264973 Chr.7: 40134431-40134431 on Build GRCh38
rs750370923 Chr.7: 40134436-40134436 on Build GRCh38
rs1038389152 Chr.7: 40134745-40134745 on Build GRCh38
rs769051260 Chr.7: 40134573-40134573 on Build GRCh38
rs774779671 Chr.7: 40134575-40134575 on Build GRCh38
rs889343631 Chr.7: 40134737-40134737 on Build GRCh38
rs751554073 Chr.7: 40134432-40134432 on Build GRCh38
rs79477505 Chr.7: 40134521-40134521 on Build GRCh38
rs1025052999 Chr.7: 40134561-40134561 on Build GRCh38

View Full Product Details