Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 30309811-30310310 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MTPAP
Entrez Gene ID
55149
Gene Name
mitochondrial poly(A) polymerase
Gene Aliases
PAPD1, SPAX4
UniGene
Hs.173946
Gene Chromosome Location
Chr.10: 30309801-30349338 on Build GRCh38
Transcripts
NM_018109.3

SNP Detail

SNP ID Chromosome Location
rs944428331 Chr.10: 30309907-30309907 on Build GRCh38
rs951749217 Chr.10: 30309886-30309886 on Build GRCh38
rs186191258 Chr.10: 30309890-30309890 on Build GRCh38
rs1011159512 Chr.10: 30310010-30310010 on Build GRCh38
rs563145479 Chr.10: 30309873-30309873 on Build GRCh38
rs572855461 Chr.10: 30310265-30310265 on Build GRCh38
rs377058770 Chr.10: 30310064-30310064 on Build GRCh38
rs996071483 Chr.10: 30310032-30310032 on Build GRCh38
rs1038207252 Chr.10: 30309911-30309911 on Build GRCh38
rs932504295 Chr.10: 30310218-30310218 on Build GRCh38
rs540009676 Chr.10: 30309974-30309974 on Build GRCh38
rs908202574 Chr.10: 30310167-30310167 on Build GRCh38
rs1004400379 Chr.10: 30310270-30310270 on Build GRCh38
rs983076281 Chr.10: 30309894-30309894 on Build GRCh38
rs12269422 Chr.10: 30310043-30310043 on Build GRCh38
rs545948200 Chr.10: 30310262-30310262 on Build GRCh38
rs551294093 Chr.10: 30309946-30309946 on Build GRCh38
rs896584936 Chr.10: 30310005-30310005 on Build GRCh38
rs965787723 Chr.10: 30310104-30310104 on Build GRCh38
rs141224668 Chr.10: 30309970-30309970 on Build GRCh38
rs905390576 Chr.10: 30310020-30310020 on Build GRCh38
rs1013647532 Chr.10: 30310006-30310006 on Build GRCh38
rs556815049 Chr.10: 30310201-30310201 on Build GRCh38
rs181660163 Chr.10: 30309884-30309884 on Build GRCh38
rs552207993 Chr.10: 30310038-30310038 on Build GRCh38
rs941258037 Chr.10: 30309897-30309897 on Build GRCh38
rs555690133 Chr.10: 30310112-30310112 on Build GRCh38
rs1043680462 Chr.10: 30309973-30309973 on Build GRCh38
rs1031405787 Chr.10: 30309997-30309997 on Build GRCh38
rs754398934 Chr.10: 30309885-30309885 on Build GRCh38
rs1050472841 Chr.10: 30309971-30309971 on Build GRCh38
rs779815243 Chr.10: 30310240-30310240 on Build GRCh38
rs575130749 Chr.10: 30310211-30310211 on Build GRCh38
rs184053230 Chr.10: 30310184-30310184 on Build GRCh38
rs935319679 Chr.10: 30309978-30309978 on Build GRCh38
rs757645927 Chr.10: 30309899-30309899 on Build GRCh38
rs995692307 Chr.10: 30310232-30310232 on Build GRCh38
rs908485765 Chr.10: 30309888-30309888 on Build GRCh38
rs979633712 Chr.10: 30310124-30310124 on Build GRCh38
rs935205735 Chr.10: 30310131-30310131 on Build GRCh38
rs565930333 Chr.10: 30310127-30310127 on Build GRCh38
rs970061024 Chr.10: 30309857-30309857 on Build GRCh38
rs190959333 Chr.10: 30309963-30309963 on Build GRCh38
rs1050856336 Chr.10: 30310229-30310229 on Build GRCh38
rs1052401575 Chr.10: 30309983-30309983 on Build GRCh38
rs534612690 Chr.10: 30309916-30309917 on Build GRCh38
rs1008912587 Chr.10: 30309991-30309991 on Build GRCh38
rs976335769 Chr.10: 30309912-30309912 on Build GRCh38
rs905273600 Chr.10: 30310220-30310220 on Build GRCh38
rs768468336 Chr.10: 30309992-30309992 on Build GRCh38
rs573686100 Chr.10: 30310114-30310114 on Build GRCh38
rs983089233 Chr.10: 30310083-30310083 on Build GRCh38
rs982493825 Chr.10: 30310155-30310155 on Build GRCh38
rs564029933 Chr.10: 30310264-30310264 on Build GRCh38
rs758120075 Chr.10: 30310181-30310181 on Build GRCh38
rs1014516791 Chr.10: 30310089-30310089 on Build GRCh38
rs11007975 Chr.10: 30310013-30310013 on Build GRCh38
rs1052453793 Chr.10: 30310151-30310151 on Build GRCh38
rs746480562 Chr.10: 30310261-30310261 on Build GRCh38
rs931871784 Chr.10: 30310122-30310122 on Build GRCh38
rs931793562 Chr.10: 30309951-30309951 on Build GRCh38

View Full Product Details