Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 23702629-23703133 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NKX2-6
Entrez Gene ID
137814
Gene Name
NK2 homeobox 6
Gene Aliases
CSX2, CTHM, NKX2F, NKX4-2
UniGene
Hs.532654
Gene Chromosome Location
Chr.8: 23702451-23706598 on Build GRCh38
Transcripts
NM_001136271.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4796560 c.599G>A p.G200D Chr.8: 23702758-23702758 on Build GRCh38
4835241 c.712G>C p.V238L Chr.8: 23702645-23702645 on Build GRCh38
5476986 c.547C>A p.R183R Chr.8: 23702810-23702810 on Build GRCh38
4410261 c.710C>T p.A237V Chr.8: 23702647-23702647 on Build GRCh38
7189446 c.665G>A p.G222D Chr.8: 23702692-23702692 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766285856 Chr.8: 23702896-23702896 on Build GRCh38
rs1011658784 Chr.8: 23702876-23702876 on Build GRCh38
rs761239489 Chr.8: 23702873-23702873 on Build GRCh38
rs1013079722 Chr.8: 23702952-23702952 on Build GRCh38
rs534202593 Chr.8: 23702893-23702893 on Build GRCh38
rs368352157 Chr.8: 23702674-23702674 on Build GRCh38
rs61743032 Chr.8: 23702998-23702998 on Build GRCh38
rs768775204 Chr.8: 23703081-23703081 on Build GRCh38
rs941121273 Chr.8: 23702732-23702732 on Build GRCh38
rs761490097 Chr.8: 23702972-23702972 on Build GRCh38
rs750361656 Chr.8: 23702889-23702889 on Build GRCh38
rs571019951 Chr.8: 23702955-23702955 on Build GRCh38
rs61740725 Chr.8: 23703059-23703059 on Build GRCh38
rs368111443 Chr.8: 23702764-23702764 on Build GRCh38
rs568127693 Chr.8: 23702989-23702989 on Build GRCh38
rs758135138 Chr.8: 23702724-23702724 on Build GRCh38
rs936421503 Chr.8: 23702659-23702659 on Build GRCh38
rs771605785 Chr.8: 23702948-23702948 on Build GRCh38
rs535566228 Chr.8: 23702992-23702992 on Build GRCh38
rs973784005 Chr.8: 23702967-23702967 on Build GRCh38
rs753434940 Chr.8: 23702688-23702688 on Build GRCh38
rs763814874 Chr.8: 23702679-23702679 on Build GRCh38
rs931354964 Chr.8: 23703017-23703017 on Build GRCh38
rs747258353 Chr.8: 23702726-23702726 on Build GRCh38
rs1004022424 Chr.8: 23702851-23702851 on Build GRCh38
rs762342897 Chr.8: 23702995-23702995 on Build GRCh38
rs913645847 Chr.8: 23702684-23702684 on Build GRCh38
rs751485832 Chr.8: 23703016-23703016 on Build GRCh38
rs538006000 Chr.8: 23702962-23702962 on Build GRCh38
rs765142500 Chr.8: 23702692-23702692 on Build GRCh38
rs759341554 Chr.8: 23703066-23703066 on Build GRCh38
rs746288922 Chr.8: 23702956-23702956 on Build GRCh38
rs929665250 Chr.8: 23702820-23702820 on Build GRCh38
rs773595026 Chr.8: 23702793-23702793 on Build GRCh38
rs901046036 Chr.8: 23703075-23703075 on Build GRCh38
rs143039156 Chr.8: 23702971-23702971 on Build GRCh38
rs776406953 Chr.8: 23702856-23702856 on Build GRCh38
rs771222551 Chr.8: 23702835-23702835 on Build GRCh38
rs756783244 Chr.8: 23702691-23702691 on Build GRCh38
rs1041889681 Chr.8: 23702720-23702720 on Build GRCh38
rs772831506 Chr.8: 23702993-23702993 on Build GRCh38
rs759945353 Chr.8: 23702842-23702842 on Build GRCh38
rs887208406 Chr.8: 23703043-23703043 on Build GRCh38
rs751396105 Chr.8: 23702902-23702902 on Build GRCh38
rs552633150 Chr.8: 23702913-23702913 on Build GRCh38
rs894435299 Chr.8: 23702802-23702802 on Build GRCh38
rs372045387 Chr.8: 23702836-23702836 on Build GRCh38
rs781453291 Chr.8: 23702911-23702911 on Build GRCh38
rs267606914 Chr.8: 23702906-23702906 on Build GRCh38
rs986507418 Chr.8: 23702994-23702994 on Build GRCh38
rs913676984 Chr.8: 23702942-23702942 on Build GRCh38
rs777859360 Chr.8: 23702917-23702917 on Build GRCh38
rs946224551 Chr.8: 23702708-23702708 on Build GRCh38
rs371546349 Chr.8: 23702743-23702743 on Build GRCh38
rs1048360478 Chr.8: 23703033-23703033 on Build GRCh38
rs548152920 Chr.8: 23702785-23702785 on Build GRCh38
rs780012501 Chr.8: 23703008-23703009 on Build GRCh38
rs918352540 Chr.8: 23702968-23702968 on Build GRCh38
rs749305533 Chr.8: 23702938-23702938 on Build GRCh38
rs587777422 Chr.8: 23702903-23702904 on Build GRCh38
rs916318048 Chr.8: 23702713-23702713 on Build GRCh38
rs754763080 Chr.8: 23702905-23702905 on Build GRCh38
rs764740389 Chr.8: 23702877-23702877 on Build GRCh38
rs991694173 Chr.8: 23702687-23702687 on Build GRCh38
rs1053580780 Chr.8: 23702681-23702681 on Build GRCh38
rs1039509893 Chr.8: 23703026-23703026 on Build GRCh38
rs774990509 Chr.8: 23702950-23702950 on Build GRCh38
rs190889748 Chr.8: 23702804-23702804 on Build GRCh38
rs374977404 Chr.8: 23702912-23702912 on Build GRCh38
rs530151840 Chr.8: 23702700-23702700 on Build GRCh38
rs557722998 Chr.8: 23703078-23703078 on Build GRCh38
rs546077329 Chr.8: 23703067-23703067 on Build GRCh38
rs879460402 Chr.8: 23702725-23702725 on Build GRCh38
rs1057165488 Chr.8: 23703088-23703088 on Build GRCh38
rs1019379812 Chr.8: 23702852-23702852 on Build GRCh38
rs958175093 Chr.8: 23703111-23703111 on Build GRCh38
rs977597161 Chr.8: 23702961-23702961 on Build GRCh38
rs757292066 Chr.8: 23702901-23702902 on Build GRCh38
rs749760844 Chr.8: 23702795-23702795 on Build GRCh38
rs374392076 Chr.8: 23702849-23702849 on Build GRCh38
rs760354322 Chr.8: 23702678-23702678 on Build GRCh38
rs1028268369 Chr.8: 23703098-23703098 on Build GRCh38
rs1039541183 Chr.8: 23702855-23702855 on Build GRCh38

View Full Product Details