Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 38820686-38821231 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NPTXR
Entrez Gene ID
23467
Gene Name
neuronal pentraxin receptor
Gene Aliases
NPR
UniGene
Hs.91622
Gene Chromosome Location
Chr.22: 38818451-38844012 on Build GRCh38
Transcripts
NM_014293.3

SNP Detail

SNP ID Chromosome Location
rs1010697915 Chr.22: 38820712-38820712 on Build GRCh38
rs893718353 Chr.22: 38821021-38821021 on Build GRCh38
rs950064076 Chr.22: 38820762-38820762 on Build GRCh38
rs566092285 Chr.22: 38820874-38820874 on Build GRCh38
rs950142313 Chr.22: 38821000-38821000 on Build GRCh38
rs905880238 Chr.22: 38820807-38820807 on Build GRCh38
rs1038676455 Chr.22: 38820813-38820813 on Build GRCh38
rs6001247 Chr.22: 38821111-38821111 on Build GRCh38
rs946574467 Chr.22: 38821023-38821023 on Build GRCh38
rs759954060 Chr.22: 38821114-38821114 on Build GRCh38
rs532304312 Chr.22: 38820714-38820714 on Build GRCh38
rs550252649 Chr.22: 38820963-38820963 on Build GRCh38
rs146814919 Chr.22: 38821019-38821019 on Build GRCh38
rs747857176 Chr.22: 38821011-38821011 on Build GRCh38
rs905979541 Chr.22: 38821066-38821066 on Build GRCh38
rs556455661 Chr.22: 38821030-38821030 on Build GRCh38
rs937329148 Chr.22: 38821016-38821016 on Build GRCh38
rs552223358 Chr.22: 38820933-38820933 on Build GRCh38
rs1007970470 Chr.22: 38821073-38821073 on Build GRCh38
rs879146111 Chr.22: 38820922-38820922 on Build GRCh38
rs988039465 Chr.22: 38821189-38821189 on Build GRCh38
rs770323499 Chr.22: 38821103-38821103 on Build GRCh38
rs529936019 Chr.22: 38820834-38820834 on Build GRCh38
rs913281859 Chr.22: 38820964-38820964 on Build GRCh38
rs746901627 Chr.22: 38820948-38820948 on Build GRCh38
rs780066547 Chr.22: 38820886-38820886 on Build GRCh38
rs1025371420 Chr.22: 38820862-38820862 on Build GRCh38
rs188060252 Chr.22: 38821174-38821174 on Build GRCh38
rs575249739 Chr.22: 38821095-38821095 on Build GRCh38
rs2235136 Chr.22: 38820911-38820911 on Build GRCh38
rs913323384 Chr.22: 38820751-38820751 on Build GRCh38
rs772811442 Chr.22: 38821045-38821045 on Build GRCh38
rs955169958 Chr.22: 38820867-38820867 on Build GRCh38
rs987196934 Chr.22: 38820938-38820938 on Build GRCh38
rs111729970 Chr.22: 38821184-38821184 on Build GRCh38
rs184298013 Chr.22: 38820763-38820763 on Build GRCh38
rs564568822 Chr.22: 38821187-38821187 on Build GRCh38
rs769449749 Chr.22: 38821020-38821020 on Build GRCh38
rs925204231 Chr.22: 38820940-38820940 on Build GRCh38
rs550616654 Chr.22: 38820792-38820792 on Build GRCh38
rs1015142243 Chr.22: 38820889-38820889 on Build GRCh38
rs570974715 Chr.22: 38820956-38820956 on Build GRCh38
rs144206642 Chr.22: 38821162-38821162 on Build GRCh38
rs1022164705 Chr.22: 38820729-38820729 on Build GRCh38
rs1047244425 Chr.22: 38821040-38821040 on Build GRCh38
rs767888472 Chr.22: 38821012-38821012 on Build GRCh38
rs973590475 Chr.22: 38820903-38820903 on Build GRCh38
rs757903573 Chr.22: 38820896-38820898 on Build GRCh38
rs1004882544 Chr.22: 38821113-38821113 on Build GRCh38
rs984121976 Chr.22: 38820832-38820832 on Build GRCh38
rs142366654 Chr.22: 38820845-38820845 on Build GRCh38
rs868267906 Chr.22: 38820749-38820749 on Build GRCh38
rs116118263 Chr.22: 38820812-38820812 on Build GRCh38
rs539596289 Chr.22: 38820919-38820919 on Build GRCh38
rs1008417385 Chr.22: 38820871-38820871 on Build GRCh38
rs146533376 Chr.22: 38821183-38821183 on Build GRCh38
rs372848640 Chr.22: 38821197-38821197 on Build GRCh38
rs1045747117 Chr.22: 38820798-38820798 on Build GRCh38

View Full Product Details