Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 33197373-33197871 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NRP1
Entrez Gene ID
8829
Gene Name
neuropilin 1
Gene Aliases
BDCA4, CD304, NP1, NRP, VEGF165R
UniGene
Hs.131704
Gene Chromosome Location
Chr.10: 33177491-33334905 on Build GRCh38
Transcripts
NM_001024628.2 NM_001024629.2 NM_001244972.1 NM_001244973.1 NM_003873.5 NR_045259.1 XM_006717521.1 XM_006717522.1 XM_006717523.2 XM_006717524.1 XM_006717525.1 XM_006717526.1 XM_011519755.2 XM_011519756.1 XM_017016865.1 XM_017016866.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5650426 c.1898C>T p.T633M Chr.10: 33197676-33197676 on Build GRCh38
1259524 c.1897A>G p.T633A Chr.10: 33197677-33197677 on Build GRCh38
3437378 c.1914C>T p.T638T Chr.10: 33197660-33197660 on Build GRCh38
4881916 c.1902C>G p.V634V Chr.10: 33197672-33197672 on Build GRCh38
4899121 c.155C>T p.T52I Chr.10: 33197660-33197660 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs900849311 Chr.10: 33197472-33197472 on Build GRCh38
rs908358769 Chr.10: 33197417-33197417 on Build GRCh38
rs578147167 Chr.10: 33197590-33197590 on Build GRCh38
rs201288994 Chr.10: 33197676-33197676 on Build GRCh38
rs563594298 Chr.10: 33197670-33197670 on Build GRCh38
rs747818891 Chr.10: 33197709-33197709 on Build GRCh38
rs975394875 Chr.10: 33197790-33197790 on Build GRCh38
rs952677848 Chr.10: 33197788-33197788 on Build GRCh38
rs775776942 Chr.10: 33197750-33197750 on Build GRCh38
rs763021813 Chr.10: 33197627-33197627 on Build GRCh38
rs369227474 Chr.10: 33197622-33197622 on Build GRCh38
rs774225703 Chr.10: 33197738-33197738 on Build GRCh38
rs906165892 Chr.10: 33197500-33197500 on Build GRCh38
rs749852353 Chr.10: 33197595-33197595 on Build GRCh38
rs373576037 Chr.10: 33197731-33197731 on Build GRCh38
rs752665390 Chr.10: 33197653-33197653 on Build GRCh38
rs957953878 Chr.10: 33197539-33197540 on Build GRCh38
rs1033573748 Chr.10: 33197519-33197519 on Build GRCh38
rs769313153 Chr.10: 33197712-33197712 on Build GRCh38
rs889360366 Chr.10: 33197536-33197536 on Build GRCh38
rs758145507 Chr.10: 33197813-33197813 on Build GRCh38
rs767787884 Chr.10: 33197641-33197641 on Build GRCh38
rs746614844 Chr.10: 33197602-33197602 on Build GRCh38
rs1051149641 Chr.10: 33197532-33197532 on Build GRCh38
rs766252120 Chr.10: 33197634-33197634 on Build GRCh38
rs1032527710 Chr.10: 33197583-33197583 on Build GRCh38
rs777471822 Chr.10: 33197718-33197718 on Build GRCh38
rs879216353 Chr.10: 33197593-33197593 on Build GRCh38
rs377496015 Chr.10: 33197597-33197597 on Build GRCh38
rs750790418 Chr.10: 33197698-33197698 on Build GRCh38
rs771821428 Chr.10: 33197748-33197748 on Build GRCh38
rs535142285 Chr.10: 33197460-33197460 on Build GRCh38
rs750413908 Chr.10: 33197622-33197623 on Build GRCh38
rs760848385 Chr.10: 33197758-33197758 on Build GRCh38
rs939786248 Chr.10: 33197451-33197451 on Build GRCh38
rs191501021 Chr.10: 33197614-33197614 on Build GRCh38
rs977498863 Chr.10: 33197598-33197598 on Build GRCh38
rs1046480885 Chr.10: 33197489-33197489 on Build GRCh38
rs10827208 Chr.10: 33197739-33197739 on Build GRCh38
rs112570033 Chr.10: 33197558-33197558 on Build GRCh38
rs780880989 Chr.10: 33197702-33197702 on Build GRCh38
rs145594886 Chr.10: 33197668-33197668 on Build GRCh38
rs747707272 Chr.10: 33197608-33197608 on Build GRCh38
rs768326830 Chr.10: 33197605-33197605 on Build GRCh38
rs553528689 Chr.10: 33197520-33197520 on Build GRCh38
rs752857728 Chr.10: 33197589-33197589 on Build GRCh38
rs759794760 Chr.10: 33197638-33197638 on Build GRCh38
rs544334675 Chr.10: 33197406-33197406 on Build GRCh38
rs377610285 Chr.10: 33197588-33197588 on Build GRCh38
rs750198312 Chr.10: 33197799-33197799 on Build GRCh38
rs867509424 Chr.10: 33197792-33197792 on Build GRCh38
rs541897812 Chr.10: 33197666-33197666 on Build GRCh38
rs373963068 Chr.10: 33197762-33197762 on Build GRCh38
rs1020824480 Chr.10: 33197582-33197582 on Build GRCh38
rs1001874031 Chr.10: 33197507-33197507 on Build GRCh38
rs1025061823 Chr.10: 33197611-33197611 on Build GRCh38
rs550087163 Chr.10: 33197671-33197671 on Build GRCh38
rs564011454 Chr.10: 33197547-33197547 on Build GRCh38
rs757782302 Chr.10: 33197674-33197674 on Build GRCh38
rs757616580 Chr.10: 33197596-33197596 on Build GRCh38
rs926558304 Chr.10: 33197770-33197770 on Build GRCh38
rs778357530 Chr.10: 33197594-33197594 on Build GRCh38
rs773192280 Chr.10: 33197615-33197615 on Build GRCh38

View Full Product Details