Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 11658632-11659175 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NTSR2
Entrez Gene ID
23620
Gene Name
neurotensin receptor 2
Gene Aliases
NTR2
UniGene
Hs.131138
Gene Chromosome Location
Chr.2: 11658178-11670206 on Build GRCh38
Transcripts
NM_012344.3 XM_005246156.3 XM_006711876.3 XM_006711877.3 XM_017003738.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5439732 c.990-1G>A p.? Chr.2: 11658723-11658723 on Build GRCh38
6192267 c.1066C>G p.L356V Chr.2: 11658646-11658646 on Build GRCh38
290507 c.1044C>T p.Y348Y Chr.2: 11658668-11658668 on Build GRCh38
4084401 c.994C>T p.L332L Chr.2: 11658718-11658718 on Build GRCh38
6153611 c.1041C>G p.F347L Chr.2: 11658671-11658671 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762738817 Chr.2: 11658971-11658971 on Build GRCh38
rs547664267 Chr.2: 11658691-11658691 on Build GRCh38
rs764229979 Chr.2: 11658694-11658696 on Build GRCh38
rs781393252 Chr.2: 11658769-11658769 on Build GRCh38
rs368923768 Chr.2: 11658740-11658740 on Build GRCh38
rs895168774 Chr.2: 11658835-11658835 on Build GRCh38
rs933890925 Chr.2: 11659122-11659122 on Build GRCh38
rs200323530 Chr.2: 11658717-11658717 on Build GRCh38
rs933512836 Chr.2: 11658803-11658803 on Build GRCh38
rs55911243 Chr.2: 11658921-11658921 on Build GRCh38
rs1030735600 Chr.2: 11658793-11658793 on Build GRCh38
rs771628693 Chr.2: 11659043-11659043 on Build GRCh38
rs890442631 Chr.2: 11659146-11659146 on Build GRCh38
rs867152745 Chr.2: 11658841-11658841 on Build GRCh38
rs775817836 Chr.2: 11658737-11658737 on Build GRCh38
rs149376121 Chr.2: 11658916-11658916 on Build GRCh38
rs893626372 Chr.2: 11658669-11658669 on Build GRCh38
rs775299094 Chr.2: 11658965-11658965 on Build GRCh38
rs375162705 Chr.2: 11658872-11658875 on Build GRCh38
rs148549427 Chr.2: 11659002-11659002 on Build GRCh38
rs369698451 Chr.2: 11659135-11659135 on Build GRCh38
rs964071313 Chr.2: 11658951-11658951 on Build GRCh38
rs764785562 Chr.2: 11658773-11658773 on Build GRCh38
rs906358893 Chr.2: 11658848-11658848 on Build GRCh38
rs143067546 Chr.2: 11658668-11658668 on Build GRCh38
rs924027568 Chr.2: 11658807-11658807 on Build GRCh38
rs375264909 Chr.2: 11659133-11659133 on Build GRCh38
rs1031032769 Chr.2: 11658966-11658966 on Build GRCh38
rs935060481 Chr.2: 11658661-11658661 on Build GRCh38
rs921124517 Chr.2: 11659089-11659089 on Build GRCh38
rs888214454 Chr.2: 11658908-11658908 on Build GRCh38
rs367870730 Chr.2: 11658699-11658699 on Build GRCh38
rs201182899 Chr.2: 11658690-11658690 on Build GRCh38
rs760604094 Chr.2: 11658676-11658676 on Build GRCh38
rs535054643 Chr.2: 11658882-11658882 on Build GRCh38
rs372105903 Chr.2: 11658743-11658743 on Build GRCh38
rs1039524786 Chr.2: 11659056-11659056 on Build GRCh38
rs746003384 Chr.2: 11658722-11658722 on Build GRCh38
rs750020399 Chr.2: 11658857-11658860 on Build GRCh38
rs772177236 Chr.2: 11658734-11658734 on Build GRCh38
rs138833286 Chr.2: 11658816-11658816 on Build GRCh38
rs963507154 Chr.2: 11658821-11658821 on Build GRCh38
rs754841949 Chr.2: 11658701-11658701 on Build GRCh38
rs761171380 Chr.2: 11658667-11658667 on Build GRCh38
rs764867425 Chr.2: 11658678-11658678 on Build GRCh38
rs543532202 Chr.2: 11659045-11659045 on Build GRCh38
rs915005757 Chr.2: 11659042-11659042 on Build GRCh38
rs80254523 Chr.2: 11658829-11658829 on Build GRCh38
rs781085816 Chr.2: 11658708-11658708 on Build GRCh38
rs936727581 Chr.2: 11659060-11659060 on Build GRCh38
rs868083320 Chr.2: 11658790-11658790 on Build GRCh38
rs775707526 Chr.2: 11658656-11658656 on Build GRCh38
rs916584847 Chr.2: 11659143-11659143 on Build GRCh38
rs569838590 Chr.2: 11658783-11658783 on Build GRCh38
rs1051059269 Chr.2: 11659142-11659142 on Build GRCh38
rs767867743 Chr.2: 11658655-11658655 on Build GRCh38
rs557328686 Chr.2: 11658850-11658850 on Build GRCh38
rs993969577 Chr.2: 11658856-11658857 on Build GRCh38
rs756773817 Chr.2: 11658677-11658677 on Build GRCh38
rs1015045585 Chr.2: 11658817-11658817 on Build GRCh38
rs762417390 Chr.2: 11658749-11658749 on Build GRCh38
rs899779517 Chr.2: 11658781-11658781 on Build GRCh38
rs557566635 Chr.2: 11658934-11658934 on Build GRCh38
rs533164027 Chr.2: 11658738-11658738 on Build GRCh38
rs956436507 Chr.2: 11658945-11658945 on Build GRCh38
rs975018089 Chr.2: 11658958-11658958 on Build GRCh38
rs145966091 Chr.2: 11658713-11658713 on Build GRCh38
rs573734272 Chr.2: 11658996-11658996 on Build GRCh38
rs890486361 Chr.2: 11658810-11658810 on Build GRCh38
rs770929971 Chr.2: 11658714-11658714 on Build GRCh38
rs774427008 Chr.2: 11658721-11658721 on Build GRCh38
rs553633110 Chr.2: 11658883-11658883 on Build GRCh38
rs1023867650 Chr.2: 11658839-11658839 on Build GRCh38

View Full Product Details