Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 225582512-225583005 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NYAP2
Entrez Gene ID
57624
Gene Name
neuronal tyrosine-phosphorylated phosphoinositide-3-kinase adaptor 2
Gene Aliases
KIAA1486
UniGene
Hs.224409
Gene Chromosome Location
Chr.2: 225399710-225703653 on Build GRCh38
Transcripts
NM_020864.1 XM_005246708.3 XM_011511524.1 XM_017004564.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6543991 c.1162C>T p.P388S Chr.2: 225582579-225582579 on Build GRCh38
3578346 c.1314C>T p.S438S Chr.2: 225582731-225582731 on Build GRCh38
4776215 c.1381C>A p.H461N Chr.2: 225582798-225582798 on Build GRCh38
4463157 c.1279C>T p.H427Y Chr.2: 225582696-225582696 on Build GRCh38
6498263 c.1587C>T p.S529S Chr.2: 225583004-225583004 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs562881458 Chr.2: 225582731-225582731 on Build GRCh38
rs898679585 Chr.2: 225582661-225582661 on Build GRCh38
rs765314985 Chr.2: 225582705-225582705 on Build GRCh38
rs750626822 Chr.2: 225582730-225582730 on Build GRCh38
rs186778268 Chr.2: 225582655-225582655 on Build GRCh38
rs376546506 Chr.2: 225582789-225582789 on Build GRCh38
rs770182739 Chr.2: 225582612-225582612 on Build GRCh38
rs367790714 Chr.2: 225582643-225582643 on Build GRCh38
rs767641991 Chr.2: 225582728-225582728 on Build GRCh38
rs760306636 Chr.2: 225582697-225582697 on Build GRCh38
rs771862567 Chr.2: 225582969-225582969 on Build GRCh38
rs745449398 Chr.2: 225582838-225582838 on Build GRCh38
rs761483711 Chr.2: 225582638-225582638 on Build GRCh38
rs749699328 Chr.2: 225582732-225582732 on Build GRCh38
rs756799729 Chr.2: 225582874-225582874 on Build GRCh38
rs761395774 Chr.2: 225582564-225582564 on Build GRCh38
rs865791029 Chr.2: 225582816-225582816 on Build GRCh38
rs759695162 Chr.2: 225582859-225582859 on Build GRCh38
rs750472721 Chr.2: 225582788-225582788 on Build GRCh38
rs766364300 Chr.2: 225582856-225582856 on Build GRCh38
rs755660299 Chr.2: 225582944-225582944 on Build GRCh38
rs770061615 Chr.2: 225582664-225582664 on Build GRCh38
rs771557195 Chr.2: 225582684-225582684 on Build GRCh38
rs747163915 Chr.2: 225582544-225582544 on Build GRCh38
rs776448171 Chr.2: 225582702-225582702 on Build GRCh38
rs964516145 Chr.2: 225582807-225582807 on Build GRCh38
rs370093403 Chr.2: 225582545-225582545 on Build GRCh38
rs772662215 Chr.2: 225582741-225582741 on Build GRCh38
rs765122978 Chr.2: 225582776-225582776 on Build GRCh38
rs774623727 Chr.2: 225582674-225582674 on Build GRCh38
rs775698420 Chr.2: 225582982-225582982 on Build GRCh38
rs767320947 Chr.2: 225582569-225582569 on Build GRCh38
rs774413383 Chr.2: 225582892-225582892 on Build GRCh38
rs775575640 Chr.2: 225582778-225582778 on Build GRCh38
rs1002913652 Chr.2: 225582738-225582738 on Build GRCh38
rs757658935 Chr.2: 225582931-225582931 on Build GRCh38
rs768062699 Chr.2: 225582629-225582629 on Build GRCh38
rs774804783 Chr.2: 225582677-225582677 on Build GRCh38
rs374591560 Chr.2: 225582557-225582557 on Build GRCh38
rs757394180 Chr.2: 225582658-225582658 on Build GRCh38
rs371641080 Chr.2: 225582668-225582668 on Build GRCh38
rs770658843 Chr.2: 225582754-225582754 on Build GRCh38
rs201024968 Chr.2: 225582870-225582870 on Build GRCh38
rs769528707 Chr.2: 225582884-225582884 on Build GRCh38
rs749141197 Chr.2: 225582888-225582888 on Build GRCh38
rs914146128 Chr.2: 225582918-225582918 on Build GRCh38
rs375542974 Chr.2: 225582734-225582734 on Build GRCh38
rs779906824 Chr.2: 225582945-225582945 on Build GRCh38
rs574157216 Chr.2: 225582653-225582653 on Build GRCh38
rs769163693 Chr.2: 225582675-225582675 on Build GRCh38
rs752903462 Chr.2: 225582924-225582924 on Build GRCh38
rs138500026 Chr.2: 225582925-225582925 on Build GRCh38
rs748940117 Chr.2: 225582959-225582959 on Build GRCh38
rs926378534 Chr.2: 225582880-225582880 on Build GRCh38
rs563650514 Chr.2: 225582966-225582966 on Build GRCh38
rs762928339 Chr.2: 225582782-225582782 on Build GRCh38
rs199867716 Chr.2: 225582844-225582844 on Build GRCh38
rs755401706 Chr.2: 225582803-225582803 on Build GRCh38
rs538675320 Chr.2: 225582546-225582546 on Build GRCh38
rs776415672 Chr.2: 225582823-225582823 on Build GRCh38
rs907521109 Chr.2: 225582903-225582903 on Build GRCh38
rs375182785 Chr.2: 225582589-225582589 on Build GRCh38
rs865814391 Chr.2: 225582802-225582802 on Build GRCh38
rs939512527 Chr.2: 225582889-225582889 on Build GRCh38
rs551391208 Chr.2: 225582735-225582735 on Build GRCh38
rs773800325 Chr.2: 225582637-225582637 on Build GRCh38
rs760442473 Chr.2: 225582644-225582644 on Build GRCh38
rs780787340 Chr.2: 225582935-225582935 on Build GRCh38
rs745540004 Chr.2: 225582877-225582877 on Build GRCh38
rs762221271 Chr.2: 225582875-225582875 on Build GRCh38
rs371936878 Chr.2: 225582912-225582912 on Build GRCh38
rs749778663 Chr.2: 225582673-225582673 on Build GRCh38
rs758472575 Chr.2: 225582582-225582582 on Build GRCh38
rs765610435 Chr.2: 225582860-225582860 on Build GRCh38
rs9917362 Chr.2: 225582590-225582590 on Build GRCh38
rs759438407 Chr.2: 225582772-225582772 on Build GRCh38
rs777853612 Chr.2: 225582584-225582584 on Build GRCh38
rs776126554 Chr.2: 225582770-225582770 on Build GRCh38
rs951402666 Chr.2: 225582792-225582792 on Build GRCh38
rs760861891 Chr.2: 225582855-225582855 on Build GRCh38
rs759222178 Chr.2: 225582703-225582703 on Build GRCh38
rs770119189 Chr.2: 225582886-225582886 on Build GRCh38
rs774130262 Chr.2: 225582854-225582854 on Build GRCh38
rs749990764 Chr.2: 225582943-225582943 on Build GRCh38
rs746672929 Chr.2: 225582752-225582752 on Build GRCh38
rs779818817 Chr.2: 225582887-225582887 on Build GRCh38
rs762429077 Chr.2: 225582554-225582554 on Build GRCh38
rs771055499 Chr.2: 225582914-225582914 on Build GRCh38
rs201650497 Chr.2: 225582563-225582563 on Build GRCh38
rs780576502 Chr.2: 225582971-225582973 on Build GRCh38
rs768501881 Chr.2: 225582960-225582960 on Build GRCh38
rs181896828 Chr.2: 225582551-225582551 on Build GRCh38
rs748665219 Chr.2: 225582805-225582805 on Build GRCh38
rs751729250 Chr.2: 225582656-225582656 on Build GRCh38
rs372160955 Chr.2: 225582609-225582609 on Build GRCh38
rs375205040 Chr.2: 225582613-225582613 on Build GRCh38
rs746284766 Chr.2: 225582611-225582611 on Build GRCh38
rs758865113 Chr.2: 225582815-225582815 on Build GRCh38
rs372148969 Chr.2: 225582767-225582767 on Build GRCh38
rs761588621 Chr.2: 225582913-225582913 on Build GRCh38
rs781625402 Chr.2: 225582662-225582662 on Build GRCh38
rs770333813 Chr.2: 225582821-225582821 on Build GRCh38
rs752672785 Chr.2: 225582718-225582718 on Build GRCh38
rs558213296 Chr.2: 225582947-225582947 on Build GRCh38
rs866736227 Chr.2: 225582759-225582759 on Build GRCh38
rs376429827 Chr.2: 225582576-225582576 on Build GRCh38
rs868521963 Chr.2: 225582764-225582764 on Build GRCh38
rs768553811 Chr.2: 225582890-225582890 on Build GRCh38
rs9917363 Chr.2: 225582791-225582791 on Build GRCh38
rs866863305 Chr.2: 225582699-225582700 on Build GRCh38
rs909587287 Chr.2: 225582813-225582813 on Build GRCh38
rs763013429 Chr.2: 225582846-225582846 on Build GRCh38
rs372494659 Chr.2: 225582865-225582865 on Build GRCh38
rs773627897 Chr.2: 225582922-225582922 on Build GRCh38
rs752009264 Chr.2: 225582872-225582872 on Build GRCh38
rs201097764 Chr.2: 225582978-225582978 on Build GRCh38
rs776044334 Chr.2: 225582553-225582553 on Build GRCh38
rs760739357 Chr.2: 225582716-225582716 on Build GRCh38
rs773361794 Chr.2: 225582542-225582542 on Build GRCh38
rs371568593 Chr.2: 225582920-225582920 on Build GRCh38
rs754094495 Chr.2: 225582857-225582857 on Build GRCh38
rs779180636 Chr.2: 225582804-225582804 on Build GRCh38
rs150128802 Chr.2: 225582820-225582820 on Build GRCh38
rs775433682 Chr.2: 225582559-225582559 on Build GRCh38
rs1045254100 Chr.2: 225582550-225582550 on Build GRCh38
rs767725801 Chr.2: 225582783-225582783 on Build GRCh38
rs772914553 Chr.2: 225582694-225582694 on Build GRCh38
rs369970591 Chr.2: 225582724-225582724 on Build GRCh38
rs772165755 Chr.2: 225582539-225582539 on Build GRCh38
rs565329041 Chr.2: 225582652-225582652 on Build GRCh38
rs753992827 Chr.2: 225582798-225582798 on Build GRCh38
rs933063222 Chr.2: 225582869-225582869 on Build GRCh38
rs778402285 Chr.2: 225582871-225582871 on Build GRCh38
rs753797313 Chr.2: 225582580-225582580 on Build GRCh38
rs776522331 Chr.2: 225582915-225582915 on Build GRCh38
rs974514188 Chr.2: 225582817-225582817 on Build GRCh38
rs191992954 Chr.2: 225582842-225582842 on Build GRCh38
rs377307715 Chr.2: 225582682-225582682 on Build GRCh38
rs768751831 Chr.2: 225582850-225582850 on Build GRCh38
rs746005558 Chr.2: 225582981-225582981 on Build GRCh38
rs746087538 Chr.2: 225582663-225582663 on Build GRCh38

View Full Product Details