Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 39672114-39672603 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PGAP3
Entrez Gene ID
93210
Gene Name
post-GPI attachment to proteins 3
Gene Aliases
AGLA546, CAB2, PERLD1, PP1498, hCOS16
UniGene
Hs.462971
Gene Chromosome Location
Chr.17: 39671122-39688070 on Build GRCh38
Transcripts
NM_001291726.1 NM_001291728.1 NM_001291730.1 NM_001291732.1 NM_001291733.1 NM_033419.4 XM_011525480.2 XM_011525481.2

SNP Detail

SNP ID Chromosome Location
rs549779237 Chr.17: 39672167-39672167 on Build GRCh38
rs553234860 Chr.17: 39672283-39672283 on Build GRCh38
rs571425527 Chr.17: 39672210-39672210 on Build GRCh38
rs971540116 Chr.17: 39672241-39672241 on Build GRCh38
rs898308738 Chr.17: 39672307-39672307 on Build GRCh38
rs958743566 Chr.17: 39672259-39672259 on Build GRCh38
rs960584218 Chr.17: 39672322-39672322 on Build GRCh38
rs980587911 Chr.17: 39672422-39672422 on Build GRCh38
rs990518542 Chr.17: 39672281-39672281 on Build GRCh38
rs923799016 Chr.17: 39672365-39672365 on Build GRCh38
rs1034442327 Chr.17: 39672552-39672552 on Build GRCh38
rs1003009962 Chr.17: 39672543-39672543 on Build GRCh38
rs959083723 Chr.17: 39672563-39672563 on Build GRCh38
rs780223575 Chr.17: 39672520-39672520 on Build GRCh38
rs576127307 Chr.17: 39672294-39672294 on Build GRCh38
rs970123930 Chr.17: 39672393-39672394 on Build GRCh38
rs141494631 Chr.17: 39672400-39672400 on Build GRCh38
rs946669737 Chr.17: 39672311-39672311 on Build GRCh38
rs910881607 Chr.17: 39672566-39672566 on Build GRCh38
rs1040081506 Chr.17: 39672132-39672132 on Build GRCh38
rs371712952 Chr.17: 39672472-39672472 on Build GRCh38
rs2941503 Chr.17: 39672492-39672492 on Build GRCh38
rs914576583 Chr.17: 39672308-39672308 on Build GRCh38
rs900182632 Chr.17: 39672140-39672140 on Build GRCh38
rs554631717 Chr.17: 39672379-39672379 on Build GRCh38
rs926090133 Chr.17: 39672464-39672464 on Build GRCh38
rs988957162 Chr.17: 39672501-39672501 on Build GRCh38
rs2952151 Chr.17: 39672243-39672243 on Build GRCh38
rs907087 Chr.17: 39672534-39672534 on Build GRCh38
rs996216010 Chr.17: 39672157-39672157 on Build GRCh38
rs1005133303 Chr.17: 39672195-39672195 on Build GRCh38
rs944518605 Chr.17: 39672372-39672372 on Build GRCh38
rs900215555 Chr.17: 39672414-39672414 on Build GRCh38
rs887985755 Chr.17: 39672188-39672188 on Build GRCh38
rs931667440 Chr.17: 39672435-39672435 on Build GRCh38
rs534891529 Chr.17: 39672280-39672280 on Build GRCh38
rs977965750 Chr.17: 39672363-39672363 on Build GRCh38
rs183208638 Chr.17: 39672244-39672244 on Build GRCh38
rs1049240911 Chr.17: 39672464-39672464 on Build GRCh38
rs1025449854 Chr.17: 39672309-39672309 on Build GRCh38
rs547147114 Chr.17: 39672233-39672233 on Build GRCh38
rs779294992 Chr.17: 39672521-39672521 on Build GRCh38
rs538942316 Chr.17: 39672218-39672218 on Build GRCh38
rs1050712609 Chr.17: 39672203-39672203 on Build GRCh38
rs754458769 Chr.17: 39672204-39672204 on Build GRCh38

View Full Product Details