Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 242219846-242220385 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLD5
Entrez Gene ID
200150
Gene Name
phospholipase D family member 5
Gene Aliases
PLDC
UniGene
Hs.672452
Gene Chromosome Location
Chr.1: 242085051-242530546 on Build GRCh38
Transcripts
NM_001195811.1 NM_001195812.1 NM_001320272.1 NM_152666.2 XM_006711752.2 XM_011544115.2 XM_011544116.2 XM_011544119.2 XM_011544120.2 XM_011544121.2 XM_011544122.2 XM_017000567.1 XM_017000568.1 XM_017000569.1 XM_017000570.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2050416 c.608-5C>T p.? Chr.1: 242220120-242220120 on Build GRCh38
679643 c.424G>T p.G142C Chr.1: 242220023-242220023 on Build GRCh38
1139111 c.620C>T p.T207M Chr.1: 242220103-242220103 on Build GRCh38
6748632 c.523G>A p.G175S Chr.1: 242220014-242220014 on Build GRCh38
4812975 c.478T>C p.S160P Chr.1: 242220059-242220059 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747819285 Chr.1: 242220157-242220157 on Build GRCh38
rs757063070 Chr.1: 242220049-242220049 on Build GRCh38
rs568515767 Chr.1: 242220111-242220111 on Build GRCh38
rs149637368 Chr.1: 242219933-242219933 on Build GRCh38
rs763359971 Chr.1: 242220108-242220108 on Build GRCh38
rs1030635425 Chr.1: 242220184-242220184 on Build GRCh38
rs747591361 Chr.1: 242219974-242219974 on Build GRCh38
rs751001547 Chr.1: 242220030-242220030 on Build GRCh38
rs1027071454 Chr.1: 242220017-242220017 on Build GRCh38
rs369597039 Chr.1: 242220102-242220102 on Build GRCh38
rs749146241 Chr.1: 242219970-242219970 on Build GRCh38
rs780929006 Chr.1: 242220152-242220152 on Build GRCh38
rs985976385 Chr.1: 242220316-242220316 on Build GRCh38
rs537939295 Chr.1: 242220306-242220307 on Build GRCh38
rs771552372 Chr.1: 242220079-242220079 on Build GRCh38
rs1049809905 Chr.1: 242219896-242219896 on Build GRCh38
rs751513380 Chr.1: 242220266-242220266 on Build GRCh38
rs373083912 Chr.1: 242219987-242219987 on Build GRCh38
rs574104344 Chr.1: 242220344-242220344 on Build GRCh38
rs58009715 Chr.1: 242220273-242220274 on Build GRCh38
rs758373657 Chr.1: 242220245-242220245 on Build GRCh38
rs750289670 Chr.1: 242220144-242220144 on Build GRCh38
rs773800524 Chr.1: 242220103-242220103 on Build GRCh38
rs1973498 Chr.1: 242219899-242219899 on Build GRCh38
rs994331871 Chr.1: 242220248-242220248 on Build GRCh38
rs137997860 Chr.1: 242220091-242220091 on Build GRCh38
rs141155291 Chr.1: 242219989-242219989 on Build GRCh38
rs144892956 Chr.1: 242220024-242220024 on Build GRCh38
rs373602787 Chr.1: 242220310-242220311 on Build GRCh38
rs765027465 Chr.1: 242220137-242220137 on Build GRCh38
rs754961540 Chr.1: 242220326-242220326 on Build GRCh38
rs752944359 Chr.1: 242220129-242220129 on Build GRCh38
rs368160969 Chr.1: 242220012-242220013 on Build GRCh38
rs746569677 Chr.1: 242220087-242220087 on Build GRCh38
rs755146825 Chr.1: 242220037-242220037 on Build GRCh38
rs945764429 Chr.1: 242219883-242219883 on Build GRCh38
rs970417261 Chr.1: 242220135-242220135 on Build GRCh38
rs546531082 Chr.1: 242219889-242219889 on Build GRCh38
rs569350964 Chr.1: 242220015-242220015 on Build GRCh38
rs888506753 Chr.1: 242220277-242220277 on Build GRCh38
rs976587414 Chr.1: 242219873-242219873 on Build GRCh38
rs771316638 Chr.1: 242220166-242220166 on Build GRCh38
rs944081153 Chr.1: 242220339-242220339 on Build GRCh38
rs7547029 Chr.1: 242220208-242220208 on Build GRCh38
rs534178349 Chr.1: 242220147-242220147 on Build GRCh38
rs199880915 Chr.1: 242220160-242220160 on Build GRCh38
rs781428918 Chr.1: 242220038-242220038 on Build GRCh38
rs140243407 Chr.1: 242220086-242220086 on Build GRCh38
rs753625409 Chr.1: 242220315-242220316 on Build GRCh38
rs762317137 Chr.1: 242220018-242220018 on Build GRCh38
rs751738939 Chr.1: 242219939-242219939 on Build GRCh38
rs1055441859 Chr.1: 242220182-242220182 on Build GRCh38
rs189693721 Chr.1: 242220218-242220218 on Build GRCh38
rs144628781 Chr.1: 242220220-242220220 on Build GRCh38
rs748484479 Chr.1: 242220099-242220099 on Build GRCh38
rs775138745 Chr.1: 242219997-242219997 on Build GRCh38
rs895382426 Chr.1: 242219934-242219934 on Build GRCh38
rs746457397 Chr.1: 242220149-242220149 on Build GRCh38
rs760256722 Chr.1: 242220000-242220000 on Build GRCh38
rs869087967 Chr.1: 242219872-242219873 on Build GRCh38
rs1014190133 Chr.1: 242219943-242219943 on Build GRCh38
rs548680899 Chr.1: 242220110-242220110 on Build GRCh38
rs767674498 Chr.1: 242220114-242220114 on Build GRCh38
rs967180797 Chr.1: 242220126-242220126 on Build GRCh38
rs751723743 Chr.1: 242220033-242220033 on Build GRCh38
rs753605572 Chr.1: 242220048-242220048 on Build GRCh38
rs552058954 Chr.1: 242220014-242220014 on Build GRCh38
rs397692938 Chr.1: 242219871-242219872 on Build GRCh38
rs554458651 Chr.1: 242220150-242220150 on Build GRCh38
rs778867183 Chr.1: 242219975-242219975 on Build GRCh38
rs540037498 Chr.1: 242220165-242220165 on Build GRCh38
rs768614434 Chr.1: 242219974-242219975 on Build GRCh38
rs369351636 Chr.1: 242219961-242219961 on Build GRCh38
rs759706257 Chr.1: 242220113-242220113 on Build GRCh38
rs1048531025 Chr.1: 242220259-242220259 on Build GRCh38
rs978515076 Chr.1: 242220128-242220128 on Build GRCh38
rs936999899 Chr.1: 242219921-242219921 on Build GRCh38
rs912984509 Chr.1: 242219879-242219879 on Build GRCh38
rs770821680 Chr.1: 242219974-242219974 on Build GRCh38
rs869234780 Chr.1: 242220331-242220335 on Build GRCh38
rs776827970 Chr.1: 242220053-242220055 on Build GRCh38
rs113967982 Chr.1: 242220307-242220311 on Build GRCh38
rs772123445 Chr.1: 242220311-242220312 on Build GRCh38
rs375225001 Chr.1: 242220057-242220057 on Build GRCh38
rs556512668 Chr.1: 242220170-242220170 on Build GRCh38
rs931242659 Chr.1: 242220353-242220353 on Build GRCh38
rs773710963 Chr.1: 242220164-242220164 on Build GRCh38
rs78458271 Chr.1: 242220335-242220335 on Build GRCh38
rs768171241 Chr.1: 242220088-242220088 on Build GRCh38
rs745808121 Chr.1: 242220071-242220071 on Build GRCh38
rs200172213 Chr.1: 242220311-242220311 on Build GRCh38
rs146236115 Chr.1: 242220016-242220017 on Build GRCh38
rs755883100 Chr.1: 242219940-242219940 on Build GRCh38
rs912638216 Chr.1: 242220322-242220322 on Build GRCh38

View Full Product Details