Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 45287700-45288225 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC642852
Entrez Gene ID
642852
Gene Name
uncharacterized LOC642852
Gene Aliases
-
UniGene
Hs.11637
Gene Chromosome Location
Chr.21: 45288052-45297354 on Build GRCh38
Transcripts
NR_026943.1 NM_015227.4 NM_133635.4 NR_004858.1 XM_006723981.3 XM_011529504.2 XM_011529505.2 XM_011529506.2 XM_011529507.2 XM_011529508.2 XM_011529509.2 XM_011529511.2 XM_011529512.2 XM_011529513.2 XM_017028306.1 XM_017028307.1 XM_017028308.1

Gene Symbol
POFUT2
Entrez Gene ID
23275
Gene Name
protein O-fucosyltransferase 2
Gene Aliases
C21orf80, FUT13
UniGene
Hs.592164
Gene Chromosome Location
Chr.21: 45263928-45287912 on Build GRCh38
Transcripts
NR_026943.1 NM_015227.4 NM_133635.4 NR_004858.1 XM_006723981.3 XM_011529504.2 XM_011529505.2 XM_011529506.2 XM_011529507.2 XM_011529508.2 XM_011529509.2 XM_011529511.2 XM_011529512.2 XM_011529513.2 XM_017028306.1 XM_017028307.1 XM_017028308.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4716348 c.110C>T p.S37L Chr.21: 45287762-45287762 on Build GRCh38
4922873 c.116C>T p.A39V Chr.21: 45287756-45287756 on Build GRCh38
4716347 c.110C>T p.S37L Chr.21: 45287762-45287762 on Build GRCh38
6586612 c.80G>C p.W27S Chr.21: 45287792-45287792 on Build GRCh38
4922872 c.116C>T p.A39V Chr.21: 45287756-45287756 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778594805 Chr.21: 45287787-45287787 on Build GRCh38
rs973802801 Chr.21: 45288186-45288187 on Build GRCh38
rs182054010 Chr.21: 45287879-45287879 on Build GRCh38
rs552831711 Chr.21: 45287884-45287884 on Build GRCh38
rs1019421998 Chr.21: 45287799-45287815 on Build GRCh38
rs187050046 Chr.21: 45288171-45288171 on Build GRCh38
rs553849885 Chr.21: 45288007-45288007 on Build GRCh38
rs373102159 Chr.21: 45287819-45287819 on Build GRCh38
rs932569560 Chr.21: 45287934-45287934 on Build GRCh38
rs758260385 Chr.21: 45287803-45287815 on Build GRCh38
rs987503659 Chr.21: 45288128-45288128 on Build GRCh38
rs996656787 Chr.21: 45287842-45287842 on Build GRCh38
rs764394521 Chr.21: 45287746-45287746 on Build GRCh38
rs777680198 Chr.21: 45287806-45287806 on Build GRCh38
rs933250189 Chr.21: 45288004-45288004 on Build GRCh38
rs943892846 Chr.21: 45288065-45288065 on Build GRCh38
rs1037979589 Chr.21: 45287927-45287927 on Build GRCh38
rs1003574259 Chr.21: 45287910-45287910 on Build GRCh38
rs866159994 Chr.21: 45287754-45287754 on Build GRCh38
rs765084050 Chr.21: 45287788-45287789 on Build GRCh38
rs909305580 Chr.21: 45287730-45287730 on Build GRCh38
rs370576674 Chr.21: 45287877-45287877 on Build GRCh38
rs1053914049 Chr.21: 45287874-45287874 on Build GRCh38
rs377224822 Chr.21: 45287845-45287845 on Build GRCh38
rs1033004844 Chr.21: 45287991-45287991 on Build GRCh38
rs763483178 Chr.21: 45287736-45287736 on Build GRCh38
rs890963298 Chr.21: 45287968-45287968 on Build GRCh38
rs760217799 Chr.21: 45287762-45287762 on Build GRCh38
rs943328612 Chr.21: 45288166-45288166 on Build GRCh38
rs535164280 Chr.21: 45288005-45288005 on Build GRCh38
rs112085313 Chr.21: 45288057-45288057 on Build GRCh38
rs774696527 Chr.21: 45287840-45287840 on Build GRCh38
rs774784587 Chr.21: 45287883-45287883 on Build GRCh38
rs757406740 Chr.21: 45287813-45287813 on Build GRCh38
rs991684151 Chr.21: 45287928-45287928 on Build GRCh38
rs908255698 Chr.21: 45287922-45287922 on Build GRCh38
rs749440477 Chr.21: 45287878-45287878 on Build GRCh38
rs1036384359 Chr.21: 45287743-45287743 on Build GRCh38
rs985125382 Chr.21: 45288207-45288207 on Build GRCh38
rs981607609 Chr.21: 45288016-45288016 on Build GRCh38
rs753276608 Chr.21: 45287779-45287779 on Build GRCh38
rs2297286 Chr.21: 45287982-45287982 on Build GRCh38
rs746867295 Chr.21: 45287811-45287811 on Build GRCh38
rs1016472125 Chr.21: 45288068-45288068 on Build GRCh38
rs764184453 Chr.21: 45287863-45287863 on Build GRCh38
rs764814144 Chr.21: 45287860-45287860 on Build GRCh38
rs575925311 Chr.21: 45288112-45288112 on Build GRCh38
rs1042085064 Chr.21: 45288203-45288203 on Build GRCh38
rs1008034215 Chr.21: 45287971-45287971 on Build GRCh38
rs1015753844 Chr.21: 45288167-45288167 on Build GRCh38
rs915115759 Chr.21: 45288183-45288183 on Build GRCh38
rs758455693 Chr.21: 45287801-45287801 on Build GRCh38
rs998938992 Chr.21: 45288082-45288082 on Build GRCh38
rs749777463 Chr.21: 45287829-45287829 on Build GRCh38
rs906303913 Chr.21: 45288205-45288205 on Build GRCh38
rs909274770 Chr.21: 45287977-45287977 on Build GRCh38
rs932286674 Chr.21: 45288048-45288048 on Build GRCh38
rs1055954930 Chr.21: 45287720-45287720 on Build GRCh38
rs775911000 Chr.21: 45287735-45287735 on Build GRCh38
rs888986561 Chr.21: 45287854-45287854 on Build GRCh38
rs972048921 Chr.21: 45287985-45287985 on Build GRCh38
rs945212395 Chr.21: 45288186-45288186 on Build GRCh38
rs941979547 Chr.21: 45287731-45287731 on Build GRCh38
rs775569698 Chr.21: 45287826-45287826 on Build GRCh38
rs1038262752 Chr.21: 45287831-45287831 on Build GRCh38
rs765049117 Chr.21: 45287906-45287906 on Build GRCh38
rs377639659 Chr.21: 45288051-45288051 on Build GRCh38
rs773494506 Chr.21: 45287895-45287895 on Build GRCh38
rs752560481 Chr.21: 45287797-45287799 on Build GRCh38
rs752294862 Chr.21: 45287862-45287862 on Build GRCh38
rs759192400 Chr.21: 45287899-45287899 on Build GRCh38
rs893150423 Chr.21: 45288108-45288108 on Build GRCh38
rs772430976 Chr.21: 45287885-45287885 on Build GRCh38
rs562946451 Chr.21: 45287775-45287775 on Build GRCh38
rs1033054356 Chr.21: 45288090-45288090 on Build GRCh38
rs776567342 Chr.21: 45287721-45287721 on Build GRCh38
rs539226180 Chr.21: 45288024-45288024 on Build GRCh38
rs200209368 Chr.21: 45287908-45287908 on Build GRCh38
rs574830414 Chr.21: 45288046-45288046 on Build GRCh38
rs746053915 Chr.21: 45287873-45287873 on Build GRCh38
rs751462559 Chr.21: 45287864-45287864 on Build GRCh38
rs530089816 Chr.21: 45287835-45287835 on Build GRCh38
rs548671933 Chr.21: 45287865-45287865 on Build GRCh38
rs754441886 Chr.21: 45287784-45287784 on Build GRCh38
rs1013484844 Chr.21: 45288063-45288063 on Build GRCh38
rs12627014 Chr.21: 45288001-45288001 on Build GRCh38
rs1049594836 Chr.21: 45287945-45287945 on Build GRCh38
rs898393148 Chr.21: 45287836-45287836 on Build GRCh38
rs766125563 Chr.21: 45287770-45287770 on Build GRCh38
rs1042033817 Chr.21: 45288034-45288034 on Build GRCh38
rs759550317 Chr.21: 45287723-45287723 on Build GRCh38
rs746083952 Chr.21: 45287817-45287817 on Build GRCh38
rs1051597841 Chr.21: 45287841-45287841 on Build GRCh38
rs1042210614 Chr.21: 45288067-45288067 on Build GRCh38
rs972181728 Chr.21: 45288084-45288084 on Build GRCh38
rs1018996272 Chr.21: 45287872-45287872 on Build GRCh38
rs762492747 Chr.21: 45287911-45287911 on Build GRCh38
rs780882458 Chr.21: 45287871-45287871 on Build GRCh38
rs1009305297 Chr.21: 45288029-45288029 on Build GRCh38
rs751974716 Chr.21: 45287756-45287756 on Build GRCh38
rs910478333 Chr.21: 45288006-45288006 on Build GRCh38
rs759718345 Chr.21: 45287728-45287728 on Build GRCh38
rs959003523 Chr.21: 45288058-45288058 on Build GRCh38
rs919344445 Chr.21: 45288110-45288110 on Build GRCh38
rs769778841 Chr.21: 45287729-45287729 on Build GRCh38
rs373925792 Chr.21: 45288017-45288017 on Build GRCh38
rs910441080 Chr.21: 45288147-45288147 on Build GRCh38
rs779709890 Chr.21: 45288054-45288054 on Build GRCh38
rs751002338 Chr.21: 45288072-45288072 on Build GRCh38
rs371337628 Chr.21: 45287773-45287773 on Build GRCh38
rs538579817 Chr.21: 45287924-45287924 on Build GRCh38
rs776304042 Chr.21: 45287856-45287856 on Build GRCh38
rs963451015 Chr.21: 45288073-45288073 on Build GRCh38
rs957648015 Chr.21: 45288000-45288000 on Build GRCh38

View Full Product Details