Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 60960374-60960913 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PUS10
Entrez Gene ID
150962
Gene Name
pseudouridylate synthase 10
Gene Aliases
CCDC139, DOBI
UniGene
Hs.368348
Gene Chromosome Location
Chr.2: 60938352-61018283 on Build GRCh38
Transcripts
NM_001322123.1 NM_001322124.1 NM_001322127.1 NM_144709.3 XM_011532568.2 XM_011532570.1 XM_011532571.1 XM_011532572.1 XM_011532573.2 XM_011532574.2 XM_011532576.2 XM_011532578.1 XM_017003428.1 XM_017003429.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
322931 c.897G>C p.R299S Chr.2: 60960495-60960495 on Build GRCh38
4314463 c.994G>A p.A332T Chr.2: 60960398-60960398 on Build GRCh38
6326354 c.994G>T p.A332S Chr.2: 60960398-60960398 on Build GRCh38
5726896 c.894C>A p.S298S Chr.2: 60960498-60960498 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs935697641 Chr.2: 60960511-60960511 on Build GRCh38
rs190387889 Chr.2: 60960661-60960661 on Build GRCh38
rs751591322 Chr.2: 60960504-60960504 on Build GRCh38
rs79703662 Chr.2: 60960649-60960649 on Build GRCh38
rs1047175637 Chr.2: 60960742-60960742 on Build GRCh38
rs770116469 Chr.2: 60960538-60960538 on Build GRCh38
rs931012193 Chr.2: 60960634-60960634 on Build GRCh38
rs771553189 Chr.2: 60960434-60960434 on Build GRCh38
rs759859618 Chr.2: 60960844-60960844 on Build GRCh38
rs772733477 Chr.2: 60960445-60960445 on Build GRCh38
rs1001455764 Chr.2: 60960850-60960850 on Build GRCh38
rs771641398 Chr.2: 60960556-60960556 on Build GRCh38
rs34578958 Chr.2: 60960862-60960862 on Build GRCh38
rs200455777 Chr.2: 60960844-60960845 on Build GRCh38
rs777746745 Chr.2: 60960499-60960499 on Build GRCh38
rs919837822 Chr.2: 60960594-60960594 on Build GRCh38
rs939968240 Chr.2: 60960549-60960549 on Build GRCh38
rs781716786 Chr.2: 60960525-60960525 on Build GRCh38
rs988567512 Chr.2: 60960633-60960633 on Build GRCh38
rs746512385 Chr.2: 60960559-60960559 on Build GRCh38
rs954129547 Chr.2: 60960801-60960801 on Build GRCh38
rs1051340173 Chr.2: 60960659-60960659 on Build GRCh38
rs576494257 Chr.2: 60960718-60960718 on Build GRCh38
rs994201532 Chr.2: 60960730-60960730 on Build GRCh38
rs1023649921 Chr.2: 60960818-60960818 on Build GRCh38
rs970357091 Chr.2: 60960843-60960844 on Build GRCh38
rs543843090 Chr.2: 60960856-60960856 on Build GRCh38
rs890005597 Chr.2: 60960665-60960665 on Build GRCh38
rs746288390 Chr.2: 60960530-60960530 on Build GRCh38
rs369069906 Chr.2: 60960489-60960489 on Build GRCh38
rs1021664162 Chr.2: 60960706-60960706 on Build GRCh38
rs564430350 Chr.2: 60960715-60960715 on Build GRCh38
rs534503672 Chr.2: 60960566-60960566 on Build GRCh38
rs369491623 Chr.2: 60960586-60960586 on Build GRCh38
rs1028290253 Chr.2: 60960765-60960765 on Build GRCh38
rs869117673 Chr.2: 60960859-60960862 on Build GRCh38
rs1056647078 Chr.2: 60960669-60960669 on Build GRCh38
rs753632354 Chr.2: 60960487-60960487 on Build GRCh38
rs377112557 Chr.2: 60960481-60960481 on Build GRCh38
rs904098121 Chr.2: 60960728-60960728 on Build GRCh38
rs913013313 Chr.2: 60960863-60960863 on Build GRCh38
rs372174292 Chr.2: 60960493-60960493 on Build GRCh38
rs765628547 Chr.2: 60960845-60960845 on Build GRCh38
rs751723845 Chr.2: 60960788-60960788 on Build GRCh38
rs1037387938 Chr.2: 60960582-60960582 on Build GRCh38
rs988541298 Chr.2: 60960861-60960861 on Build GRCh38
rs11690328 Chr.2: 60960867-60960867 on Build GRCh38
rs760504857 Chr.2: 60960486-60960486 on Build GRCh38
rs925795336 Chr.2: 60960408-60960408 on Build GRCh38
rs747824781 Chr.2: 60960542-60960542 on Build GRCh38
rs1009720260 Chr.2: 60960700-60960700 on Build GRCh38
rs67745317 Chr.2: 60960844-60960844 on Build GRCh38
rs938408825 Chr.2: 60960652-60960652 on Build GRCh38
rs757720080 Chr.2: 60960522-60960522 on Build GRCh38
rs760530572 Chr.2: 60960462-60960462 on Build GRCh38
rs961530232 Chr.2: 60960855-60960855 on Build GRCh38
rs770421082 Chr.2: 60960843-60960844 on Build GRCh38
rs776598524 Chr.2: 60960568-60960568 on Build GRCh38
rs1044004269 Chr.2: 60960647-60960647 on Build GRCh38
rs896674472 Chr.2: 60960679-60960679 on Build GRCh38
rs573709064 Chr.2: 60960539-60960539 on Build GRCh38
rs569199154 Chr.2: 60960731-60960731 on Build GRCh38
rs972592200 Chr.2: 60960857-60960857 on Build GRCh38
rs1000875997 Chr.2: 60960733-60960733 on Build GRCh38
rs912362633 Chr.2: 60960483-60960483 on Build GRCh38
rs986855686 Chr.2: 60960810-60960810 on Build GRCh38
rs368859825 Chr.2: 60960400-60960400 on Build GRCh38
rs773992213 Chr.2: 60960407-60960407 on Build GRCh38
rs896470613 Chr.2: 60960758-60960758 on Build GRCh38
rs761356820 Chr.2: 60960416-60960416 on Build GRCh38
rs768706252 Chr.2: 60960842-60960844 on Build GRCh38
rs185625936 Chr.2: 60960658-60960658 on Build GRCh38
rs752944700 Chr.2: 60960498-60960498 on Build GRCh38

View Full Product Details