Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 53088093-53088587 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RFT1
Entrez Gene ID
91869
Gene Name
RFT1 homolog
Gene Aliases
CDG1N
UniGene
Hs.631910
Gene Chromosome Location
Chr.3: 53071151-53130469 on Build GRCh38
Transcripts
NM_052859.3 XM_006713384.3 XM_011534214.2 XM_011534215.2 XM_017007460.1

SNP Detail

SNP ID Chromosome Location
rs964446256 Chr.3: 53088370-53088370 on Build GRCh38
rs189103179 Chr.3: 53088424-53088424 on Build GRCh38
rs942011479 Chr.3: 53088238-53088238 on Build GRCh38
rs2581782 Chr.3: 53088241-53088241 on Build GRCh38
rs181386256 Chr.3: 53088406-53088406 on Build GRCh38
rs565093798 Chr.3: 53088519-53088519 on Build GRCh38
rs72965386 Chr.3: 53088209-53088209 on Build GRCh38
rs376801362 Chr.3: 53088365-53088365 on Build GRCh38
rs72965387 Chr.3: 53088458-53088458 on Build GRCh38
rs899898266 Chr.3: 53088240-53088240 on Build GRCh38
rs1039851936 Chr.3: 53088451-53088451 on Build GRCh38
rs191315221 Chr.3: 53088227-53088227 on Build GRCh38
rs142330220 Chr.3: 53088514-53088514 on Build GRCh38
rs933523810 Chr.3: 53088199-53088199 on Build GRCh38
rs1002738171 Chr.3: 53088312-53088312 on Build GRCh38
rs764098415 Chr.3: 53088419-53088419 on Build GRCh38
rs2581783 Chr.3: 53088242-53088242 on Build GRCh38
rs964822817 Chr.3: 53088150-53088150 on Build GRCh38
rs77145844 Chr.3: 53088364-53088364 on Build GRCh38
rs908158217 Chr.3: 53088217-53088217 on Build GRCh38
rs776308285 Chr.3: 53088317-53088317 on Build GRCh38
rs892591770 Chr.3: 53088487-53088487 on Build GRCh38
rs900014416 Chr.3: 53088457-53088457 on Build GRCh38
rs963214351 Chr.3: 53088561-53088561 on Build GRCh38
rs2581784 Chr.3: 53088354-53088354 on Build GRCh38
rs916566285 Chr.3: 53088560-53088560 on Build GRCh38
rs1024889990 Chr.3: 53088293-53088293 on Build GRCh38
rs1030065768 Chr.3: 53088410-53088410 on Build GRCh38
rs907029082 Chr.3: 53088296-53088296 on Build GRCh38
rs944314862 Chr.3: 53088434-53088434 on Build GRCh38
rs974898449 Chr.3: 53088153-53088153 on Build GRCh38
rs973161311 Chr.3: 53088366-53088366 on Build GRCh38
rs879844698 Chr.3: 53088241-53088242 on Build GRCh38
rs527944970 Chr.3: 53088189-53088189 on Build GRCh38
rs1008390811 Chr.3: 53088181-53088181 on Build GRCh38
rs923429511 Chr.3: 53088180-53088180 on Build GRCh38
rs189410181 Chr.3: 53088328-53088328 on Build GRCh38
rs1009516449 Chr.3: 53088501-53088501 on Build GRCh38
rs914295605 Chr.3: 53088149-53088149 on Build GRCh38
rs71615889 Chr.3: 53088241-53088242 on Build GRCh38
rs886058708 Chr.3: 53088561-53088561 on Build GRCh38
rs1025426277 Chr.3: 53088532-53088532 on Build GRCh38
rs537972804 Chr.3: 53088245-53088245 on Build GRCh38
rs956715170 Chr.3: 53088251-53088251 on Build GRCh38
rs747635315 Chr.3: 53088353-53088353 on Build GRCh38
rs1012750139 Chr.3: 53088253-53088253 on Build GRCh38
rs184538568 Chr.3: 53088265-53088265 on Build GRCh38
rs1051465160 Chr.3: 53088412-53088412 on Build GRCh38
rs970652407 Chr.3: 53088535-53088535 on Build GRCh38
rs942285156 Chr.3: 53088546-53088546 on Build GRCh38
rs762743158 Chr.3: 53088371-53088371 on Build GRCh38
rs527683198 Chr.3: 53088550-53088550 on Build GRCh38
rs147750853 Chr.3: 53088362-53088362 on Build GRCh38
rs1033489111 Chr.3: 53088551-53088551 on Build GRCh38
rs186703235 Chr.3: 53088218-53088218 on Build GRCh38
rs542830987 Chr.3: 53088375-53088375 on Build GRCh38
rs1031576220 Chr.3: 53088334-53088334 on Build GRCh38
rs918891226 Chr.3: 53088372-53088372 on Build GRCh38
rs908106084 Chr.3: 53088528-53088528 on Build GRCh38
rs1053802963 Chr.3: 53088477-53088477 on Build GRCh38
rs185163052 Chr.3: 53088408-53088408 on Build GRCh38

View Full Product Details