Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 41027152-41027653 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SCMH1
Entrez Gene ID
22955
Gene Name
sex comb on midleg homolog 1 (Drosophila)
Gene Aliases
Scml3
UniGene
Hs.571874
Gene Chromosome Location
Chr.1: 41027199-41242155 on Build GRCh38
Transcripts
NM_001031694.2 NM_001172218.1 NM_001172219.1 NM_001172220.1 NM_001172221.1 NM_001172222.2 NM_012236.3 NR_135092.1 XM_006710462.2 XM_006710464.1 XM_011541032.2 XM_011541033.2 XM_011541034.1 XM_011541035.2 XM_011541036.2 XM_011541039.1 XM_011541043.2 XM_011541044.1 XM_011541045.2 XM_017000698.1 XM_017000699.1 XM_017000700.1 XM_017000701.1 XM_017000702.1 XM_017000703.1 XM_017000704.1 XM_017000705.1 XM_017000706.1 XM_017000707.1 XM_017000708.1 XM_017000709.1 XM_017000710.1 XM_017000711.1 XM_017000712.1 XM_017000713.1 NR_037868.1

Gene Symbol
SLFNL1-AS1
Entrez Gene ID
100507178
Gene Name
SLFNL1 antisense RNA 1
Gene Aliases
-
UniGene
Hs.660056
Gene Chromosome Location
Chr.1: 41014590-41043890 on Build GRCh38
Transcripts
NM_001031694.2 NM_001172218.1 NM_001172219.1 NM_001172220.1 NM_001172221.1 NM_001172222.2 NM_012236.3 NR_135092.1 XM_006710462.2 XM_006710464.1 XM_011541032.2 XM_011541033.2 XM_011541034.1 XM_011541035.2 XM_011541036.2 XM_011541039.1 XM_011541043.2 XM_011541044.1 XM_011541045.2 XM_017000698.1 XM_017000699.1 XM_017000700.1 XM_017000701.1 XM_017000702.1 XM_017000703.1 XM_017000704.1 XM_017000705.1 XM_017000706.1 XM_017000707.1 XM_017000708.1 XM_017000709.1 XM_017000710.1 XM_017000711.1 XM_017000712.1 XM_017000713.1 NR_037868.1

SNP Detail

SNP ID Chromosome Location
rs570524309 Chr.1: 41027259-41027259 on Build GRCh38
rs762810500 Chr.1: 41027308-41027308 on Build GRCh38
rs971016442 Chr.1: 41027533-41027533 on Build GRCh38
rs997732119 Chr.1: 41027413-41027413 on Build GRCh38
rs3157 Chr.1: 41027439-41027439 on Build GRCh38
rs41312034 Chr.1: 41027473-41027473 on Build GRCh38
rs774109137 Chr.1: 41027360-41027360 on Build GRCh38
rs979819696 Chr.1: 41027210-41027210 on Build GRCh38
rs535012436 Chr.1: 41027478-41027478 on Build GRCh38
rs956937779 Chr.1: 41027256-41027256 on Build GRCh38
rs773074027 Chr.1: 41027236-41027236 on Build GRCh38
rs937080650 Chr.1: 41027586-41027586 on Build GRCh38
rs74071122 Chr.1: 41027260-41027260 on Build GRCh38
rs953569802 Chr.1: 41027466-41027466 on Build GRCh38
rs557712612 Chr.1: 41027482-41027482 on Build GRCh38
rs1040483885 Chr.1: 41027433-41027433 on Build GRCh38
rs1016802314 Chr.1: 41027477-41027477 on Build GRCh38
rs958522015 Chr.1: 41027242-41027242 on Build GRCh38
rs553603863 Chr.1: 41027322-41027322 on Build GRCh38
rs1031293886 Chr.1: 41027563-41027563 on Build GRCh38
rs142406364 Chr.1: 41027356-41027356 on Build GRCh38
rs918080391 Chr.1: 41027295-41027295 on Build GRCh38
rs545631613 Chr.1: 41027534-41027534 on Build GRCh38
rs910764035 Chr.1: 41027613-41027613 on Build GRCh38
rs1005411816 Chr.1: 41027232-41027232 on Build GRCh38
rs988101644 Chr.1: 41027522-41027524 on Build GRCh38
rs990370841 Chr.1: 41027378-41027378 on Build GRCh38
rs541264907 Chr.1: 41027522-41027522 on Build GRCh38
rs537141399 Chr.1: 41027525-41027525 on Build GRCh38
rs771048478 Chr.1: 41027532-41027532 on Build GRCh38
rs957676277 Chr.1: 41027573-41027573 on Build GRCh38
rs990814617 Chr.1: 41027587-41027587 on Build GRCh38
rs750832140 Chr.1: 41027610-41027610 on Build GRCh38
rs769744584 Chr.1: 41027219-41027219 on Build GRCh38
rs887047968 Chr.1: 41027464-41027464 on Build GRCh38
rs943551391 Chr.1: 41027617-41027617 on Build GRCh38
rs573522531 Chr.1: 41027355-41027355 on Build GRCh38
rs953503819 Chr.1: 41027208-41027208 on Build GRCh38
rs557111906 Chr.1: 41027207-41027207 on Build GRCh38
rs971042799 Chr.1: 41027323-41027323 on Build GRCh38
rs977033748 Chr.1: 41027327-41027327 on Build GRCh38
rs559264930 Chr.1: 41027602-41027602 on Build GRCh38
rs923378211 Chr.1: 41027626-41027626 on Build GRCh38
rs1023974658 Chr.1: 41027278-41027278 on Build GRCh38
rs901904399 Chr.1: 41027434-41027434 on Build GRCh38
rs373712988 Chr.1: 41027387-41027387 on Build GRCh38
rs1036240987 Chr.1: 41027393-41027393 on Build GRCh38
rs976218414 Chr.1: 41027618-41027618 on Build GRCh38
rs910701011 Chr.1: 41027382-41027382 on Build GRCh38
rs1047746865 Chr.1: 41027443-41027443 on Build GRCh38
rs190968896 Chr.1: 41027184-41027184 on Build GRCh38
rs572152477 Chr.1: 41027498-41027498 on Build GRCh38
rs901770417 Chr.1: 41027401-41027401 on Build GRCh38
rs1012715410 Chr.1: 41027484-41027484 on Build GRCh38

View Full Product Details