Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 148990008-148990503 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SH3TC2
Entrez Gene ID
79628
Gene Name
SH3 domain and tetratricopeptide repeats 2
Gene Aliases
CMT4C, MNMN
UniGene
Hs.483784
Gene Chromosome Location
Chr.5: 148982150-149063174 on Build GRCh38
Transcripts
NM_024577.3

SNP Detail

SNP ID Chromosome Location
rs985375497 Chr.5: 148990463-148990463 on Build GRCh38
rs919585509 Chr.5: 148990438-148990438 on Build GRCh38
rs992706954 Chr.5: 148990395-148990395 on Build GRCh38
rs997281074 Chr.5: 148990243-148990243 on Build GRCh38
rs958312779 Chr.5: 148990297-148990297 on Build GRCh38
rs10043086 Chr.5: 148990315-148990315 on Build GRCh38
rs575838244 Chr.5: 148990159-148990159 on Build GRCh38
rs949454263 Chr.5: 148990060-148990060 on Build GRCh38
rs1002120470 Chr.5: 148990139-148990139 on Build GRCh38
rs577465518 Chr.5: 148990282-148990282 on Build GRCh38
rs557486021 Chr.5: 148990033-148990033 on Build GRCh38
rs776282002 Chr.5: 148990230-148990230 on Build GRCh38
rs758906397 Chr.5: 148990261-148990261 on Build GRCh38
rs949531167 Chr.5: 148990327-148990327 on Build GRCh38
rs772828814 Chr.5: 148990400-148990400 on Build GRCh38
rs372234197 Chr.5: 148990253-148990253 on Build GRCh38
rs540922872 Chr.5: 148990328-148990328 on Build GRCh38
rs1015372895 Chr.5: 148990398-148990398 on Build GRCh38
rs762925324 Chr.5: 148990396-148990396 on Build GRCh38
rs775243177 Chr.5: 148990383-148990383 on Build GRCh38
rs557514393 Chr.5: 148990150-148990150 on Build GRCh38
rs960924989 Chr.5: 148990381-148990381 on Build GRCh38
rs926675230 Chr.5: 148990423-148990423 on Build GRCh38
rs912444172 Chr.5: 148990472-148990472 on Build GRCh38
rs973721788 Chr.5: 148990223-148990223 on Build GRCh38
rs1033805572 Chr.5: 148990100-148990100 on Build GRCh38
rs973775975 Chr.5: 148990417-148990417 on Build GRCh38
rs143623684 Chr.5: 148990300-148990300 on Build GRCh38
rs1030064403 Chr.5: 148990252-148990252 on Build GRCh38
rs1046467370 Chr.5: 148990340-148990340 on Build GRCh38
rs886060118 Chr.5: 148990083-148990083 on Build GRCh38
rs770560130 Chr.5: 148990181-148990181 on Build GRCh38
rs552475810 Chr.5: 148990175-148990175 on Build GRCh38
rs564255234 Chr.5: 148990231-148990231 on Build GRCh38
rs1052607507 Chr.5: 148990049-148990049 on Build GRCh38
rs1048385739 Chr.5: 148990456-148990456 on Build GRCh38
rs368005281 Chr.5: 148990234-148990234 on Build GRCh38
rs914013985 Chr.5: 148990056-148990056 on Build GRCh38
rs1006504735 Chr.5: 148990164-148990164 on Build GRCh38
rs939715353 Chr.5: 148990203-148990203 on Build GRCh38
rs1046434773 Chr.5: 148990076-148990076 on Build GRCh38
rs919867514 Chr.5: 148990224-148990224 on Build GRCh38
rs985761637 Chr.5: 148990270-148990270 on Build GRCh38
rs938045271 Chr.5: 148990439-148990439 on Build GRCh38
rs5872106 Chr.5: 148990108-148990109 on Build GRCh38
rs17795097 Chr.5: 148990090-148990090 on Build GRCh38
rs17109198 Chr.5: 148990149-148990149 on Build GRCh38

View Full Product Details