Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 96323979-96324487 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC35D2
Entrez Gene ID
11046
Gene Name
solute carrier family 35 member D2
Gene Aliases
HFRC1, SQV7L, UGTrel8, hfrc
UniGene
Hs.494556
Gene Chromosome Location
Chr.9: 96313437-96383710 on Build GRCh38
Transcripts
NM_001286990.1 NM_007001.2 NR_104627.1 XM_005251673.1 XM_005251674.1 XM_005251675.2 XM_005251676.1 XM_005251678.4 XM_006716939.2 XM_011518164.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1111596 c.806C>T p.T269M Chr.9: 96324116-96324116 on Build GRCh38
1464248 c.800C>T p.A267V Chr.9: 96324122-96324122 on Build GRCh38
4772440 c.827_828delTC p.I276fs*48 Chr.9: 96324094-96324095 on Build GRCh38
5653297 c.824C>G p.A275G Chr.9: 96324098-96324098 on Build GRCh38
3927875 c.806C>A p.T269K Chr.9: 96324116-96324116 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs756967899 Chr.9: 96324130-96324130 on Build GRCh38
rs78220385 Chr.9: 96324168-96324168 on Build GRCh38
rs1008541070 Chr.9: 96324246-96324246 on Build GRCh38
rs7026081 Chr.9: 96324054-96324054 on Build GRCh38
rs186139409 Chr.9: 96324105-96324105 on Build GRCh38
rs186438845 Chr.9: 96324013-96324013 on Build GRCh38
rs949989533 Chr.9: 96324397-96324397 on Build GRCh38
rs539084624 Chr.9: 96324211-96324211 on Build GRCh38
rs373785289 Chr.9: 96324416-96324416 on Build GRCh38
rs183282247 Chr.9: 96324189-96324189 on Build GRCh38
rs145858517 Chr.9: 96324148-96324148 on Build GRCh38
rs7038558 Chr.9: 96324268-96324268 on Build GRCh38
rs557792513 Chr.9: 96324212-96324212 on Build GRCh38
rs145290742 Chr.9: 96324116-96324116 on Build GRCh38
rs749434366 Chr.9: 96324184-96324184 on Build GRCh38
rs377214875 Chr.9: 96324061-96324061 on Build GRCh38
rs749109859 Chr.9: 96324162-96324162 on Build GRCh38
rs891952567 Chr.9: 96324170-96324170 on Build GRCh38
rs564019171 Chr.9: 96324390-96324390 on Build GRCh38
rs780177959 Chr.9: 96324004-96324004 on Build GRCh38
rs960082916 Chr.9: 96324375-96324375 on Build GRCh38
rs987139582 Chr.9: 96324018-96324018 on Build GRCh38
rs1054946168 Chr.9: 96324423-96324423 on Build GRCh38
rs73534915 Chr.9: 96324319-96324319 on Build GRCh38
rs771199477 Chr.9: 96324063-96324063 on Build GRCh38
rs753448649 Chr.9: 96324128-96324128 on Build GRCh38
rs775052556 Chr.9: 96324364-96324364 on Build GRCh38
rs147730425 Chr.9: 96324255-96324255 on Build GRCh38
rs572556943 Chr.9: 96324329-96324329 on Build GRCh38
rs1051288140 Chr.9: 96324456-96324456 on Build GRCh38
rs371994293 Chr.9: 96324123-96324123 on Build GRCh38
rs181735925 Chr.9: 96324003-96324003 on Build GRCh38
rs905932359 Chr.9: 96324149-96324149 on Build GRCh38
rs761270064 Chr.9: 96324296-96324296 on Build GRCh38
rs920394548 Chr.9: 96324286-96324286 on Build GRCh38
rs750225475 Chr.9: 96324138-96324138 on Build GRCh38
rs72621424 Chr.9: 96324000-96324000 on Build GRCh38
rs969922064 Chr.9: 96324137-96324137 on Build GRCh38
rs41281898 Chr.9: 96324210-96324210 on Build GRCh38
rs76549121 Chr.9: 96324461-96324461 on Build GRCh38
rs767017118 Chr.9: 96324387-96324387 on Build GRCh38
rs147885801 Chr.9: 96324134-96324134 on Build GRCh38
rs553309549 Chr.9: 96324269-96324269 on Build GRCh38
rs964400199 Chr.9: 96324254-96324254 on Build GRCh38
rs748210789 Chr.9: 96324040-96324040 on Build GRCh38
rs746179418 Chr.9: 96324200-96324200 on Build GRCh38
rs1017301705 Chr.9: 96324250-96324250 on Build GRCh38
rs936038708 Chr.9: 96324420-96324420 on Build GRCh38
rs891881528 Chr.9: 96324436-96324436 on Build GRCh38
rs773417700 Chr.9: 96324058-96324058 on Build GRCh38
rs67992409 Chr.9: 96324440-96324440 on Build GRCh38
rs556879561 Chr.9: 96324019-96324019 on Build GRCh38
rs1044313944 Chr.9: 96324400-96324400 on Build GRCh38
rs200143326 Chr.9: 96324115-96324115 on Build GRCh38
rs780001329 Chr.9: 96324158-96324158 on Build GRCh38
rs920406348 Chr.9: 96324060-96324060 on Build GRCh38
rs35505854 Chr.9: 96324163-96324163 on Build GRCh38
rs777699840 Chr.9: 96324183-96324183 on Build GRCh38
rs747574055 Chr.9: 96324132-96324132 on Build GRCh38
rs371638212 Chr.9: 96324064-96324068 on Build GRCh38
rs530956725 Chr.9: 96324075-96324075 on Build GRCh38
rs999462985 Chr.9: 96324303-96324303 on Build GRCh38
rs78649010 Chr.9: 96324352-96324352 on Build GRCh38
rs115818542 Chr.9: 96324196-96324196 on Build GRCh38

View Full Product Details