Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 141212363-141212855 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC45A4
Entrez Gene ID
57210
Gene Name
solute carrier family 45 member 4
Gene Aliases
-
UniGene
Hs.372492
Gene Chromosome Location
Chr.8: 141207166-141308321 on Build GRCh38
Transcripts
NM_001080431.2 NM_001286646.1 NM_001286648.1 XM_011517169.2 XM_011517170.2 XM_011517171.2 XM_011517172.2 XM_011517173.2 XM_011517174.2 XM_011517176.2 XM_011517177.2 XM_011517178.2 XM_011517179.2 XM_017013648.1 XM_017013649.1 XM_017013650.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7197319 c.2061C>T p.D687D Chr.8: 141212437-141212437 on Build GRCh38
6782502 c.2017A>G p.I673V Chr.8: 141212481-141212481 on Build GRCh38
4532763 c.2070G>A p.G690G Chr.8: 141212428-141212428 on Build GRCh38
5539220 c.2112C>T p.F704F Chr.8: 141212386-141212386 on Build GRCh38
6782466 c.1916G>A p.G639E Chr.8: 141212429-141212429 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs188546735 Chr.8: 141212811-141212811 on Build GRCh38
rs967905338 Chr.8: 141212738-141212738 on Build GRCh38
rs1009372000 Chr.8: 141212688-141212688 on Build GRCh38
rs747487303 Chr.8: 141212384-141212384 on Build GRCh38
rs911351017 Chr.8: 141212796-141212796 on Build GRCh38
rs541739901 Chr.8: 141212617-141212617 on Build GRCh38
rs201350370 Chr.8: 141212532-141212532 on Build GRCh38
rs535903836 Chr.8: 141212436-141212436 on Build GRCh38
rs141399161 Chr.8: 141212434-141212434 on Build GRCh38
rs370725358 Chr.8: 141212470-141212470 on Build GRCh38
rs528274678 Chr.8: 141212610-141212610 on Build GRCh38
rs775831890 Chr.8: 141212412-141212412 on Build GRCh38
rs748389233 Chr.8: 141212587-141212587 on Build GRCh38
rs370119880 Chr.8: 141212417-141212417 on Build GRCh38
rs1012807319 Chr.8: 141212651-141212651 on Build GRCh38
rs974284636 Chr.8: 141212790-141212790 on Build GRCh38
rs1005240559 Chr.8: 141212624-141212625 on Build GRCh38
rs771581638 Chr.8: 141212393-141212393 on Build GRCh38
rs978835334 Chr.8: 141212789-141212790 on Build GRCh38
rs745922068 Chr.8: 141212460-141212460 on Build GRCh38
rs773051311 Chr.8: 141212504-141212504 on Build GRCh38
rs558892917 Chr.8: 141212586-141212586 on Build GRCh38
rs1003248582 Chr.8: 141212667-141212667 on Build GRCh38
rs749394330 Chr.8: 141212579-141212579 on Build GRCh38
rs563777436 Chr.8: 141212695-141212695 on Build GRCh38
rs564741188 Chr.8: 141212756-141212756 on Build GRCh38
rs749469321 Chr.8: 141212497-141212497 on Build GRCh38
rs781708424 Chr.8: 141212562-141212562 on Build GRCh38
rs919773440 Chr.8: 141212801-141212801 on Build GRCh38
rs749986057 Chr.8: 141212723-141212723 on Build GRCh38
rs1007918340 Chr.8: 141212698-141212698 on Build GRCh38
rs764740270 Chr.8: 141212538-141212538 on Build GRCh38
rs759757666 Chr.8: 141212401-141212401 on Build GRCh38
rs368362326 Chr.8: 141212539-141212539 on Build GRCh38
rs778146517 Chr.8: 141212440-141212440 on Build GRCh38
rs930262573 Chr.8: 141212819-141212819 on Build GRCh38
rs904066684 Chr.8: 141212656-141212656 on Build GRCh38
rs543563635 Chr.8: 141212737-141212737 on Build GRCh38
rs956656121 Chr.8: 141212687-141212687 on Build GRCh38
rs757835914 Chr.8: 141212445-141212445 on Build GRCh38
rs560132156 Chr.8: 141212624-141212624 on Build GRCh38
rs547273014 Chr.8: 141212824-141212824 on Build GRCh38
rs1012488607 Chr.8: 141212387-141212387 on Build GRCh38
rs574873462 Chr.8: 141212658-141212658 on Build GRCh38
rs375296684 Chr.8: 141212421-141212421 on Build GRCh38
rs758050138 Chr.8: 141212802-141212802 on Build GRCh38
rs765701630 Chr.8: 141212767-141212767 on Build GRCh38
rs886479038 Chr.8: 141212611-141212611 on Build GRCh38
rs764551113 Chr.8: 141212602-141212602 on Build GRCh38
rs963847264 Chr.8: 141212776-141212776 on Build GRCh38
rs376612191 Chr.8: 141212439-141212439 on Build GRCh38
rs1043915418 Chr.8: 141212652-141212652 on Build GRCh38
rs148067808 Chr.8: 141212419-141212419 on Build GRCh38
rs781775162 Chr.8: 141212446-141212446 on Build GRCh38
rs530924997 Chr.8: 141212716-141212716 on Build GRCh38
rs770111752 Chr.8: 141212409-141212409 on Build GRCh38
rs760498596 Chr.8: 141212521-141212521 on Build GRCh38
rs776499015 Chr.8: 141212530-141212530 on Build GRCh38
rs763722556 Chr.8: 141212541-141212541 on Build GRCh38
rs758855318 Chr.8: 141212833-141212833 on Build GRCh38
rs760647905 Chr.8: 141212420-141212420 on Build GRCh38
rs976788557 Chr.8: 141212807-141212807 on Build GRCh38
rs769953358 Chr.8: 141212461-141212461 on Build GRCh38
rs939410118 Chr.8: 141212612-141212612 on Build GRCh38
rs185173401 Chr.8: 141212580-141212580 on Build GRCh38
rs201879517 Chr.8: 141212526-141212526 on Build GRCh38
rs955084841 Chr.8: 141212437-141212437 on Build GRCh38
rs780169612 Chr.8: 141212578-141212578 on Build GRCh38
rs757747989 Chr.8: 141212556-141212556 on Build GRCh38
rs774188540 Chr.8: 141212624-141212627 on Build GRCh38
rs955075026 Chr.8: 141212686-141212686 on Build GRCh38
rs372268272 Chr.8: 141212551-141212551 on Build GRCh38
rs756700238 Chr.8: 141212575-141212575 on Build GRCh38
rs776838096 Chr.8: 141212397-141212397 on Build GRCh38
rs1035307867 Chr.8: 141212668-141212668 on Build GRCh38
rs75952226 Chr.8: 141212834-141212834 on Build GRCh38
rs374347918 Chr.8: 141212500-141212500 on Build GRCh38
rs999781892 Chr.8: 141212659-141212659 on Build GRCh38
rs947464317 Chr.8: 141212514-141212514 on Build GRCh38
rs754529727 Chr.8: 141212433-141212433 on Build GRCh38
rs546818039 Chr.8: 141212747-141212747 on Build GRCh38
rs1030620307 Chr.8: 141212680-141212680 on Build GRCh38
rs750986966 Chr.8: 141212577-141212586 on Build GRCh38
rs541887543 Chr.8: 141212669-141212669 on Build GRCh38
rs1023369356 Chr.8: 141212633-141212633 on Build GRCh38
rs775435684 Chr.8: 141212476-141212476 on Build GRCh38
rs753310131 Chr.8: 141212424-141212424 on Build GRCh38
rs746309380 Chr.8: 141212574-141212574 on Build GRCh38
rs770670211 Chr.8: 141212605-141212605 on Build GRCh38
rs895299554 Chr.8: 141212637-141212637 on Build GRCh38

View Full Product Details