Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 11147124-11147363 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPC24
Entrez Gene ID
147841
Gene Name
SPC24, NDC80 kinetochore complex component
Gene Aliases
SPBC24
UniGene
Hs.381225
Gene Chromosome Location
Chr.19: 11145494-11155997 on Build GRCh38
Transcripts
NM_001317031.1 NM_001317033.1 NM_182513.3 XM_005259753.3 XM_011527702.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5765626 c.528C>A p.D176E Chr.19: 11147249-11147249 on Build GRCh38
7211445 c.492T>A p.H164Q Chr.19: 11147285-11147285 on Build GRCh38
3821911 c.594G>T p.*198Y Chr.19: 11147183-11147183 on Build GRCh38
5388595 c.546G>A p.R182R Chr.19: 11147231-11147231 on Build GRCh38
5593243 c.522C>T p.H174H Chr.19: 11147255-11147255 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769587754 Chr.19: 11147150-11147150 on Build GRCh38
rs755895844 Chr.19: 11147294-11147294 on Build GRCh38
rs980070711 Chr.19: 11147251-11147251 on Build GRCh38
rs1051489655 Chr.19: 11147212-11147212 on Build GRCh38
rs539671057 Chr.19: 11147173-11147173 on Build GRCh38
rs771083717 Chr.19: 11147148-11147148 on Build GRCh38
rs867220436 Chr.19: 11147239-11147239 on Build GRCh38
rs183836474 Chr.19: 11147172-11147172 on Build GRCh38
rs372332361 Chr.19: 11147333-11147333 on Build GRCh38
rs747613176 Chr.19: 11147317-11147317 on Build GRCh38
rs888892156 Chr.19: 11147225-11147225 on Build GRCh38
rs745657132 Chr.19: 11147163-11147163 on Build GRCh38
rs749342385 Chr.19: 11147146-11147146 on Build GRCh38
rs768601401 Chr.19: 11147315-11147315 on Build GRCh38
rs139615751 Chr.19: 11147316-11147316 on Build GRCh38
rs557924675 Chr.19: 11147175-11147175 on Build GRCh38
rs192507355 Chr.19: 11147164-11147164 on Build GRCh38
rs188013688 Chr.19: 11147191-11147191 on Build GRCh38
rs943003878 Chr.19: 11147265-11147265 on Build GRCh38
rs779141076 Chr.19: 11147149-11147149 on Build GRCh38
rs779072119 Chr.19: 11147281-11147281 on Build GRCh38
rs375986607 Chr.19: 11147192-11147192 on Build GRCh38
rs746764270 Chr.19: 11147314-11147314 on Build GRCh38
rs753821558 Chr.19: 11147318-11147319 on Build GRCh38
rs562200114 Chr.19: 11147219-11147219 on Build GRCh38
rs756722263 Chr.19: 11147245-11147245 on Build GRCh38
rs771902851 Chr.19: 11147288-11147288 on Build GRCh38
rs200206619 Chr.19: 11147189-11147189 on Build GRCh38
rs369672908 Chr.19: 11147236-11147236 on Build GRCh38
rs968027339 Chr.19: 11147247-11147247 on Build GRCh38
rs746090767 Chr.19: 11147283-11147283 on Build GRCh38
rs960809690 Chr.19: 11147208-11147208 on Build GRCh38
rs921554850 Chr.19: 11147269-11147269 on Build GRCh38
rs945651460 Chr.19: 11147176-11147176 on Build GRCh38
rs775465289 Chr.19: 11147171-11147171 on Build GRCh38
rs1037808352 Chr.19: 11147286-11147286 on Build GRCh38
rs778378194 Chr.19: 11147260-11147260 on Build GRCh38
rs749933933 Chr.19: 11147278-11147278 on Build GRCh38

View Full Product Details