Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 134248550-134249061 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
THYN1
Entrez Gene ID
29087
Gene Name
thymocyte nuclear protein 1
Gene Aliases
HSPC144, MDS012, MY105, THY28, THY28KD
UniGene
Hs.13645
Gene Chromosome Location
Chr.11: 134248278-134253401 on Build GRCh38
Transcripts
NM_001037304.1 NM_001037305.1 NM_014174.2 NM_199297.1 NM_199298.1 XM_005271527.3 XM_005271528.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4803672 c.546T>G p.H182Q Chr.11: 134248894-134248894 on Build GRCh38
6345228 c.527C>A p.A176D Chr.11: 134248913-134248913 on Build GRCh38
3415701 c.507G>T p.M169I Chr.11: 134248933-134248933 on Build GRCh38
7180820 c.481-1G>A p.? Chr.11: 134248960-134248960 on Build GRCh38
466589 c.617A>G p.Q206R Chr.11: 134248823-134248823 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1029038727 Chr.11: 134248747-134248748 on Build GRCh38
rs1003935384 Chr.11: 134248876-134248876 on Build GRCh38
rs542710089 Chr.11: 134248929-134248929 on Build GRCh38
rs747087764 Chr.11: 134248984-134248987 on Build GRCh38
rs141146037 Chr.11: 134248644-134248644 on Build GRCh38
rs3816440 Chr.11: 134248640-134248640 on Build GRCh38
rs939087045 Chr.11: 134248986-134248986 on Build GRCh38
rs765074945 Chr.11: 134248905-134248905 on Build GRCh38
rs368822230 Chr.11: 134248992-134248992 on Build GRCh38
rs758994402 Chr.11: 134248598-134248598 on Build GRCh38
rs753894768 Chr.11: 134248933-134248933 on Build GRCh38
rs200675658 Chr.11: 134248759-134248759 on Build GRCh38
rs779530492 Chr.11: 134248842-134248842 on Build GRCh38
rs749403330 Chr.11: 134248840-134248840 on Build GRCh38
rs960857412 Chr.11: 134248639-134248639 on Build GRCh38
rs781269466 Chr.11: 134248811-134248811 on Build GRCh38
rs561446670 Chr.11: 134248940-134248940 on Build GRCh38
rs112958221 Chr.11: 134248827-134248827 on Build GRCh38
rs372783800 Chr.11: 134248980-134248980 on Build GRCh38
rs778742640 Chr.11: 134248843-134248844 on Build GRCh38
rs1029861807 Chr.11: 134248573-134248573 on Build GRCh38
rs936199406 Chr.11: 134248794-134248794 on Build GRCh38
rs778011490 Chr.11: 134248932-134248932 on Build GRCh38
rs1040368542 Chr.11: 134248818-134248818 on Build GRCh38
rs1033482564 Chr.11: 134248624-134248624 on Build GRCh38
rs375504006 Chr.11: 134248899-134248899 on Build GRCh38
rs765257186 Chr.11: 134248785-134248785 on Build GRCh38
rs750159540 Chr.11: 134248792-134248792 on Build GRCh38
rs758344503 Chr.11: 134248891-134248892 on Build GRCh38
rs946016730 Chr.11: 134248750-134248750 on Build GRCh38
rs763679759 Chr.11: 134248835-134248836 on Build GRCh38
rs777781079 Chr.11: 134248836-134248836 on Build GRCh38
rs548126995 Chr.11: 134249027-134249027 on Build GRCh38
rs758850568 Chr.11: 134248950-134248950 on Build GRCh38
rs750444928 Chr.11: 134248908-134248908 on Build GRCh38
rs781036754 Chr.11: 134248817-134248817 on Build GRCh38
rs199857469 Chr.11: 134248944-134248944 on Build GRCh38
rs760350363 Chr.11: 134248864-134248864 on Build GRCh38
rs189393672 Chr.11: 134248819-134248819 on Build GRCh38
rs201261788 Chr.11: 134248928-134248928 on Build GRCh38
rs886774163 Chr.11: 134248585-134248585 on Build GRCh38
rs766109535 Chr.11: 134248922-134248922 on Build GRCh38
rs1000160445 Chr.11: 134248688-134248689 on Build GRCh38
rs193129683 Chr.11: 134248963-134248963 on Build GRCh38
rs898173742 Chr.11: 134248948-134248948 on Build GRCh38
rs977391403 Chr.11: 134248780-134248780 on Build GRCh38
rs779899925 Chr.11: 134248893-134248894 on Build GRCh38
rs756346064 Chr.11: 134248820-134248820 on Build GRCh38
rs774102257 Chr.11: 134249007-134249007 on Build GRCh38
rs753416467 Chr.11: 134248781-134248781 on Build GRCh38
rs1010427158 Chr.11: 134248620-134248620 on Build GRCh38
rs780396096 Chr.11: 134248675-134248675 on Build GRCh38
rs536247045 Chr.11: 134249031-134249031 on Build GRCh38
rs752148066 Chr.11: 134248736-134248736 on Build GRCh38
rs540952909 Chr.11: 134248892-134248892 on Build GRCh38
rs776010722 Chr.11: 134248856-134248856 on Build GRCh38
rs755612333 Chr.11: 134248817-134248818 on Build GRCh38
rs762806982 Chr.11: 134248916-134248916 on Build GRCh38
rs904787294 Chr.11: 134249029-134249029 on Build GRCh38
rs745646426 Chr.11: 134248892-134248894 on Build GRCh38
rs147965930 Chr.11: 134249028-134249028 on Build GRCh38
rs200017797 Chr.11: 134248849-134248849 on Build GRCh38
rs926370507 Chr.11: 134248574-134248574 on Build GRCh38
rs780522398 Chr.11: 134248962-134248962 on Build GRCh38
rs771312843 Chr.11: 134248841-134248841 on Build GRCh38
rs758274347 Chr.11: 134248799-134248799 on Build GRCh38
rs775348452 Chr.11: 134248771-134248771 on Build GRCh38
rs763640365 Chr.11: 134248776-134248776 on Build GRCh38
rs753572499 Chr.11: 134248838-134248839 on Build GRCh38
rs1057487415 Chr.11: 134248597-134248597 on Build GRCh38
rs202070344 Chr.11: 134248896-134248896 on Build GRCh38
rs184515474 Chr.11: 134248656-134248656 on Build GRCh38
rs962577001 Chr.11: 134248601-134248601 on Build GRCh38
rs764294451 Chr.11: 134249011-134249011 on Build GRCh38
rs11223733 Chr.11: 134248694-134248694 on Build GRCh38
rs1002116904 Chr.11: 134248951-134248951 on Build GRCh38
rs369864471 Chr.11: 134248979-134248979 on Build GRCh38
rs376005049 Chr.11: 134248991-134248991 on Build GRCh38
rs917344291 Chr.11: 134248741-134248741 on Build GRCh38
rs369908466 Chr.11: 134248761-134248761 on Build GRCh38
rs532841374 Chr.11: 134249009-134249009 on Build GRCh38
rs768450231 Chr.11: 134248878-134248878 on Build GRCh38
rs372522779 Chr.11: 134248995-134248995 on Build GRCh38
rs766335101 Chr.11: 134248997-134248997 on Build GRCh38
rs1003860200 Chr.11: 134248586-134248586 on Build GRCh38
rs981097725 Chr.11: 134248608-134248608 on Build GRCh38
rs761611526 Chr.11: 134248782-134248782 on Build GRCh38
rs558368911 Chr.11: 134248846-134248846 on Build GRCh38
rs925962058 Chr.11: 134248783-134248783 on Build GRCh38
rs756825520 Chr.11: 134248841-134248842 on Build GRCh38

View Full Product Details