Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 26972971-26973521 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TM7SF3
Entrez Gene ID
51768
Gene Name
transmembrane 7 superfamily member 3
Gene Aliases
-
UniGene
Hs.438641
Gene Chromosome Location
Chr.12: 26966519-27014427 on Build GRCh38
Transcripts
NM_016551.2 XM_005253391.3 XM_011520690.2 XM_017019463.1 XM_017019464.1 XM_017019465.1 XM_017019466.1

SNP Detail

SNP ID Chromosome Location
rs201631165 Chr.12: 26973084-26973085 on Build GRCh38
rs974109296 Chr.12: 26973216-26973216 on Build GRCh38
rs779189556 Chr.12: 26973091-26973094 on Build GRCh38
rs998912506 Chr.12: 26973073-26973073 on Build GRCh38
rs925059683 Chr.12: 26973298-26973298 on Build GRCh38
rs1032192023 Chr.12: 26973091-26973091 on Build GRCh38
rs1127782 Chr.12: 26973263-26973263 on Build GRCh38
rs963997741 Chr.12: 26973174-26973174 on Build GRCh38
rs116479892 Chr.12: 26973006-26973006 on Build GRCh38
rs997812428 Chr.12: 26973123-26973123 on Build GRCh38
rs540967603 Chr.12: 26973109-26973109 on Build GRCh38
rs997464992 Chr.12: 26973340-26973340 on Build GRCh38
rs187394514 Chr.12: 26973051-26973051 on Build GRCh38
rs746682140 Chr.12: 26973472-26973472 on Build GRCh38
rs200654752 Chr.12: 26973190-26973190 on Build GRCh38
rs1013090019 Chr.12: 26973327-26973327 on Build GRCh38
rs1055181696 Chr.12: 26973001-26973001 on Build GRCh38
rs933666658 Chr.12: 26973236-26973236 on Build GRCh38
rs957712782 Chr.12: 26973383-26973383 on Build GRCh38
rs1056648136 Chr.12: 26973008-26973008 on Build GRCh38
rs971380181 Chr.12: 26973020-26973020 on Build GRCh38
rs5797207 Chr.12: 26973175-26973176 on Build GRCh38
rs115604315 Chr.12: 26973179-26973179 on Build GRCh38
rs200227032 Chr.12: 26973178-26973179 on Build GRCh38
rs779433569 Chr.12: 26973124-26973124 on Build GRCh38
rs1042145691 Chr.12: 26973074-26973074 on Build GRCh38
rs1016019139 Chr.12: 26973466-26973466 on Build GRCh38
rs941519413 Chr.12: 26973285-26973285 on Build GRCh38
rs772444150 Chr.12: 26973366-26973366 on Build GRCh38
rs926588070 Chr.12: 26973230-26973230 on Build GRCh38
rs558941933 Chr.12: 26973197-26973197 on Build GRCh38
rs900421426 Chr.12: 26973118-26973118 on Build GRCh38
rs536363479 Chr.12: 26973179-26973180 on Build GRCh38
rs907189756 Chr.12: 26973346-26973346 on Build GRCh38
rs562364385 Chr.12: 26973131-26973131 on Build GRCh38
rs945130996 Chr.12: 26973029-26973031 on Build GRCh38
rs548786385 Chr.12: 26973019-26973022 on Build GRCh38
rs989751776 Chr.12: 26973161-26973161 on Build GRCh38
rs886314610 Chr.12: 26973388-26973388 on Build GRCh38
rs116540653 Chr.12: 26973335-26973335 on Build GRCh38
rs529513989 Chr.12: 26973169-26973169 on Build GRCh38
rs528181915 Chr.12: 26973073-26973074 on Build GRCh38
rs546539452 Chr.12: 26973372-26973372 on Build GRCh38
rs1042200008 Chr.12: 26973277-26973277 on Build GRCh38
rs975333442 Chr.12: 26973198-26973198 on Build GRCh38
rs1038542281 Chr.12: 26973310-26973310 on Build GRCh38
rs201947616 Chr.12: 26973192-26973192 on Build GRCh38
rs963080527 Chr.12: 26973206-26973207 on Build GRCh38
rs533486599 Chr.12: 26973245-26973245 on Build GRCh38
rs879932743 Chr.12: 26973116-26973116 on Build GRCh38
rs894722551 Chr.12: 26973321-26973321 on Build GRCh38
rs534502624 Chr.12: 26973367-26973367 on Build GRCh38
rs531377930 Chr.12: 26973023-26973023 on Build GRCh38
rs397743639 Chr.12: 26973189-26973190 on Build GRCh38
rs756113266 Chr.12: 26973085-26973088 on Build GRCh38
rs147991243 Chr.12: 26973237-26973237 on Build GRCh38
rs922222389 Chr.12: 26973202-26973202 on Build GRCh38
rs1051625705 Chr.12: 26973386-26973386 on Build GRCh38
rs1000533563 Chr.12: 26973093-26973093 on Build GRCh38
rs918117241 Chr.12: 26973241-26973241 on Build GRCh38
rs999275159 Chr.12: 26973360-26973360 on Build GRCh38
rs1038707213 Chr.12: 26973311-26973311 on Build GRCh38
rs1022173326 Chr.12: 26973422-26973422 on Build GRCh38
rs959668665 Chr.12: 26973232-26973232 on Build GRCh38
rs771884448 Chr.12: 26973389-26973389 on Build GRCh38
rs568349459 Chr.12: 26973439-26973439 on Build GRCh38
rs573454135 Chr.12: 26973090-26973090 on Build GRCh38
rs372660402 Chr.12: 26973412-26973412 on Build GRCh38

View Full Product Details