Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 39666035-39666530 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM178A
Entrez Gene ID
130733
Gene Name
transmembrane protein 178A
Gene Aliases
TMEM178
UniGene
Hs.40808
Gene Chromosome Location
Chr.2: 39665087-39717963 on Build GRCh38
Transcripts
NM_001167959.1 NM_152390.2 XM_005264144.1 XM_005264145.1 XM_017003369.1 XM_017003370.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5615081 c.74C>A p.A25D Chr.2: 39666048-39666048 on Build GRCh38
5643385 c.250C>T p.R84C Chr.2: 39666224-39666224 on Build GRCh38
5692030 c.167A>G p.K56R Chr.2: 39666141-39666141 on Build GRCh38
1614845 c.252C>T p.R84R Chr.2: 39666226-39666226 on Build GRCh38
1668864 c.113G>T p.R38L Chr.2: 39666087-39666087 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs909435934 Chr.2: 39666395-39666395 on Build GRCh38
rs755453696 Chr.2: 39666080-39666080 on Build GRCh38
rs372463281 Chr.2: 39666298-39666298 on Build GRCh38
rs17852679 Chr.2: 39666293-39666293 on Build GRCh38
rs942206142 Chr.2: 39666403-39666403 on Build GRCh38
rs992200047 Chr.2: 39666076-39666076 on Build GRCh38
rs973589814 Chr.2: 39666447-39666447 on Build GRCh38
rs1018422610 Chr.2: 39666145-39666145 on Build GRCh38
rs768483450 Chr.2: 39666291-39666291 on Build GRCh38
rs909976512 Chr.2: 39666507-39666507 on Build GRCh38
rs919411275 Chr.2: 39666460-39666460 on Build GRCh38
rs866283035 Chr.2: 39666334-39666334 on Build GRCh38
rs778007609 Chr.2: 39666095-39666095 on Build GRCh38
rs201650119 Chr.2: 39666364-39666364 on Build GRCh38
rs1037864024 Chr.2: 39666183-39666183 on Build GRCh38
rs867303091 Chr.2: 39666312-39666312 on Build GRCh38
rs927957182 Chr.2: 39666483-39666483 on Build GRCh38
rs756440294 Chr.2: 39666094-39666094 on Build GRCh38
rs764798692 Chr.2: 39666176-39666176 on Build GRCh38
rs377194329 Chr.2: 39666355-39666355 on Build GRCh38
rs561534498 Chr.2: 39666107-39666107 on Build GRCh38
rs986288603 Chr.2: 39666424-39666424 on Build GRCh38
rs530068371 Chr.2: 39666435-39666435 on Build GRCh38
rs572019351 Chr.2: 39666214-39666214 on Build GRCh38
rs866895590 Chr.2: 39666309-39666309 on Build GRCh38
rs966029736 Chr.2: 39666454-39666454 on Build GRCh38
rs540455776 Chr.2: 39666262-39666262 on Build GRCh38
rs867522587 Chr.2: 39666135-39666135 on Build GRCh38
rs967291876 Chr.2: 39666384-39666384 on Build GRCh38
rs918827360 Chr.2: 39666468-39666468 on Build GRCh38
rs776324570 Chr.2: 39666163-39666163 on Build GRCh38
rs761016291 Chr.2: 39666358-39666358 on Build GRCh38
rs772742698 Chr.2: 39666352-39666352 on Build GRCh38
rs764285669 Chr.2: 39666247-39666247 on Build GRCh38
rs773111806 Chr.2: 39666229-39666229 on Build GRCh38
rs746856012 Chr.2: 39666288-39666288 on Build GRCh38
rs779936684 Chr.2: 39666283-39666283 on Build GRCh38
rs1002717806 Chr.2: 39666265-39666265 on Build GRCh38
rs1030426303 Chr.2: 39666412-39666412 on Build GRCh38
rs981003584 Chr.2: 39666387-39666387 on Build GRCh38
rs754090258 Chr.2: 39666259-39666259 on Build GRCh38
rs963118620 Chr.2: 39666438-39666438 on Build GRCh38
rs758160308 Chr.2: 39666062-39666063 on Build GRCh38
rs768008771 Chr.2: 39666082-39666082 on Build GRCh38
rs752108827 Chr.2: 39666075-39666075 on Build GRCh38
rs745816255 Chr.2: 39666117-39666117 on Build GRCh38
rs929556482 Chr.2: 39666466-39666466 on Build GRCh38
rs750456040 Chr.2: 39666268-39666268 on Build GRCh38
rs200656944 Chr.2: 39666165-39666165 on Build GRCh38
rs866442443 Chr.2: 39666290-39666290 on Build GRCh38
rs747840220 Chr.2: 39666299-39666299 on Build GRCh38
rs573490688 Chr.2: 39666127-39666127 on Build GRCh38
rs930001223 Chr.2: 39666472-39666472 on Build GRCh38
rs749363566 Chr.2: 39666103-39666103 on Build GRCh38
rs948125005 Chr.2: 39666508-39666508 on Build GRCh38
rs879562620 Chr.2: 39666230-39666230 on Build GRCh38
rs768234209 Chr.2: 39666137-39666137 on Build GRCh38
rs772869428 Chr.2: 39666313-39666313 on Build GRCh38
rs977032988 Chr.2: 39666467-39666467 on Build GRCh38
rs778771460 Chr.2: 39666111-39666111 on Build GRCh38
rs879640592 Chr.2: 39666475-39666475 on Build GRCh38
rs984114225 Chr.2: 39666498-39666498 on Build GRCh38
rs762509248 Chr.2: 39666333-39666333 on Build GRCh38
rs754096822 Chr.2: 39666383-39666383 on Build GRCh38
rs753047006 Chr.2: 39666083-39666083 on Build GRCh38
rs769270706 Chr.2: 39666310-39666310 on Build GRCh38
rs772779720 Chr.2: 39666206-39666206 on Build GRCh38
rs766835827 Chr.2: 39666071-39666071 on Build GRCh38
rs771927547 Chr.2: 39666122-39666122 on Build GRCh38
rs868530808 Chr.2: 39666104-39666104 on Build GRCh38
rs954382179 Chr.2: 39666413-39666413 on Build GRCh38
rs779066424 Chr.2: 39666263-39666263 on Build GRCh38
rs761065168 Chr.2: 39666233-39666233 on Build GRCh38
rs998717537 Chr.2: 39666397-39666397 on Build GRCh38
rs542249675 Chr.2: 39666471-39666471 on Build GRCh38
rs761363620 Chr.2: 39666070-39666070 on Build GRCh38

View Full Product Details