Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 132757217-132757552 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM71
Entrez Gene ID
137835
Gene Name
transmembrane protein 71
Gene Aliases
-
UniGene
Hs.293842
Gene Chromosome Location
Chr.8: 132709945-132760668 on Build GRCh38
Transcripts
NM_001145153.1 NM_144649.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7183184 c.96C>T p.F32F Chr.8: 132757239-132757239 on Build GRCh38
3645229 c.54G>A p.L18L Chr.8: 132757281-132757281 on Build GRCh38
3645230 c.54G>A p.L18L Chr.8: 132757281-132757281 on Build GRCh38
4912810 c.86C>T p.T29M Chr.8: 132757249-132757249 on Build GRCh38
4912811 c.86C>T p.T29M Chr.8: 132757249-132757249 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1014921753 Chr.8: 132757249-132757249 on Build GRCh38
rs145273992 Chr.8: 132757284-132757284 on Build GRCh38
rs11778229 Chr.8: 132757379-132757379 on Build GRCh38
rs979290488 Chr.8: 132757523-132757523 on Build GRCh38
rs746973011 Chr.8: 132757294-132757294 on Build GRCh38
rs73708367 Chr.8: 132757492-132757492 on Build GRCh38
rs540674121 Chr.8: 132757395-132757395 on Build GRCh38
rs778652234 Chr.8: 132757283-132757283 on Build GRCh38
rs955285625 Chr.8: 132757480-132757480 on Build GRCh38
rs769995387 Chr.8: 132757245-132757245 on Build GRCh38
rs372314612 Chr.8: 132757271-132757271 on Build GRCh38
rs775641773 Chr.8: 132757247-132757247 on Build GRCh38
rs570998471 Chr.8: 132757315-132757316 on Build GRCh38
rs375727318 Chr.8: 132757280-132757280 on Build GRCh38
rs955708822 Chr.8: 132757434-132757435 on Build GRCh38
rs762882432 Chr.8: 132757250-132757250 on Build GRCh38
rs151072192 Chr.8: 132757290-132757290 on Build GRCh38
rs139679751 Chr.8: 132757254-132757254 on Build GRCh38
rs754684047 Chr.8: 132757281-132757281 on Build GRCh38
rs757809891 Chr.8: 132757286-132757286 on Build GRCh38
rs1037124178 Chr.8: 132757263-132757263 on Build GRCh38
rs759807621 Chr.8: 132757244-132757244 on Build GRCh38
rs961139945 Chr.8: 132757349-132757349 on Build GRCh38
rs968912996 Chr.8: 132757476-132757476 on Build GRCh38
rs866909827 Chr.8: 132757261-132757261 on Build GRCh38
rs773835006 Chr.8: 132757308-132757309 on Build GRCh38
rs111242269 Chr.8: 132757459-132757459 on Build GRCh38
rs770837897 Chr.8: 132757311-132757311 on Build GRCh38
rs968270319 Chr.8: 132757520-132757520 on Build GRCh38
rs768115797 Chr.8: 132757277-132757277 on Build GRCh38
rs776377657 Chr.8: 132757503-132757503 on Build GRCh38
rs147397125 Chr.8: 132757242-132757242 on Build GRCh38
rs544743873 Chr.8: 132757484-132757484 on Build GRCh38
rs959622901 Chr.8: 132757509-132757509 on Build GRCh38
rs188804494 Chr.8: 132757448-132757448 on Build GRCh38
rs762378372 Chr.8: 132757256-132757256 on Build GRCh38
rs138117721 Chr.8: 132757309-132757310 on Build GRCh38
rs555278198 Chr.8: 132757324-132757324 on Build GRCh38
rs1021322298 Chr.8: 132757465-132757465 on Build GRCh38

View Full Product Details