Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.Y: 9399560-9400056 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TSPY3
Entrez Gene ID
728137
Gene Name
testis specific protein, Y-linked 3
Gene Aliases
CT78
UniGene
Hs.556121
Gene Chromosome Location
Chr.Y: 9398421-9401224 on Build GRCh38
Transcripts
NM_001077697.2 XM_005272742.4 XM_011544004.2

SNP Detail

SNP ID Chromosome Location
rs771675793 Chr.Y: 9400036-9400036 on Build GRCh38
rs778618256 Chr.Y: 9399970-9399971 on Build GRCh38
rs778589299 Chr.Y: 9400023-9400023 on Build GRCh38
rs747771474 Chr.Y: 9400025-9400025 on Build GRCh38
rs754154065 Chr.Y: 9399983-9399983 on Build GRCh38
rs755328003 Chr.Y: 9400005-9400005 on Build GRCh38
rs304805 Chr.Y: 9400037-9400037 on Build GRCh38

View Full Product Details