Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 167341846-167342355 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TTLL2
Entrez Gene ID
83887
Gene Name
tubulin tyrosine ligase like 2
Gene Aliases
C6orf104, NYD-TSPG, dJ366N23.3
UniGene
Hs.520554
Gene Chromosome Location
Chr.6: 167325086-167342710 on Build GRCh38
Transcripts
NM_031949.4 XM_006715572.3

SNP Detail

SNP ID Chromosome Location
rs997628452 Chr.6: 167342100-167342100 on Build GRCh38
rs575350467 Chr.6: 167342236-167342236 on Build GRCh38
rs2981978 Chr.6: 167341945-167341945 on Build GRCh38
rs759546513 Chr.6: 167342264-167342264 on Build GRCh38
rs937685137 Chr.6: 167342312-167342312 on Build GRCh38
rs925068929 Chr.6: 167341937-167341937 on Build GRCh38
rs926250132 Chr.6: 167342247-167342247 on Build GRCh38
rs937590405 Chr.6: 167342069-167342069 on Build GRCh38
rs1030358989 Chr.6: 167341900-167341900 on Build GRCh38
rs753336689 Chr.6: 167342224-167342224 on Build GRCh38
rs986086139 Chr.6: 167341923-167341923 on Build GRCh38
rs958089881 Chr.6: 167342169-167342169 on Build GRCh38
rs1055973219 Chr.6: 167342076-167342077 on Build GRCh38
rs540499661 Chr.6: 167341944-167341944 on Build GRCh38
rs76006598 Chr.6: 167341910-167341910 on Build GRCh38
rs942224933 Chr.6: 167342077-167342077 on Build GRCh38
rs915876890 Chr.6: 167342137-167342137 on Build GRCh38
rs556996626 Chr.6: 167342227-167342227 on Build GRCh38
rs934921456 Chr.6: 167342261-167342261 on Build GRCh38
rs1046035801 Chr.6: 167342057-167342057 on Build GRCh38
rs542300050 Chr.6: 167342077-167342078 on Build GRCh38
rs533931856 Chr.6: 167342017-167342017 on Build GRCh38
rs142986266 Chr.6: 167342246-167342246 on Build GRCh38
rs771332928 Chr.6: 167342133-167342134 on Build GRCh38
rs544223196 Chr.6: 167342173-167342173 on Build GRCh38
rs970496956 Chr.6: 167342240-167342240 on Build GRCh38
rs889124620 Chr.6: 167342138-167342138 on Build GRCh38
rs568250971 Chr.6: 167342099-167342099 on Build GRCh38
rs1021237823 Chr.6: 167342231-167342231 on Build GRCh38
rs760333202 Chr.6: 167342060-167342060 on Build GRCh38
rs2981979 Chr.6: 167341956-167341956 on Build GRCh38
rs949183500 Chr.6: 167342055-167342055 on Build GRCh38
rs1054655616 Chr.6: 167341878-167341878 on Build GRCh38
rs1054623581 Chr.6: 167342324-167342324 on Build GRCh38
rs1007449907 Chr.6: 167342146-167342146 on Build GRCh38
rs913671805 Chr.6: 167341967-167341967 on Build GRCh38
rs549388773 Chr.6: 167342085-167342085 on Build GRCh38
rs987976182 Chr.6: 167342178-167342178 on Build GRCh38
rs560453070 Chr.6: 167342326-167342326 on Build GRCh38
rs1007758466 Chr.6: 167341905-167341905 on Build GRCh38
rs892866287 Chr.6: 167341887-167341887 on Build GRCh38
rs981662780 Chr.6: 167342221-167342221 on Build GRCh38
rs901513961 Chr.6: 167341922-167341922 on Build GRCh38
rs78097800 Chr.6: 167342168-167342168 on Build GRCh38
rs566663170 Chr.6: 167341968-167341968 on Build GRCh38
rs766007003 Chr.6: 167342066-167342066 on Build GRCh38
rs1016551425 Chr.6: 167342076-167342076 on Build GRCh38
rs962819300 Chr.6: 167342078-167342078 on Build GRCh38
rs781062102 Chr.6: 167341986-167341986 on Build GRCh38

View Full Product Details