Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 75681395-75681937 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TTLL5
Entrez Gene ID
23093
Gene Name
tubulin tyrosine ligase like 5
Gene Aliases
CORD19, KIAA0998, STAMP
UniGene
Hs.709609
Gene Chromosome Location
Chr.14: 75661208-75955082 on Build GRCh38
Transcripts
NM_015072.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4578020 c.185G>A p.R62H Chr.14: 75681548-75681548 on Build GRCh38
433387 c.211C>G p.R71G Chr.14: 75681574-75681574 on Build GRCh38
4911237 c.182-1G>T p.? Chr.14: 75681544-75681544 on Build GRCh38
1371180 c.224G>A p.R75H Chr.14: 75681587-75681587 on Build GRCh38
5948369 c.215C>T p.T72M Chr.14: 75681578-75681578 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs962320996 Chr.14: 75681822-75681822 on Build GRCh38
rs757176297 Chr.14: 75681594-75681594 on Build GRCh38
rs771356937 Chr.14: 75681668-75681668 on Build GRCh38
rs368944443 Chr.14: 75681814-75681814 on Build GRCh38
rs1042138407 Chr.14: 75681708-75681708 on Build GRCh38
rs574151007 Chr.14: 75681445-75681445 on Build GRCh38
rs530244229 Chr.14: 75681488-75681488 on Build GRCh38
rs549156164 Chr.14: 75681878-75681878 on Build GRCh38
rs542766469 Chr.14: 75681487-75681487 on Build GRCh38
rs990141346 Chr.14: 75681911-75681913 on Build GRCh38
rs369997391 Chr.14: 75681595-75681595 on Build GRCh38
rs776986442 Chr.14: 75681670-75681670 on Build GRCh38
rs186137096 Chr.14: 75681713-75681713 on Build GRCh38
rs756369159 Chr.14: 75681500-75681500 on Build GRCh38
rs891754595 Chr.14: 75681694-75681694 on Build GRCh38
rs780987658 Chr.14: 75681494-75681494 on Build GRCh38
rs762611824 Chr.14: 75681674-75681674 on Build GRCh38
rs995960190 Chr.14: 75681738-75681738 on Build GRCh38
rs969853716 Chr.14: 75681853-75681853 on Build GRCh38
rs992700420 Chr.14: 75681836-75681836 on Build GRCh38
rs980891740 Chr.14: 75681861-75681861 on Build GRCh38
rs554109852 Chr.14: 75681807-75681807 on Build GRCh38
rs745686995 Chr.14: 75681498-75681498 on Build GRCh38
rs774297805 Chr.14: 75681545-75681545 on Build GRCh38
rs149514506 Chr.14: 75681908-75681908 on Build GRCh38
rs778267207 Chr.14: 75681913-75681913 on Build GRCh38
rs886276476 Chr.14: 75681831-75681831 on Build GRCh38
rs747364578 Chr.14: 75681658-75681658 on Build GRCh38
rs936704237 Chr.14: 75681579-75681579 on Build GRCh38
rs994705020 Chr.14: 75681438-75681438 on Build GRCh38
rs768847182 Chr.14: 75681542-75681542 on Build GRCh38
rs373520758 Chr.14: 75681645-75681645 on Build GRCh38
rs775184984 Chr.14: 75681575-75681575 on Build GRCh38
rs761983463 Chr.14: 75681421-75681421 on Build GRCh38
rs756030583 Chr.14: 75681599-75681599 on Build GRCh38
rs773528252 Chr.14: 75681564-75681564 on Build GRCh38
rs563439430 Chr.14: 75681424-75681424 on Build GRCh38
rs915401180 Chr.14: 75681661-75681661 on Build GRCh38
rs567793589 Chr.14: 75681821-75681821 on Build GRCh38
rs758345139 Chr.14: 75681813-75681813 on Build GRCh38
rs765479385 Chr.14: 75681627-75681627 on Build GRCh38
rs925394190 Chr.14: 75681879-75681879 on Build GRCh38
rs748629838 Chr.14: 75681548-75681548 on Build GRCh38
rs948170380 Chr.14: 75681547-75681547 on Build GRCh38
rs772480677 Chr.14: 75681552-75681552 on Build GRCh38
rs138075327 Chr.14: 75681886-75681886 on Build GRCh38
rs773757522 Chr.14: 75681654-75681654 on Build GRCh38
rs918093689 Chr.14: 75681838-75681838 on Build GRCh38
rs565110992 Chr.14: 75681698-75681698 on Build GRCh38
rs772600489 Chr.14: 75681650-75681650 on Build GRCh38
rs145135000 Chr.14: 75681578-75681578 on Build GRCh38
rs750217504 Chr.14: 75681596-75681596 on Build GRCh38
rs1045431257 Chr.14: 75681553-75681553 on Build GRCh38
rs751047559 Chr.14: 75681587-75681587 on Build GRCh38
rs1054948930 Chr.14: 75681754-75681754 on Build GRCh38
rs566972730 Chr.14: 75681881-75681881 on Build GRCh38
rs758473802 Chr.14: 75681780-75681780 on Build GRCh38
rs761068583 Chr.14: 75681571-75681571 on Build GRCh38
rs1028569941 Chr.14: 75681898-75681898 on Build GRCh38
rs780419945 Chr.14: 75681608-75681608 on Build GRCh38
rs114450239 Chr.14: 75681493-75681493 on Build GRCh38
rs1040216389 Chr.14: 75681702-75681702 on Build GRCh38
rs201717756 Chr.14: 75681633-75681633 on Build GRCh38
rs779003256 Chr.14: 75681644-75681644 on Build GRCh38
rs763596307 Chr.14: 75681586-75681586 on Build GRCh38
rs939047453 Chr.14: 75681732-75681732 on Build GRCh38
rs755167226 Chr.14: 75681631-75681631 on Build GRCh38
rs780469230 Chr.14: 75681502-75681502 on Build GRCh38
rs998224365 Chr.14: 75681888-75681888 on Build GRCh38
rs749364412 Chr.14: 75681515-75681515 on Build GRCh38
rs1028637048 Chr.14: 75681764-75681764 on Build GRCh38
rs551679767 Chr.14: 75681696-75681696 on Build GRCh38
rs536745010 Chr.14: 75681852-75681852 on Build GRCh38

View Full Product Details