Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 67182748-67183283 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TYW1
Entrez Gene ID
55253
Gene Name
tRNA-yW synthesizing protein 1 homolog
Gene Aliases
RSAFD1, TYW1A, YPL207W
UniGene
Hs.520917
Gene Chromosome Location
Chr.7: 66996805-67239520 on Build GRCh38
Transcripts
NM_018264.3 NR_134540.1 XM_011516372.2 XM_017012392.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3663300 c.1705C>T p.R569* Chr.7: 67183132-67183132 on Build GRCh38
1451873 c.1765G>A p.A589T Chr.7: 67183192-67183192 on Build GRCh38
4418296 c.1743C>T p.N581N Chr.7: 67183170-67183170 on Build GRCh38
398356 c.1774G>T p.V592L Chr.7: 67183201-67183201 on Build GRCh38
3882107 c.1725G>A p.T575T Chr.7: 67183152-67183152 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs146322625 Chr.7: 67183206-67183206 on Build GRCh38
rs185015015 Chr.7: 67183097-67183097 on Build GRCh38
rs4145351 Chr.7: 67182897-67182897 on Build GRCh38
rs1028893463 Chr.7: 67182779-67182779 on Build GRCh38
rs777259269 Chr.7: 67183210-67183210 on Build GRCh38
rs779626948 Chr.7: 67183168-67183168 on Build GRCh38
rs372725266 Chr.7: 67183252-67183252 on Build GRCh38
rs547114863 Chr.7: 67183257-67183257 on Build GRCh38
rs982907706 Chr.7: 67183018-67183018 on Build GRCh38
rs1011384096 Chr.7: 67182863-67182863 on Build GRCh38
rs568698065 Chr.7: 67182788-67182788 on Build GRCh38
rs1036088331 Chr.7: 67182909-67182910 on Build GRCh38
rs755994594 Chr.7: 67183179-67183179 on Build GRCh38
rs2948336 Chr.7: 67183043-67183043 on Build GRCh38
rs531820949 Chr.7: 67183191-67183191 on Build GRCh38
rs754509946 Chr.7: 67183171-67183171 on Build GRCh38
rs570253417 Chr.7: 67182902-67182902 on Build GRCh38
rs200770817 Chr.7: 67183095-67183095 on Build GRCh38
rs57386346 Chr.7: 67183042-67183042 on Build GRCh38
rs577442165 Chr.7: 67183114-67183114 on Build GRCh38
rs4145352 Chr.7: 67182879-67182879 on Build GRCh38
rs777735942 Chr.7: 67183232-67183232 on Build GRCh38
rs764577174 Chr.7: 67183155-67183155 on Build GRCh38
rs369471651 Chr.7: 67183250-67183250 on Build GRCh38
rs772930972 Chr.7: 67183106-67183106 on Build GRCh38
rs184644676 Chr.7: 67183245-67183245 on Build GRCh38
rs376519717 Chr.7: 67183176-67183176 on Build GRCh38
rs920143668 Chr.7: 67182987-67182987 on Build GRCh38
rs148898519 Chr.7: 67183236-67183236 on Build GRCh38
rs770736900 Chr.7: 67183193-67183193 on Build GRCh38
rs755589026 Chr.7: 67183228-67183228 on Build GRCh38
rs767034362 Chr.7: 67183224-67183224 on Build GRCh38
rs957207058 Chr.7: 67182825-67182825 on Build GRCh38
rs1050344286 Chr.7: 67183096-67183096 on Build GRCh38
rs907648530 Chr.7: 67183029-67183029 on Build GRCh38
rs542363709 Chr.7: 67183197-67183197 on Build GRCh38
rs752028264 Chr.7: 67183019-67183019 on Build GRCh38
rs192287125 Chr.7: 67183214-67183214 on Build GRCh38
rs763256344 Chr.7: 67183108-67183108 on Build GRCh38
rs555970988 Chr.7: 67183033-67183033 on Build GRCh38
rs2298398 Chr.7: 67183151-67183151 on Build GRCh38
rs778712871 Chr.7: 67183249-67183249 on Build GRCh38
rs749076683 Chr.7: 67183192-67183192 on Build GRCh38
rs192923958 Chr.7: 67183013-67183013 on Build GRCh38
rs775665185 Chr.7: 67183200-67183200 on Build GRCh38
rs145674605 Chr.7: 67183227-67183227 on Build GRCh38
rs958709120 Chr.7: 67182911-67182911 on Build GRCh38
rs6943708 Chr.7: 67183082-67183082 on Build GRCh38
rs373327693 Chr.7: 67183137-67183137 on Build GRCh38
rs754598678 Chr.7: 67183222-67183222 on Build GRCh38
rs546341528 Chr.7: 67183128-67183128 on Build GRCh38
rs147474467 Chr.7: 67183133-67183133 on Build GRCh38
rs761207173 Chr.7: 67183147-67183147 on Build GRCh38
rs991713510 Chr.7: 67182957-67182957 on Build GRCh38
rs2687038 Chr.7: 67183262-67183262 on Build GRCh38
rs4145350 Chr.7: 67183170-67183170 on Build GRCh38
rs746545783 Chr.7: 67183089-67183089 on Build GRCh38
rs748088780 Chr.7: 67183177-67183177 on Build GRCh38
rs2687039 Chr.7: 67183104-67183104 on Build GRCh38
rs375543606 Chr.7: 67183194-67183194 on Build GRCh38
rs140515560 Chr.7: 67183186-67183186 on Build GRCh38
rs754218013 Chr.7: 67183156-67183156 on Build GRCh38
rs766661238 Chr.7: 67182910-67182911 on Build GRCh38
rs562934984 Chr.7: 67183131-67183131 on Build GRCh38
rs547866608 Chr.7: 67182867-67182867 on Build GRCh38
rs144362734 Chr.7: 67183201-67183201 on Build GRCh38
rs753261616 Chr.7: 67183143-67183143 on Build GRCh38
rs757614057 Chr.7: 67183164-67183164 on Build GRCh38
rs373627237 Chr.7: 67183083-67183083 on Build GRCh38

View Full Product Details