Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 149448522-149449038 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZC3H12D
Entrez Gene ID
340152
Gene Name
zinc finger CCCH-type containing 12D
Gene Aliases
C6orf95, MCPIP4, TFL, dJ281H8.1, p34
UniGene
Hs.632618
Gene Chromosome Location
Chr.6: 149447630-149485012 on Build GRCh38
Transcripts
NM_207360.2

SNP Detail

SNP ID Chromosome Location
rs901678894 Chr.6: 149448647-149448647 on Build GRCh38
rs751700621 Chr.6: 149448709-149448709 on Build GRCh38
rs540557204 Chr.6: 149448936-149448936 on Build GRCh38
rs1050316044 Chr.6: 149448996-149448996 on Build GRCh38
rs887304927 Chr.6: 149448664-149448664 on Build GRCh38
rs940171643 Chr.6: 149448960-149448960 on Build GRCh38
rs573481848 Chr.6: 149448745-149448745 on Build GRCh38
rs926930151 Chr.6: 149448549-149448549 on Build GRCh38
rs879669955 Chr.6: 149448992-149448992 on Build GRCh38
rs574948518 Chr.6: 149448858-149448858 on Build GRCh38
rs539708725 Chr.6: 149448782-149448782 on Build GRCh38
rs2789496 Chr.6: 149448907-149448907 on Build GRCh38
rs1050347113 Chr.6: 149448669-149448669 on Build GRCh38
rs12197676 Chr.6: 149448837-149448837 on Build GRCh38
rs564091998 Chr.6: 149448910-149448910 on Build GRCh38
rs781580673 Chr.6: 149448811-149448811 on Build GRCh38
rs868363998 Chr.6: 149448739-149448739 on Build GRCh38
rs757714558 Chr.6: 149448799-149448799 on Build GRCh38
rs970929662 Chr.6: 149448812-149448812 on Build GRCh38
rs1054112524 Chr.6: 149448576-149448576 on Build GRCh38
rs922702054 Chr.6: 149448967-149448967 on Build GRCh38
rs1041491038 Chr.6: 149448966-149448966 on Build GRCh38
rs191612075 Chr.6: 149448930-149448930 on Build GRCh38
rs528483409 Chr.6: 149448937-149448937 on Build GRCh38
rs564858138 Chr.6: 149448980-149448980 on Build GRCh38
rs778055290 Chr.6: 149448667-149448667 on Build GRCh38
rs932767614 Chr.6: 149448968-149448968 on Build GRCh38
rs984383063 Chr.6: 149448928-149448928 on Build GRCh38
rs73779356 Chr.6: 149448659-149448659 on Build GRCh38
rs940057132 Chr.6: 149448668-149448668 on Build GRCh38
rs937022913 Chr.6: 149448569-149448569 on Build GRCh38
rs751262154 Chr.6: 149448642-149448642 on Build GRCh38
rs889096934 Chr.6: 149448676-149448676 on Build GRCh38
rs527368013 Chr.6: 149448824-149448824 on Build GRCh38
rs923068459 Chr.6: 149448953-149448953 on Build GRCh38
rs548685370 Chr.6: 149448580-149448580 on Build GRCh38
rs1049731643 Chr.6: 149448978-149448978 on Build GRCh38
rs187572257 Chr.6: 149448654-149448654 on Build GRCh38
rs992117811 Chr.6: 149448559-149448559 on Build GRCh38
rs572737144 Chr.6: 149448817-149448817 on Build GRCh38
rs773787719 Chr.6: 149448949-149448949 on Build GRCh38
rs1034784009 Chr.6: 149448848-149448848 on Build GRCh38
rs1041151161 Chr.6: 149448738-149448738 on Build GRCh38
rs535978977 Chr.6: 149448771-149448771 on Build GRCh38
rs548648368 Chr.6: 149449013-149449013 on Build GRCh38
rs996864904 Chr.6: 149448775-149448775 on Build GRCh38
rs761348426 Chr.6: 149448987-149448987 on Build GRCh38
rs916159888 Chr.6: 149448620-149448620 on Build GRCh38
rs553659723 Chr.6: 149448787-149448787 on Build GRCh38
rs62621196 Chr.6: 149448931-149448931 on Build GRCh38
rs534051485 Chr.6: 149448643-149448643 on Build GRCh38
rs541467636 Chr.6: 149448832-149448832 on Build GRCh38
rs557540905 Chr.6: 149448711-149448711 on Build GRCh38
rs993679125 Chr.6: 149448794-149448794 on Build GRCh38
rs1025526361 Chr.6: 149448798-149448798 on Build GRCh38
rs959419549 Chr.6: 149448859-149448859 on Build GRCh38
rs779679023 Chr.6: 149448884-149448884 on Build GRCh38
rs1020716672 Chr.6: 149448807-149448807 on Build GRCh38
rs1010755876 Chr.6: 149449014-149449014 on Build GRCh38
rs990500978 Chr.6: 149448867-149448867 on Build GRCh38
rs914349059 Chr.6: 149448613-149448613 on Build GRCh38
rs1006250515 Chr.6: 149448736-149448736 on Build GRCh38

View Full Product Details