Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 153345326-153345837 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF275
Entrez Gene ID
10838
Gene Name
zinc finger protein 275
Gene Aliases
-
UniGene
Hs.348963
Gene Chromosome Location
Chr.X: 153334155-153352926 on Build GRCh38
Transcripts
NM_001080485.3

SNP Detail

SNP ID Chromosome Location
rs144271371 Chr.X: 153345673-153345673 on Build GRCh38
rs782140192 Chr.X: 153345528-153345528 on Build GRCh38
rs75458321 Chr.X: 153345736-153345736 on Build GRCh38
rs781893185 Chr.X: 153345601-153345601 on Build GRCh38
rs12847869 Chr.X: 153345756-153345756 on Build GRCh38
rs3213466 Chr.X: 153345527-153345527 on Build GRCh38
rs926367772 Chr.X: 153345510-153345510 on Build GRCh38
rs187160433 Chr.X: 153345393-153345393 on Build GRCh38
rs1021182353 Chr.X: 153345433-153345433 on Build GRCh38
rs782246507 Chr.X: 153345471-153345471 on Build GRCh38
rs371051380 Chr.X: 153345522-153345522 on Build GRCh38
rs782321250 Chr.X: 153345561-153345561 on Build GRCh38
rs782573874 Chr.X: 153345354-153345354 on Build GRCh38
rs78686240 Chr.X: 153345750-153345750 on Build GRCh38
rs782571664 Chr.X: 153345624-153345624 on Build GRCh38
rs782314309 Chr.X: 153345507-153345507 on Build GRCh38
rs78085283 Chr.X: 153345734-153345734 on Build GRCh38
rs371097541 Chr.X: 153345715-153345715 on Build GRCh38
rs782584865 Chr.X: 153345521-153345521 on Build GRCh38
rs35044224 Chr.X: 153345800-153345800 on Build GRCh38
rs782202877 Chr.X: 153345496-153345496 on Build GRCh38
rs781841112 Chr.X: 153345638-153345638 on Build GRCh38
rs1051886672 Chr.X: 153345690-153345690 on Build GRCh38
rs370534782 Chr.X: 153345548-153345548 on Build GRCh38
rs374358643 Chr.X: 153345643-153345643 on Build GRCh38
rs782633795 Chr.X: 153345670-153345670 on Build GRCh38
rs781973258 Chr.X: 153345698-153345698 on Build GRCh38
rs782055394 Chr.X: 153345727-153345727 on Build GRCh38
rs12858008 Chr.X: 153345737-153345737 on Build GRCh38
rs76790272 Chr.X: 153345759-153345759 on Build GRCh38
rs782416354 Chr.X: 153345570-153345570 on Build GRCh38
rs3213465 Chr.X: 153345489-153345489 on Build GRCh38
rs5969963 Chr.X: 153345771-153345771 on Build GRCh38
rs192726030 Chr.X: 153345478-153345478 on Build GRCh38
rs12850397 Chr.X: 153345755-153345755 on Build GRCh38
rs782123691 Chr.X: 153345590-153345590 on Build GRCh38
rs782808492 Chr.X: 153345592-153345592 on Build GRCh38
rs368950000 Chr.X: 153345578-153345578 on Build GRCh38
rs919446252 Chr.X: 153345648-153345648 on Build GRCh38
rs942249345 Chr.X: 153345576-153345576 on Build GRCh38
rs12850385 Chr.X: 153345741-153345741 on Build GRCh38
rs372182880 Chr.X: 153345620-153345620 on Build GRCh38
rs185084050 Chr.X: 153345587-153345587 on Build GRCh38
rs781877476 Chr.X: 153345588-153345588 on Build GRCh38
rs782366999 Chr.X: 153345525-153345525 on Build GRCh38
rs76073837 Chr.X: 153345753-153345753 on Build GRCh38

View Full Product Details