Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 73891657-73892184 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF410
Entrez Gene ID
57862
Gene Name
zinc finger protein 410
Gene Aliases
APA-1, APA1
UniGene
Hs.270869
Gene Chromosome Location
Chr.14: 73886615-73932288 on Build GRCh38
Transcripts
NM_001242924.1 NM_001242926.1 NM_001242927.1 NM_001242928.1 NM_021188.2 NR_040251.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3815280 c.8C>T p.S3L Chr.14: 73892183-73892183 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs766405818 Chr.14: 73892007-73892007 on Build GRCh38
rs1008995283 Chr.14: 73891959-73891959 on Build GRCh38
rs890118085 Chr.14: 73891957-73891957 on Build GRCh38
rs756698259 Chr.14: 73892158-73892158 on Build GRCh38
rs548550504 Chr.14: 73891971-73891971 on Build GRCh38
rs561415909 Chr.14: 73891902-73891902 on Build GRCh38
rs774490841 Chr.14: 73891803-73891803 on Build GRCh38
rs191132237 Chr.14: 73891682-73891682 on Build GRCh38
rs757999945 Chr.14: 73892125-73892130 on Build GRCh38
rs778568081 Chr.14: 73892061-73892061 on Build GRCh38
rs929069825 Chr.14: 73891936-73891936 on Build GRCh38
rs980336280 Chr.14: 73891896-73891897 on Build GRCh38
rs559813252 Chr.14: 73891711-73891711 on Build GRCh38
rs760373284 Chr.14: 73891997-73891997 on Build GRCh38
rs757085575 Chr.14: 73891997-73891998 on Build GRCh38
rs140665239 Chr.14: 73892001-73892002 on Build GRCh38
rs746397386 Chr.14: 73892043-73892043 on Build GRCh38
rs781775351 Chr.14: 73892042-73892042 on Build GRCh38
rs762933624 Chr.14: 73892120-73892120 on Build GRCh38
rs35307600 Chr.14: 73891889-73891889 on Build GRCh38
rs11540490 Chr.14: 73892041-73892041 on Build GRCh38
rs759648754 Chr.14: 73891765-73891765 on Build GRCh38
rs776517544 Chr.14: 73892020-73892020 on Build GRCh38
rs750133067 Chr.14: 73892032-73892035 on Build GRCh38
rs905049588 Chr.14: 73891799-73891799 on Build GRCh38
rs530963010 Chr.14: 73891837-73891837 on Build GRCh38
rs1034558384 Chr.14: 73892110-73892110 on Build GRCh38
rs965013208 Chr.14: 73891896-73891896 on Build GRCh38
rs750747095 Chr.14: 73892151-73892151 on Build GRCh38
rs763801845 Chr.14: 73891725-73891726 on Build GRCh38
rs1028017659 Chr.14: 73891943-73891943 on Build GRCh38
rs772634695 Chr.14: 73892064-73892064 on Build GRCh38
rs201025600 Chr.14: 73891708-73891709 on Build GRCh38
rs927541997 Chr.14: 73891897-73891897 on Build GRCh38
rs62003711 Chr.14: 73891895-73891895 on Build GRCh38
rs11540492 Chr.14: 73891909-73891909 on Build GRCh38
rs561728098 Chr.14: 73891854-73891854 on Build GRCh38
rs747549608 Chr.14: 73892062-73892062 on Build GRCh38
rs371715301 Chr.14: 73892142-73892142 on Build GRCh38
rs189104598 Chr.14: 73892082-73892082 on Build GRCh38
rs764169309 Chr.14: 73892032-73892032 on Build GRCh38
rs200605259 Chr.14: 73892007-73892007 on Build GRCh38
rs771553833 Chr.14: 73891990-73891990 on Build GRCh38
rs917254057 Chr.14: 73891886-73891886 on Build GRCh38
rs1038548544 Chr.14: 73891932-73891932 on Build GRCh38
rs1047960392 Chr.14: 73891946-73891946 on Build GRCh38
rs752023691 Chr.14: 73892040-73892040 on Build GRCh38
rs752735544 Chr.14: 73892028-73892028 on Build GRCh38
rs773073394 Chr.14: 73891991-73891991 on Build GRCh38
rs764447000 Chr.14: 73892122-73892122 on Build GRCh38
rs765325517 Chr.14: 73892023-73892023 on Build GRCh38
rs927426852 Chr.14: 73891684-73891684 on Build GRCh38
rs530474265 Chr.14: 73891916-73891916 on Build GRCh38
rs1000686230 Chr.14: 73891801-73891801 on Build GRCh38
rs149671462 Chr.14: 73891765-73891765 on Build GRCh38
rs1017505504 Chr.14: 73891728-73891728 on Build GRCh38
rs1009856978 Chr.14: 73891878-73891878 on Build GRCh38
rs565401843 Chr.14: 73891819-73891819 on Build GRCh38
rs77953765 Chr.14: 73892102-73892102 on Build GRCh38
rs543640675 Chr.14: 73891748-73891748 on Build GRCh38
rs920306499 Chr.14: 73891935-73891936 on Build GRCh38
rs746859689 Chr.14: 73892081-73892081 on Build GRCh38
rs573359734 Chr.14: 73891714-73891714 on Build GRCh38
rs772734308 Chr.14: 73892067-73892067 on Build GRCh38
rs900429958 Chr.14: 73892015-73892015 on Build GRCh38
rs999953038 Chr.14: 73892059-73892059 on Build GRCh38
rs771210920 Chr.14: 73891740-73891740 on Build GRCh38
rs940917885 Chr.14: 73891712-73891712 on Build GRCh38
rs776362558 Chr.14: 73892085-73892085 on Build GRCh38
rs758651596 Chr.14: 73892029-73892029 on Build GRCh38
rs896444562 Chr.14: 73891725-73891725 on Build GRCh38
rs892399687 Chr.14: 73891843-73891843 on Build GRCh38
rs952437114 Chr.14: 73892022-73892022 on Build GRCh38
rs769653093 Chr.14: 73892092-73892092 on Build GRCh38
rs761264356 Chr.14: 73892097-73892097 on Build GRCh38
rs559660624 Chr.14: 73892091-73892091 on Build GRCh38
rs749431306 Chr.14: 73891691-73891691 on Build GRCh38
rs891662104 Chr.14: 73892066-73892066 on Build GRCh38
rs537441738 Chr.14: 73891712-73891713 on Build GRCh38
rs183799215 Chr.14: 73891769-73891769 on Build GRCh38
rs1013710937 Chr.14: 73891727-73891727 on Build GRCh38
rs377327866 Chr.14: 73892048-73892048 on Build GRCh38
rs780916703 Chr.14: 73892019-73892019 on Build GRCh38
rs756646778 Chr.14: 73892057-73892057 on Build GRCh38
rs762289142 Chr.14: 73892144-73892144 on Build GRCh38
rs768022764 Chr.14: 73892147-73892147 on Build GRCh38
rs909332659 Chr.14: 73892076-73892076 on Build GRCh38
rs143996625 Chr.14: 73891726-73891727 on Build GRCh38

View Full Product Details