Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 44712544-44713050 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF502
Entrez Gene ID
91392
Gene Name
zinc finger protein 502
Gene Aliases
-
UniGene
Hs.224843
Gene Chromosome Location
Chr.3: 44712643-44723831 on Build GRCh38
Transcripts
NM_001134440.1 NM_001134441.1 NM_001134442.1 NM_001282880.1 NM_033210.4

SNP Detail

SNP ID Chromosome Location
rs967025047 Chr.3: 44712607-44712607 on Build GRCh38
rs940019496 Chr.3: 44712682-44712682 on Build GRCh38
rs866916563 Chr.3: 44712652-44712652 on Build GRCh38
rs544450858 Chr.3: 44712850-44712850 on Build GRCh38
rs139755879 Chr.3: 44712693-44712693 on Build GRCh38
rs1028896081 Chr.3: 44712609-44712609 on Build GRCh38
rs1050227797 Chr.3: 44712947-44712947 on Build GRCh38
rs868463987 Chr.3: 44712928-44712928 on Build GRCh38
rs936851652 Chr.3: 44712674-44712674 on Build GRCh38
rs961495531 Chr.3: 44712991-44712994 on Build GRCh38
rs908578928 Chr.3: 44712675-44712675 on Build GRCh38
rs1037648346 Chr.3: 44712912-44712912 on Build GRCh38
rs1015702419 Chr.3: 44712768-44712768 on Build GRCh38
rs894242356 Chr.3: 44712593-44712593 on Build GRCh38
rs958418813 Chr.3: 44712809-44712809 on Build GRCh38
rs561880279 Chr.3: 44712691-44712691 on Build GRCh38
rs528353805 Chr.3: 44712663-44712663 on Build GRCh38
rs995959712 Chr.3: 44712601-44712601 on Build GRCh38
rs374193820 Chr.3: 44712888-44712889 on Build GRCh38
rs899275300 Chr.3: 44712914-44712914 on Build GRCh38
rs762445014 Chr.3: 44712876-44712876 on Build GRCh38
rs932039495 Chr.3: 44712915-44712915 on Build GRCh38
rs1046836756 Chr.3: 44712696-44712696 on Build GRCh38
rs1004936879 Chr.3: 44712581-44712581 on Build GRCh38
rs961443300 Chr.3: 44712781-44712781 on Build GRCh38
rs574280973 Chr.3: 44712802-44712802 on Build GRCh38
rs559626012 Chr.3: 44712636-44712636 on Build GRCh38
rs1005292565 Chr.3: 44712706-44712706 on Build GRCh38
rs1020228814 Chr.3: 44713020-44713020 on Build GRCh38
rs934407567 Chr.3: 44712875-44712875 on Build GRCh38
rs940732651 Chr.3: 44712886-44712886 on Build GRCh38
rs534167000 Chr.3: 44712762-44712762 on Build GRCh38
rs570124969 Chr.3: 44712743-44712743 on Build GRCh38
rs967104106 Chr.3: 44712782-44712782 on Build GRCh38
rs915774516 Chr.3: 44712823-44712823 on Build GRCh38
rs1015754800 Chr.3: 44712990-44712991 on Build GRCh38
rs755989643 Chr.3: 44712738-44712738 on Build GRCh38
rs753861533 Chr.3: 44712862-44712862 on Build GRCh38
rs1008426890 Chr.3: 44712973-44712973 on Build GRCh38
rs556025495 Chr.3: 44712792-44712792 on Build GRCh38
rs891341554 Chr.3: 44712941-44712941 on Build GRCh38
rs776992318 Chr.3: 44712707-44712707 on Build GRCh38
rs556700066 Chr.3: 44712883-44712883 on Build GRCh38
rs537978476 Chr.3: 44712754-44712754 on Build GRCh38
rs757216641 Chr.3: 44712864-44712864 on Build GRCh38
rs996875229 Chr.3: 44712901-44712901 on Build GRCh38
rs565872718 Chr.3: 44712727-44712727 on Build GRCh38
rs963222721 Chr.3: 44712599-44712599 on Build GRCh38
rs955676203 Chr.3: 44712654-44712654 on Build GRCh38
rs6771160 Chr.3: 44712685-44712685 on Build GRCh38
rs535415101 Chr.3: 44712812-44712812 on Build GRCh38
rs185946990 Chr.3: 44712702-44712702 on Build GRCh38
rs75266471 Chr.3: 44713014-44713014 on Build GRCh38
rs959404544 Chr.3: 44713022-44713022 on Build GRCh38
rs1016517008 Chr.3: 44712594-44712594 on Build GRCh38
rs1056073821 Chr.3: 44712692-44712692 on Build GRCh38
rs569298367 Chr.3: 44712708-44712708 on Build GRCh38
rs974148262 Chr.3: 44712998-44712998 on Build GRCh38
rs551514661 Chr.3: 44712717-44712717 on Build GRCh38
rs990996191 Chr.3: 44712853-44712853 on Build GRCh38
rs752603591 Chr.3: 44712700-44712700 on Build GRCh38
rs967533847 Chr.3: 44712656-44712656 on Build GRCh38
rs978819754 Chr.3: 44712798-44712798 on Build GRCh38
rs115032414 Chr.3: 44712649-44712649 on Build GRCh38
rs925682986 Chr.3: 44712665-44712665 on Build GRCh38
rs919924488 Chr.3: 44712791-44712791 on Build GRCh38
rs954640884 Chr.3: 44712747-44712747 on Build GRCh38
rs975575779 Chr.3: 44712646-44712646 on Build GRCh38
rs1052775172 Chr.3: 44712902-44712902 on Build GRCh38
rs539565237 Chr.3: 44712709-44712709 on Build GRCh38
rs954610263 Chr.3: 44712610-44712610 on Build GRCh38
rs916714226 Chr.3: 44712854-44712854 on Build GRCh38
rs890333871 Chr.3: 44712966-44712966 on Build GRCh38

View Full Product Details