Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 37389830-37390373 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF527
Entrez Gene ID
84503
Gene Name
zinc finger protein 527
Gene Aliases
-
UniGene
Hs.590940
Gene Chromosome Location
Chr.19: 37371055-37393066 on Build GRCh38
Transcripts
NM_032453.1 XM_005259328.4 XM_005259329.3 XM_017027380.1 XM_017027381.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3728518 c.1801C>G p.Q601E Chr.19: 37389850-37389850 on Build GRCh38
4744147 c.1824G>A p.T608T Chr.19: 37389873-37389873 on Build GRCh38
5625222 c.1793G>C p.R598T Chr.19: 37389842-37389842 on Build GRCh38
286901 c.1782C>A p.V594V Chr.19: 37389831-37389831 on Build GRCh38
126433 c.1811A>G p.H604R Chr.19: 37389860-37389860 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772299274 Chr.19: 37389890-37389890 on Build GRCh38
rs913258647 Chr.19: 37390083-37390083 on Build GRCh38
rs760419974 Chr.19: 37389905-37389905 on Build GRCh38
rs980284033 Chr.19: 37390303-37390303 on Build GRCh38
rs1030202494 Chr.19: 37390109-37390109 on Build GRCh38
rs766309859 Chr.19: 37390320-37390320 on Build GRCh38
rs954331335 Chr.19: 37390112-37390112 on Build GRCh38
rs377309382 Chr.19: 37389916-37389916 on Build GRCh38
rs536084472 Chr.19: 37390134-37390134 on Build GRCh38
rs927365690 Chr.19: 37389999-37389999 on Build GRCh38
rs1042299217 Chr.19: 37390100-37390100 on Build GRCh38
rs1004518966 Chr.19: 37390279-37390279 on Build GRCh38
rs746189982 Chr.19: 37390218-37390218 on Build GRCh38
rs973456822 Chr.19: 37390184-37390184 on Build GRCh38
rs537144865 Chr.19: 37390236-37390236 on Build GRCh38
rs895744345 Chr.19: 37389937-37389937 on Build GRCh38
rs186622721 Chr.19: 37390180-37390180 on Build GRCh38
rs752840637 Chr.19: 37390265-37390265 on Build GRCh38
rs985671299 Chr.19: 37390113-37390113 on Build GRCh38
rs537050103 Chr.19: 37389928-37389928 on Build GRCh38
rs1017254789 Chr.19: 37390153-37390153 on Build GRCh38
rs749838561 Chr.19: 37389862-37389862 on Build GRCh38
rs891844395 Chr.19: 37390025-37390025 on Build GRCh38
rs386388969 Chr.19: 37390035-37390036 on Build GRCh38
rs200322627 Chr.19: 37389872-37389872 on Build GRCh38
rs945262757 Chr.19: 37390089-37390089 on Build GRCh38
rs938796564 Chr.19: 37390039-37390039 on Build GRCh38
rs900241353 Chr.19: 37390232-37390232 on Build GRCh38
rs950486437 Chr.19: 37390221-37390221 on Build GRCh38
rs1031772927 Chr.19: 37389998-37389998 on Build GRCh38
rs773374842 Chr.19: 37389899-37389899 on Build GRCh38
rs755911825 Chr.19: 37390318-37390318 on Build GRCh38
rs1016449540 Chr.19: 37390290-37390290 on Build GRCh38
rs386388970 Chr.19: 37390045-37390046 on Build GRCh38
rs950967371 Chr.19: 37389877-37389877 on Build GRCh38
rs992824071 Chr.19: 37390053-37390053 on Build GRCh38
rs992941820 Chr.19: 37390319-37390319 on Build GRCh38
rs936479502 Chr.19: 37390121-37390121 on Build GRCh38
rs1000710958 Chr.19: 37389987-37389987 on Build GRCh38
rs33996770 Chr.19: 37390034-37390035 on Build GRCh38
rs386365555 Chr.19: 37390046-37390047 on Build GRCh38
rs532006208 Chr.19: 37390219-37390219 on Build GRCh38
rs191121542 Chr.19: 37390275-37390275 on Build GRCh38
rs780899303 Chr.19: 37389905-37389906 on Build GRCh38
rs141536000 Chr.19: 37389972-37389972 on Build GRCh38
rs147075017 Chr.19: 37390295-37390295 on Build GRCh38
rs78381139 Chr.19: 37389978-37389978 on Build GRCh38
rs1039287582 Chr.19: 37390181-37390181 on Build GRCh38
rs202094634 Chr.19: 37389854-37389854 on Build GRCh38
rs998671487 Chr.19: 37390321-37390321 on Build GRCh38
rs558822749 Chr.19: 37390309-37390309 on Build GRCh38
rs765295556 Chr.19: 37390060-37390060 on Build GRCh38
rs904510276 Chr.19: 37389985-37389985 on Build GRCh38
rs948579830 Chr.19: 37389945-37389945 on Build GRCh38
rs779034783 Chr.19: 37389873-37389873 on Build GRCh38
rs917084587 Chr.19: 37390249-37390249 on Build GRCh38
rs997259999 Chr.19: 37390248-37390248 on Build GRCh38
rs748162386 Chr.19: 37389882-37389882 on Build GRCh38
rs992678623 Chr.19: 37390235-37390235 on Build GRCh38

View Full Product Details