Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 245770-246265 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF596
Entrez Gene ID
169270
Gene Name
zinc finger protein 596
Gene Aliases
-
UniGene
Hs.591388
Gene Chromosome Location
Chr.8: 232137-247340 on Build GRCh38
Transcripts
NM_001042415.2 NM_001042416.2 NM_001287254.1 NM_001287255.1 NM_001287256.1 NM_001287399.1 NM_173539.3 XM_011534744.1 XM_017013166.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1097832 c.1373G>T p.R458I Chr.8: 246220-246220 on Build GRCh38
3737840 c.1222C>T p.R408* Chr.8: 246069-246069 on Build GRCh38
1097833 c.1384T>G p.F462V Chr.8: 246231-246231 on Build GRCh38
5063017 c.924A>T p.G308G Chr.8: 245771-245771 on Build GRCh38
1097830 c.1312G>T p.E438* Chr.8: 246159-246159 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs149423885 Chr.8: 245948-245948 on Build GRCh38
rs765627223 Chr.8: 245855-245855 on Build GRCh38
rs1026292733 Chr.8: 246206-246206 on Build GRCh38
rs762387236 Chr.8: 245925-245925 on Build GRCh38
rs370301818 Chr.8: 246004-246004 on Build GRCh38
rs116191154 Chr.8: 245879-245879 on Build GRCh38
rs755229489 Chr.8: 246007-246007 on Build GRCh38
rs567054359 Chr.8: 246069-246069 on Build GRCh38
rs137958651 Chr.8: 245916-245916 on Build GRCh38
rs541906501 Chr.8: 245900-245900 on Build GRCh38
rs759624995 Chr.8: 246072-246072 on Build GRCh38
rs780800565 Chr.8: 246112-246112 on Build GRCh38
rs867899295 Chr.8: 245983-245983 on Build GRCh38
rs149728676 Chr.8: 245841-245841 on Build GRCh38
rs1045412234 Chr.8: 246106-246106 on Build GRCh38
rs759278467 Chr.8: 245986-245986 on Build GRCh38
rs758274674 Chr.8: 246098-246098 on Build GRCh38
rs73521331 Chr.8: 245990-245990 on Build GRCh38
rs747471421 Chr.8: 246128-246128 on Build GRCh38
rs1007766291 Chr.8: 246160-246160 on Build GRCh38
rs201947775 Chr.8: 246197-246197 on Build GRCh38
rs749896247 Chr.8: 246159-246159 on Build GRCh38
rs773899860 Chr.8: 245827-245827 on Build GRCh38
rs1055483196 Chr.8: 245905-245905 on Build GRCh38
rs950814561 Chr.8: 246002-246002 on Build GRCh38
rs140067348 Chr.8: 245985-245985 on Build GRCh38
rs201186640 Chr.8: 246013-246013 on Build GRCh38
rs373764251 Chr.8: 246025-246025 on Build GRCh38
rs751424005 Chr.8: 246154-246154 on Build GRCh38
rs774558389 Chr.8: 246047-246047 on Build GRCh38
rs751478981 Chr.8: 246103-246103 on Build GRCh38
rs774258415 Chr.8: 246237-246237 on Build GRCh38
rs768548272 Chr.8: 245904-245904 on Build GRCh38
rs753462766 Chr.8: 246168-246168 on Build GRCh38
rs768837485 Chr.8: 245989-245989 on Build GRCh38
rs950878363 Chr.8: 246217-246217 on Build GRCh38
rs201577009 Chr.8: 246029-246029 on Build GRCh38
rs773765404 Chr.8: 246063-246063 on Build GRCh38
rs755243515 Chr.8: 246109-246110 on Build GRCh38
rs757239263 Chr.8: 246157-246157 on Build GRCh38
rs763092549 Chr.8: 246147-246147 on Build GRCh38
rs117305688 Chr.8: 246121-246121 on Build GRCh38
rs1039298477 Chr.8: 246170-246170 on Build GRCh38
rs765541303 Chr.8: 246073-246073 on Build GRCh38
rs773568973 Chr.8: 246126-246126 on Build GRCh38
rs775466505 Chr.8: 246230-246230 on Build GRCh38
rs929835422 Chr.8: 245835-245835 on Build GRCh38
rs746562692 Chr.8: 245966-245966 on Build GRCh38
rs773304108 Chr.8: 246243-246245 on Build GRCh38
rs902093587 Chr.8: 246176-246176 on Build GRCh38
rs878942778 Chr.8: 245854-245854 on Build GRCh38
rs758241317 Chr.8: 246011-246011 on Build GRCh38
rs142354791 Chr.8: 246139-246139 on Build GRCh38
rs879126623 Chr.8: 246095-246095 on Build GRCh38
rs150499628 Chr.8: 245927-245927 on Build GRCh38
rs202032273 Chr.8: 245910-245910 on Build GRCh38
rs776720104 Chr.8: 246071-246071 on Build GRCh38
rs145328181 Chr.8: 245903-245903 on Build GRCh38
rs746012343 Chr.8: 245890-245890 on Build GRCh38
rs757294239 Chr.8: 246104-246104 on Build GRCh38
rs780293195 Chr.8: 245938-245938 on Build GRCh38
rs761298475 Chr.8: 245836-245836 on Build GRCh38
rs940615939 Chr.8: 246003-246003 on Build GRCh38
rs370719960 Chr.8: 246171-246171 on Build GRCh38
rs529474239 Chr.8: 246077-246077 on Build GRCh38
rs751954153 Chr.8: 245921-245921 on Build GRCh38
rs747679162 Chr.8: 245909-245909 on Build GRCh38
rs201785288 Chr.8: 246056-246056 on Build GRCh38
rs772073733 Chr.8: 245959-245959 on Build GRCh38
rs141773746 Chr.8: 245895-245895 on Build GRCh38
rs752483498 Chr.8: 245940-245940 on Build GRCh38
rs760809279 Chr.8: 245996-245996 on Build GRCh38
rs778780281 Chr.8: 245946-245946 on Build GRCh38
rs775330275 Chr.8: 245954-245954 on Build GRCh38
rs991365775 Chr.8: 246091-246091 on Build GRCh38
rs931790463 Chr.8: 246214-246214 on Build GRCh38
rs781346837 Chr.8: 245889-245889 on Build GRCh38
rs188238096 Chr.8: 245934-245934 on Build GRCh38
rs756701581 Chr.8: 246119-246119 on Build GRCh38
rs775727492 Chr.8: 246140-246140 on Build GRCh38
rs751220230 Chr.8: 246114-246114 on Build GRCh38
rs183667362 Chr.8: 245821-245821 on Build GRCh38
rs769847673 Chr.8: 246223-246223 on Build GRCh38
rs1007735301 Chr.8: 245975-245975 on Build GRCh38
rs745479203 Chr.8: 246031-246031 on Build GRCh38
rs936037894 Chr.8: 245935-245935 on Build GRCh38
rs776584203 Chr.8: 245884-245884 on Build GRCh38
rs755706426 Chr.8: 246166-246166 on Build GRCh38
rs151239104 Chr.8: 246183-246183 on Build GRCh38
rs200553223 Chr.8: 245800-245800 on Build GRCh38
rs746378595 Chr.8: 246221-246221 on Build GRCh38
rs186344589 Chr.8: 246198-246198 on Build GRCh38
rs753138989 Chr.8: 245867-245867 on Build GRCh38
rs78666370 Chr.8: 246229-246229 on Build GRCh38
rs962957390 Chr.8: 246028-246028 on Build GRCh38
rs745433308 Chr.8: 246116-246116 on Build GRCh38
rs987299348 Chr.8: 246225-246225 on Build GRCh38
rs147693045 Chr.8: 246027-246027 on Build GRCh38
rs201973017 Chr.8: 245956-245956 on Build GRCh38
rs770706627 Chr.8: 245978-245978 on Build GRCh38
rs1024909944 Chr.8: 246105-246105 on Build GRCh38
rs922440203 Chr.8: 245812-245812 on Build GRCh38
rs866731953 Chr.8: 245808-245808 on Build GRCh38
rs777927639 Chr.8: 246023-246023 on Build GRCh38
rs748585147 Chr.8: 246048-246048 on Build GRCh38
rs561813298 Chr.8: 245907-245907 on Build GRCh38
rs766572673 Chr.8: 246000-246000 on Build GRCh38
rs769509507 Chr.8: 246036-246036 on Build GRCh38
rs763998457 Chr.8: 246101-246101 on Build GRCh38
rs754785761 Chr.8: 245941-245941 on Build GRCh38
rs765240628 Chr.8: 246059-246059 on Build GRCh38
rs528278097 Chr.8: 245823-245823 on Build GRCh38
rs772791560 Chr.8: 245843-245843 on Build GRCh38
rs368718146 Chr.8: 245920-245920 on Build GRCh38
rs370749334 Chr.8: 245876-245876 on Build GRCh38
rs763481538 Chr.8: 245914-245914 on Build GRCh38
rs994542217 Chr.8: 246042-246042 on Build GRCh38
rs139472874 Chr.8: 245930-245930 on Build GRCh38
rs778528538 Chr.8: 246008-246008 on Build GRCh38
rs754137939 Chr.8: 245884-245886 on Build GRCh38
rs530836868 Chr.8: 245911-245911 on Build GRCh38
rs778059989 Chr.8: 246187-246187 on Build GRCh38
rs752555246 Chr.8: 246010-246010 on Build GRCh38
rs145555549 Chr.8: 245868-245868 on Build GRCh38
rs756306914 Chr.8: 245931-245931 on Build GRCh38
rs766377726 Chr.8: 246070-246070 on Build GRCh38
rs748446791 Chr.8: 246120-246120 on Build GRCh38
rs771446971 Chr.8: 246133-246133 on Build GRCh38
rs779442990 Chr.8: 246167-246167 on Build GRCh38
rs868678419 Chr.8: 245810-245810 on Build GRCh38
rs762177709 Chr.8: 245992-245992 on Build GRCh38
rs766831324 Chr.8: 245806-245810 on Build GRCh38
rs768119203 Chr.8: 245993-245993 on Build GRCh38
rs750470074 Chr.8: 245995-245995 on Build GRCh38
rs760413673 Chr.8: 245852-245852 on Build GRCh38
rs375404674 Chr.8: 245892-245892 on Build GRCh38
rs556671635 Chr.8: 246242-246242 on Build GRCh38
rs759732316 Chr.8: 246135-246135 on Build GRCh38
rs760769107 Chr.8: 245813-245813 on Build GRCh38
rs78181259 Chr.8: 245960-245960 on Build GRCh38
rs763943484 Chr.8: 246153-246153 on Build GRCh38
rs200366695 Chr.8: 246050-246050 on Build GRCh38
rs781753767 Chr.8: 246216-246216 on Build GRCh38

View Full Product Details