Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 46474481-46475003 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KRBOX4
Entrez Gene ID
55634
Gene Name
KRAB box domain containing 4
Gene Aliases
ZNF673
UniGene
Hs.632800
Gene Chromosome Location
Chr.X: 46447189-46474639 on Build GRCh38
Transcripts
NM_001129898.1 NM_001129899.1 NM_001129900.1 NM_017776.2

SNP Detail

SNP ID Chromosome Location
rs985836437 Chr.X: 46474596-46474596 on Build GRCh38
rs889667237 Chr.X: 46474950-46474950 on Build GRCh38
rs1038848445 Chr.X: 46474749-46474749 on Build GRCh38
rs193006756 Chr.X: 46474933-46474933 on Build GRCh38
rs760419203 Chr.X: 46474712-46474712 on Build GRCh38
rs752129903 Chr.X: 46474711-46474711 on Build GRCh38
rs550197040 Chr.X: 46474546-46474546 on Build GRCh38
rs1055157227 Chr.X: 46474840-46474840 on Build GRCh38
rs767489226 Chr.X: 46474790-46474790 on Build GRCh38
rs950250167 Chr.X: 46474700-46474700 on Build GRCh38
rs750120334 Chr.X: 46474562-46474563 on Build GRCh38
rs1042188548 Chr.X: 46474868-46474868 on Build GRCh38
rs922808410 Chr.X: 46474609-46474609 on Build GRCh38
rs978648157 Chr.X: 46474928-46474928 on Build GRCh38
rs745378199 Chr.X: 46474573-46474573 on Build GRCh38
rs11091187 Chr.X: 46474727-46474727 on Build GRCh38
rs767419125 Chr.X: 46474632-46474633 on Build GRCh38
rs144966899 Chr.X: 46474674-46474674 on Build GRCh38
rs12560021 Chr.X: 46474862-46474862 on Build GRCh38
rs184726189 Chr.X: 46474974-46474974 on Build GRCh38
rs1005697301 Chr.X: 46474687-46474687 on Build GRCh38
rs962730692 Chr.X: 46474662-46474662 on Build GRCh38
rs942349148 Chr.X: 46474514-46474514 on Build GRCh38
rs1051504143 Chr.X: 46474702-46474702 on Build GRCh38
rs1034987045 Chr.X: 46474859-46474859 on Build GRCh38
rs893514349 Chr.X: 46474860-46474860 on Build GRCh38
rs902329151 Chr.X: 46474908-46474908 on Build GRCh38
rs998403137 Chr.X: 46474918-46474918 on Build GRCh38
rs937614107 Chr.X: 46474768-46474768 on Build GRCh38
rs775258235 Chr.X: 46474613-46474616 on Build GRCh38
rs946440002 Chr.X: 46474866-46474866 on Build GRCh38
rs922855349 Chr.X: 46474567-46474567 on Build GRCh38
rs150236852 Chr.X: 46474580-46474580 on Build GRCh38
rs905308881 Chr.X: 46474839-46474839 on Build GRCh38
rs58041336 Chr.X: 46474688-46474688 on Build GRCh38
rs371106394 Chr.X: 46474616-46474619 on Build GRCh38
rs753694245 Chr.X: 46474930-46474930 on Build GRCh38

View Full Product Details