Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 15836437-15836971 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLJ37453
Entrez Gene ID
729614
Gene Name
uncharacterized LOC729614
Gene Aliases
-
UniGene
Hs.119298
Gene Chromosome Location
Chr.1: 15834215-15848147 on Build GRCh38
Transcripts
NR_024279.1

SNP Detail

SNP ID Chromosome Location
rs529380739 Chr.1: 15836801-15836801 on Build GRCh38
rs995920675 Chr.1: 15836508-15836508 on Build GRCh38
rs904626390 Chr.1: 15836611-15836611 on Build GRCh38
rs1000242225 Chr.1: 15836641-15836641 on Build GRCh38
rs966697307 Chr.1: 15836542-15836542 on Build GRCh38
rs933248812 Chr.1: 15836652-15836652 on Build GRCh38
rs370360877 Chr.1: 15836935-15836935 on Build GRCh38
rs929485246 Chr.1: 15836560-15836560 on Build GRCh38
rs980385580 Chr.1: 15836489-15836489 on Build GRCh38
rs778132600 Chr.1: 15836892-15836892 on Build GRCh38
rs1002035239 Chr.1: 15836832-15836832 on Build GRCh38
rs1004545939 Chr.1: 15836728-15836728 on Build GRCh38
rs1051592737 Chr.1: 15836664-15836664 on Build GRCh38
rs970655729 Chr.1: 15836669-15836669 on Build GRCh38
rs550785332 Chr.1: 15836736-15836736 on Build GRCh38
rs549284632 Chr.1: 15836874-15836874 on Build GRCh38
rs898897954 Chr.1: 15836752-15836752 on Build GRCh38
rs970838688 Chr.1: 15836478-15836478 on Build GRCh38
rs1025333384 Chr.1: 15836660-15836660 on Build GRCh38
rs980415332 Chr.1: 15836706-15836706 on Build GRCh38
rs182118569 Chr.1: 15836505-15836505 on Build GRCh38
rs917833151 Chr.1: 15836471-15836471 on Build GRCh38
rs551627046 Chr.1: 15836838-15836838 on Build GRCh38
rs1009399402 Chr.1: 15836654-15836654 on Build GRCh38
rs933153976 Chr.1: 15836831-15836831 on Build GRCh38
rs377459746 Chr.1: 15836482-15836482 on Build GRCh38
rs950138785 Chr.1: 15836927-15836927 on Build GRCh38
rs6691557 Chr.1: 15836760-15836760 on Build GRCh38
rs780570977 Chr.1: 15836814-15836814 on Build GRCh38
rs112138935 Chr.1: 15836865-15836865 on Build GRCh38
rs936524586 Chr.1: 15836619-15836619 on Build GRCh38
rs1025222419 Chr.1: 15836797-15836797 on Build GRCh38
rs112314149 Chr.1: 15836769-15836769 on Build GRCh38
rs912144389 Chr.1: 15836789-15836789 on Build GRCh38
rs958162547 Chr.1: 15836815-15836815 on Build GRCh38
rs1039025249 Chr.1: 15836602-15836602 on Build GRCh38
rs978099511 Chr.1: 15836805-15836805 on Build GRCh38
rs921741468 Chr.1: 15836645-15836645 on Build GRCh38
rs1001661112 Chr.1: 15836768-15836768 on Build GRCh38
rs778037155 Chr.1: 15836518-15836518 on Build GRCh38
rs148753642 Chr.1: 15836850-15836850 on Build GRCh38
rs1053650819 Chr.1: 15836827-15836827 on Build GRCh38
rs978615038 Chr.1: 15836574-15836574 on Build GRCh38
rs372774718 Chr.1: 15836517-15836517 on Build GRCh38
rs919388968 Chr.1: 15836531-15836531 on Build GRCh38
rs973163532 Chr.1: 15836740-15836740 on Build GRCh38
rs1038886570 Chr.1: 15836629-15836629 on Build GRCh38
rs963056637 Chr.1: 15836833-15836833 on Build GRCh38
rs747223418 Chr.1: 15836794-15836794 on Build GRCh38
rs926190264 Chr.1: 15836898-15836898 on Build GRCh38
rs549361659 Chr.1: 15836828-15836828 on Build GRCh38
rs533337610 Chr.1: 15836687-15836687 on Build GRCh38
rs954401824 Chr.1: 15836946-15836946 on Build GRCh38
rs553718918 Chr.1: 15836931-15836931 on Build GRCh38
rs1046659924 Chr.1: 15836929-15836930 on Build GRCh38
rs1053112274 Chr.1: 15836911-15836913 on Build GRCh38
rs1033853417 Chr.1: 15836783-15836783 on Build GRCh38
rs80256350 Chr.1: 15836485-15836485 on Build GRCh38
rs936010016 Chr.1: 15836901-15836901 on Build GRCh38
rs545308299 Chr.1: 15836530-15836530 on Build GRCh38
rs1052486873 Chr.1: 15836561-15836561 on Build GRCh38
rs531665717 Chr.1: 15836528-15836528 on Build GRCh38
rs527859939 Chr.1: 15836894-15836894 on Build GRCh38
rs1029194514 Chr.1: 15836625-15836625 on Build GRCh38
rs187140274 Chr.1: 15836829-15836829 on Build GRCh38
rs943609428 Chr.1: 15836880-15836880 on Build GRCh38
rs35945411 Chr.1: 15836482-15836483 on Build GRCh38
rs890696289 Chr.1: 15836582-15836582 on Build GRCh38
rs940376402 Chr.1: 15836944-15836944 on Build GRCh38
rs757512760 Chr.1: 15836607-15836607 on Build GRCh38
rs71002946 Chr.1: 15836499-15836500 on Build GRCh38
rs765542010 Chr.1: 15836873-15836873 on Build GRCh38
rs886147454 Chr.1: 15836720-15836720 on Build GRCh38
rs959486657 Chr.1: 15836788-15836788 on Build GRCh38
rs775942243 Chr.1: 15836823-15836824 on Build GRCh38
rs1051668144 Chr.1: 15836834-15836834 on Build GRCh38
rs562698495 Chr.1: 15836459-15836459 on Build GRCh38
rs758686740 Chr.1: 15836904-15836904 on Build GRCh38
rs956652697 Chr.1: 15836469-15836469 on Build GRCh38
rs144227816 Chr.1: 15836600-15836600 on Build GRCh38
rs975199958 Chr.1: 15836825-15836825 on Build GRCh38
rs891887114 Chr.1: 15836917-15836917 on Build GRCh38
rs1000188430 Chr.1: 15836820-15836820 on Build GRCh38

View Full Product Details