Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 99147646-99148152 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC100129620
Entrez Gene ID
100129620
Gene Name
uncharacterized LOC100129620
Gene Aliases
-
UniGene
Hs.588631
Gene Chromosome Location
Chr.1: 99004276-99148852 on Build GRCh38
Transcripts
NR_033940.1

SNP Detail

SNP ID Chromosome Location
rs1053177121 Chr.1: 99147681-99147681 on Build GRCh38
rs1012872845 Chr.1: 99148036-99148036 on Build GRCh38
rs549556782 Chr.1: 99147903-99147903 on Build GRCh38
rs748750543 Chr.1: 99147827-99147827 on Build GRCh38
rs117894196 Chr.1: 99147687-99147687 on Build GRCh38
rs891558503 Chr.1: 99147682-99147682 on Build GRCh38
rs531811327 Chr.1: 99147984-99147984 on Build GRCh38
rs781357767 Chr.1: 99147808-99147808 on Build GRCh38
rs965630938 Chr.1: 99147915-99147915 on Build GRCh38
rs112447843 Chr.1: 99147684-99147684 on Build GRCh38
rs17119715 Chr.1: 99148034-99148034 on Build GRCh38
rs1018601482 Chr.1: 99148125-99148125 on Build GRCh38
rs536892102 Chr.1: 99148087-99148087 on Build GRCh38
rs906038586 Chr.1: 99147706-99147706 on Build GRCh38
rs146566302 Chr.1: 99147987-99147987 on Build GRCh38
rs556771191 Chr.1: 99148107-99148107 on Build GRCh38
rs576237322 Chr.1: 99148114-99148114 on Build GRCh38
rs571913827 Chr.1: 99147828-99147828 on Build GRCh38
rs905587467 Chr.1: 99148063-99148063 on Build GRCh38
rs921474928 Chr.1: 99147971-99147971 on Build GRCh38
rs113964853 Chr.1: 99147788-99147788 on Build GRCh38
rs1043112519 Chr.1: 99147997-99147997 on Build GRCh38
rs987108941 Chr.1: 99147890-99147890 on Build GRCh38
rs190171130 Chr.1: 99147868-99147868 on Build GRCh38
rs1026852475 Chr.1: 99147904-99147904 on Build GRCh38
rs144300307 Chr.1: 99147912-99147912 on Build GRCh38
rs141238419 Chr.1: 99148024-99148024 on Build GRCh38
rs898956744 Chr.1: 99148040-99148040 on Build GRCh38
rs766180631 Chr.1: 99147757-99147757 on Build GRCh38
rs912883502 Chr.1: 99148026-99148026 on Build GRCh38
rs1026033088 Chr.1: 99148102-99148102 on Build GRCh38
rs779509363 Chr.1: 99148006-99148006 on Build GRCh38
rs758651856 Chr.1: 99148039-99148039 on Build GRCh38
rs544704921 Chr.1: 99147741-99147741 on Build GRCh38
rs896097105 Chr.1: 99148125-99148128 on Build GRCh38
rs372554466 Chr.1: 99147707-99147707 on Build GRCh38
rs150982000 Chr.1: 99147800-99147800 on Build GRCh38
rs548118947 Chr.1: 99148032-99148032 on Build GRCh38
rs186880582 Chr.1: 99148014-99148014 on Build GRCh38
rs943925457 Chr.1: 99148013-99148013 on Build GRCh38
rs181569544 Chr.1: 99147869-99147869 on Build GRCh38
rs542179190 Chr.1: 99147929-99147929 on Build GRCh38
rs150186819 Chr.1: 99148088-99148088 on Build GRCh38
rs139651635 Chr.1: 99147857-99147857 on Build GRCh38
rs947545325 Chr.1: 99147974-99147974 on Build GRCh38

View Full Product Details