Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 100281079-100281614 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MIR553
Entrez Gene ID
693138
Gene Name
microRNA 553
Gene Aliases
MIRN553, hsa-mir-553
UniGene
Gene Chromosome Location
Chr.1: 100281241-100281308 on Build GRCh38
Transcripts
NR_030279.1 NM_001130841.1 NM_003729.3 XM_005271298.4 XM_005271299.4 XM_017002669.1

Gene Symbol
RTCA
Entrez Gene ID
8634
Gene Name
RNA 3'-terminal phosphate cyclase
Gene Aliases
RPC, RTC1, RTCD1
UniGene
Hs.484222
Gene Chromosome Location
Chr.1: 100266158-100292769 on Build GRCh38
Transcripts
NR_030279.1 NM_001130841.1 NM_003729.3 XM_005271298.4 XM_005271299.4 XM_017002669.1

SNP Detail

SNP ID Chromosome Location
rs760153585 Chr.1: 100281215-100281215 on Build GRCh38
rs1001165586 Chr.1: 100281307-100281307 on Build GRCh38
rs956734304 Chr.1: 100281315-100281315 on Build GRCh38
rs559168796 Chr.1: 100281291-100281291 on Build GRCh38
rs568293497 Chr.1: 100281128-100281128 on Build GRCh38
rs139920725 Chr.1: 100281442-100281442 on Build GRCh38
rs762706612 Chr.1: 100281240-100281240 on Build GRCh38
rs779152303 Chr.1: 100281492-100281492 on Build GRCh38
rs985812154 Chr.1: 100281529-100281529 on Build GRCh38
rs770513106 Chr.1: 100281377-100281377 on Build GRCh38
rs190622705 Chr.1: 100281258-100281258 on Build GRCh38
rs150968989 Chr.1: 100281323-100281326 on Build GRCh38
rs1001484497 Chr.1: 100281558-100281558 on Build GRCh38
rs1000162035 Chr.1: 100281423-100281423 on Build GRCh38
rs781561579 Chr.1: 100281328-100281328 on Build GRCh38
rs546204898 Chr.1: 100281552-100281552 on Build GRCh38
rs150551481 Chr.1: 100281167-100281167 on Build GRCh38
rs778192927 Chr.1: 100281302-100281302 on Build GRCh38
rs767449823 Chr.1: 100281246-100281246 on Build GRCh38
rs112891767 Chr.1: 100281299-100281299 on Build GRCh38
rs761261667 Chr.1: 100281337-100281338 on Build GRCh38
rs182887234 Chr.1: 100281409-100281409 on Build GRCh38
rs1052721013 Chr.1: 100281559-100281559 on Build GRCh38
rs927148723 Chr.1: 100281400-100281400 on Build GRCh38
rs563795365 Chr.1: 100281514-100281514 on Build GRCh38
rs559727053 Chr.1: 100281571-100281571 on Build GRCh38
rs539064757 Chr.1: 100281279-100281279 on Build GRCh38
rs866763513 Chr.1: 100281444-100281444 on Build GRCh38
rs901998722 Chr.1: 100281235-100281235 on Build GRCh38
rs113552542 Chr.1: 100281356-100281356 on Build GRCh38
rs372814095 Chr.1: 100281325-100281328 on Build GRCh38
rs930567669 Chr.1: 100281212-100281212 on Build GRCh38
rs766012377 Chr.1: 100281317-100281321 on Build GRCh38
rs745486825 Chr.1: 100281352-100281352 on Build GRCh38
rs374474735 Chr.1: 100281312-100281312 on Build GRCh38
rs764810041 Chr.1: 100281351-100281351 on Build GRCh38
rs969488211 Chr.1: 100281357-100281357 on Build GRCh38
rs1040129818 Chr.1: 100281233-100281233 on Build GRCh38
rs187279045 Chr.1: 100281183-100281183 on Build GRCh38
rs892432046 Chr.1: 100281568-100281568 on Build GRCh38
rs751360626 Chr.1: 100281481-100281481 on Build GRCh38
rs747516614 Chr.1: 100281304-100281304 on Build GRCh38
rs112789988 Chr.1: 100281508-100281508 on Build GRCh38
rs775490802 Chr.1: 100281215-100281215 on Build GRCh38
rs188674230 Chr.1: 100281507-100281507 on Build GRCh38
rs763973554 Chr.1: 100281385-100281385 on Build GRCh38
rs944341385 Chr.1: 100281550-100281553 on Build GRCh38
rs755274212 Chr.1: 100281260-100281260 on Build GRCh38
rs572632934 Chr.1: 100281292-100281292 on Build GRCh38
rs761644778 Chr.1: 100281224-100281224 on Build GRCh38
rs1050054264 Chr.1: 100281160-100281160 on Build GRCh38
rs762773615 Chr.1: 100281192-100281192 on Build GRCh38
rs1025001566 Chr.1: 100281572-100281572 on Build GRCh38
rs554859663 Chr.1: 100281430-100281430 on Build GRCh38
rs1014063937 Chr.1: 100281351-100281351 on Build GRCh38
rs371608722 Chr.1: 100281219-100281219 on Build GRCh38
rs577275685 Chr.1: 100281516-100281516 on Build GRCh38
rs923011893 Chr.1: 100281506-100281506 on Build GRCh38
rs1050552743 Chr.1: 100281549-100281549 on Build GRCh38
rs370592334 Chr.1: 100281515-100281515 on Build GRCh38

View Full Product Details