Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 165581227-165581723 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC400794
Entrez Gene ID
400794
Gene Name
uncharacterized LOC400794
Gene Aliases
-
UniGene
Hs.374847
Gene Chromosome Location
Chr.1: 165476842-165582155 on Build GRCh38
Transcripts
NR_026744.2

SNP Detail

SNP ID Chromosome Location
rs964324839 Chr.1: 165581610-165581610 on Build GRCh38
rs920132726 Chr.1: 165581630-165581630 on Build GRCh38
rs41267554 Chr.1: 165581417-165581417 on Build GRCh38
rs911474332 Chr.1: 165581605-165581605 on Build GRCh38
rs897359328 Chr.1: 165581380-165581380 on Build GRCh38
rs41267552 Chr.1: 165581397-165581397 on Build GRCh38
rs182105567 Chr.1: 165581586-165581586 on Build GRCh38
rs538867734 Chr.1: 165581536-165581536 on Build GRCh38
rs544179789 Chr.1: 165581596-165581596 on Build GRCh38
rs553207556 Chr.1: 165581458-165581458 on Build GRCh38
rs186571483 Chr.1: 165581599-165581599 on Build GRCh38
rs41267558 Chr.1: 165581657-165581657 on Build GRCh38
rs530803382 Chr.1: 165581412-165581412 on Build GRCh38
rs987023275 Chr.1: 165581418-165581418 on Build GRCh38
rs1002839200 Chr.1: 165581507-165581507 on Build GRCh38
rs970274774 Chr.1: 165581422-165581422 on Build GRCh38
rs974635975 Chr.1: 165581617-165581617 on Build GRCh38
rs953207064 Chr.1: 165581694-165581694 on Build GRCh38
rs895058061 Chr.1: 165581667-165581667 on Build GRCh38
rs866994033 Chr.1: 165581669-165581669 on Build GRCh38
rs898882621 Chr.1: 165581602-165581602 on Build GRCh38
rs747465704 Chr.1: 165581362-165581362 on Build GRCh38
rs978152475 Chr.1: 165581391-165581391 on Build GRCh38
rs1049001306 Chr.1: 165581608-165581608 on Build GRCh38
rs565235466 Chr.1: 165581705-165581705 on Build GRCh38
rs536710035 Chr.1: 165581452-165581452 on Build GRCh38
rs919998829 Chr.1: 165581477-165581477 on Build GRCh38
rs778535313 Chr.1: 165581281-165581281 on Build GRCh38
rs890204090 Chr.1: 165581346-165581346 on Build GRCh38
rs1004554693 Chr.1: 165581349-165581349 on Build GRCh38
rs190840242 Chr.1: 165581312-165581312 on Build GRCh38
rs559843292 Chr.1: 165581654-165581654 on Build GRCh38
rs907186651 Chr.1: 165581492-165581492 on Build GRCh38
rs150183842 Chr.1: 165581403-165581403 on Build GRCh38
rs930107506 Chr.1: 165581478-165581478 on Build GRCh38
rs1047489218 Chr.1: 165581479-165581479 on Build GRCh38
rs41267560 Chr.1: 165581666-165581666 on Build GRCh38
rs566646693 Chr.1: 165581464-165581464 on Build GRCh38
rs576989510 Chr.1: 165581609-165581609 on Build GRCh38
rs1012489848 Chr.1: 165581523-165581523 on Build GRCh38
rs41267556 Chr.1: 165581604-165581604 on Build GRCh38
rs138725571 Chr.1: 165581430-165581430 on Build GRCh38
rs1056275524 Chr.1: 165581508-165581508 on Build GRCh38
rs938643701 Chr.1: 165581659-165581659 on Build GRCh38
rs1051347209 Chr.1: 165581339-165581339 on Build GRCh38
rs921733464 Chr.1: 165581683-165581683 on Build GRCh38
rs940346952 Chr.1: 165581588-165581588 on Build GRCh38
rs116383715 Chr.1: 165581647-165581647 on Build GRCh38
rs554895597 Chr.1: 165581567-165581567 on Build GRCh38
rs141109226 Chr.1: 165581259-165581259 on Build GRCh38
rs895067818 Chr.1: 165581510-165581510 on Build GRCh38
rs942807965 Chr.1: 165581438-165581438 on Build GRCh38
rs1022162438 Chr.1: 165581537-165581537 on Build GRCh38
rs999306329 Chr.1: 165581560-165581560 on Build GRCh38
rs533434882 Chr.1: 165581442-165581459 on Build GRCh38
rs967862130 Chr.1: 165581356-165581356 on Build GRCh38
rs968189576 Chr.1: 165581553-165581553 on Build GRCh38
rs948024484 Chr.1: 165581672-165581672 on Build GRCh38

View Full Product Details