Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 168375216-168375737 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MIR557
Entrez Gene ID
693142
Gene Name
microRNA 557
Gene Aliases
MIRN557, hsa-mir-557
UniGene
Gene Chromosome Location
Chr.1: 168375524-168375621 on Build GRCh38
Transcripts
NR_030284.1

SNP Detail

SNP ID Chromosome Location
rs1028216788 Chr.1: 168375387-168375387 on Build GRCh38
rs374318773 Chr.1: 168375592-168375592 on Build GRCh38
rs751073728 Chr.1: 168375528-168375528 on Build GRCh38
rs750782939 Chr.1: 168375553-168375553 on Build GRCh38
rs936228194 Chr.1: 168375357-168375357 on Build GRCh38
rs772314468 Chr.1: 168375274-168375274 on Build GRCh38
rs374126458 Chr.1: 168375538-168375538 on Build GRCh38
rs932102277 Chr.1: 168375469-168375469 on Build GRCh38
rs368745434 Chr.1: 168375640-168375640 on Build GRCh38
rs957518597 Chr.1: 168375702-168375702 on Build GRCh38
rs535003152 Chr.1: 168375516-168375516 on Build GRCh38
rs191063554 Chr.1: 168375704-168375704 on Build GRCh38
rs747105489 Chr.1: 168375498-168375498 on Build GRCh38
rs990319562 Chr.1: 168375705-168375705 on Build GRCh38
rs570134892 Chr.1: 168375424-168375424 on Build GRCh38
rs761689751 Chr.1: 168375511-168375511 on Build GRCh38
rs775086473 Chr.1: 168375626-168375626 on Build GRCh38
rs548703388 Chr.1: 168375370-168375370 on Build GRCh38
rs755123880 Chr.1: 168375664-168375664 on Build GRCh38
rs961314940 Chr.1: 168375296-168375296 on Build GRCh38
rs370414755 Chr.1: 168375513-168375513 on Build GRCh38
rs746084303 Chr.1: 168375474-168375474 on Build GRCh38
rs762472175 Chr.1: 168375630-168375630 on Build GRCh38
rs990350783 Chr.1: 168375416-168375416 on Build GRCh38
rs368301801 Chr.1: 168375567-168375567 on Build GRCh38
rs766601513 Chr.1: 168375652-168375652 on Build GRCh38
rs913512133 Chr.1: 168375423-168375423 on Build GRCh38
rs1023977062 Chr.1: 168375644-168375644 on Build GRCh38
rs1024465399 Chr.1: 168375321-168375321 on Build GRCh38
rs765713092 Chr.1: 168375548-168375548 on Build GRCh38
rs774076663 Chr.1: 168375614-168375614 on Build GRCh38
rs765766669 Chr.1: 168375634-168375634 on Build GRCh38
rs758613020 Chr.1: 168375557-168375557 on Build GRCh38
rs753241083 Chr.1: 168375478-168375478 on Build GRCh38
rs373381430 Chr.1: 168375383-168375383 on Build GRCh38
rs770977808 Chr.1: 168375561-168375561 on Build GRCh38
rs781262389 Chr.1: 168375666-168375666 on Build GRCh38
rs780052771 Chr.1: 168375479-168375479 on Build GRCh38
rs576041174 Chr.1: 168375668-168375668 on Build GRCh38
rs972464533 Chr.1: 168375299-168375299 on Build GRCh38
rs781208997 Chr.1: 168375570-168375570 on Build GRCh38
rs768615763 Chr.1: 168375502-168375502 on Build GRCh38
rs1040411699 Chr.1: 168375426-168375426 on Build GRCh38
rs762525030 Chr.1: 168375517-168375517 on Build GRCh38
rs980638548 Chr.1: 168375336-168375336 on Build GRCh38
rs759095667 Chr.1: 168375530-168375530 on Build GRCh38
rs546829402 Chr.1: 168375523-168375523 on Build GRCh38
rs74123929 Chr.1: 168375322-168375322 on Build GRCh38
rs773121288 Chr.1: 168375418-168375418 on Build GRCh38
rs1005412646 Chr.1: 168375582-168375582 on Build GRCh38
rs985459701 Chr.1: 168375440-168375441 on Build GRCh38
rs568324323 Chr.1: 168375532-168375532 on Build GRCh38
rs751809406 Chr.1: 168375658-168375658 on Build GRCh38
rs889696119 Chr.1: 168375401-168375401 on Build GRCh38
rs185760604 Chr.1: 168375434-168375434 on Build GRCh38
rs1049407550 Chr.1: 168375709-168375709 on Build GRCh38
rs559581021 Chr.1: 168375280-168375280 on Build GRCh38
rs927485877 Chr.1: 168375353-168375353 on Build GRCh38
rs748082545 Chr.1: 168375577-168375577 on Build GRCh38
rs759227475 Chr.1: 168375615-168375615 on Build GRCh38
rs767143289 Chr.1: 168375625-168375625 on Build GRCh38
rs769661027 Chr.1: 168375581-168375581 on Build GRCh38
rs530947422 Chr.1: 168375427-168375427 on Build GRCh38
rs763287481 Chr.1: 168375653-168375653 on Build GRCh38
rs903479597 Chr.1: 168375355-168375355 on Build GRCh38
rs772282173 Chr.1: 168375477-168375477 on Build GRCh38
rs78825966 Chr.1: 168375591-168375591 on Build GRCh38
rs747103957 Chr.1: 168375562-168375562 on Build GRCh38
rs372765041 Chr.1: 168375695-168375695 on Build GRCh38
rs758765822 Chr.1: 168375647-168375647 on Build GRCh38
rs924957072 Chr.1: 168375662-168375662 on Build GRCh38
rs757776730 Chr.1: 168375540-168375540 on Build GRCh38
rs754430920 Chr.1: 168375535-168375535 on Build GRCh38
rs571151277 Chr.1: 168375506-168375506 on Build GRCh38

View Full Product Details