Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 207822368-207822875 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1orf132
Entrez Gene ID
100128537
Gene Name
chromosome 1 open reading frame 132
Gene Aliases
-
UniGene
Hs.445414
Gene Chromosome Location
Chr.1: 207801518-207822703 on Build GRCh38
Transcripts
NR_135298.1 NR_135299.1 NR_026817.1

Gene Symbol
LOC148696
Entrez Gene ID
148696
Gene Name
uncharacterized LOC148696
Gene Aliases
-
UniGene
Hs.125511
Gene Chromosome Location
Chr.1: 207818379-207822596 on Build GRCh38
Transcripts
NR_135298.1 NR_135299.1 NR_026817.1

SNP Detail

SNP ID Chromosome Location
rs1015003592 Chr.1: 207822719-207822719 on Build GRCh38
rs1003118070 Chr.1: 207822631-207822631 on Build GRCh38
rs923154317 Chr.1: 207822764-207822764 on Build GRCh38
rs1050537729 Chr.1: 207822602-207822602 on Build GRCh38
rs907812370 Chr.1: 207822436-207822436 on Build GRCh38
rs900658265 Chr.1: 207822483-207822483 on Build GRCh38
rs1013994719 Chr.1: 207822786-207822786 on Build GRCh38
rs961360144 Chr.1: 207822724-207822724 on Build GRCh38
rs925912094 Chr.1: 207822397-207822397 on Build GRCh38
rs80129642 Chr.1: 207822644-207822644 on Build GRCh38
rs960478151 Chr.1: 207822393-207822393 on Build GRCh38
rs553857373 Chr.1: 207822391-207822391 on Build GRCh38
rs903475843 Chr.1: 207822630-207822630 on Build GRCh38
rs117937945 Chr.1: 207822805-207822805 on Build GRCh38
rs1013476718 Chr.1: 207822664-207822664 on Build GRCh38
rs747358528 Chr.1: 207822398-207822398 on Build GRCh38
rs894648628 Chr.1: 207822648-207822648 on Build GRCh38
rs530963119 Chr.1: 207822718-207822718 on Build GRCh38
rs984832576 Chr.1: 207822415-207822415 on Build GRCh38
rs576656239 Chr.1: 207822701-207822701 on Build GRCh38
rs914691091 Chr.1: 207822832-207822832 on Build GRCh38
rs947423910 Chr.1: 207822617-207822617 on Build GRCh38
rs931835164 Chr.1: 207822797-207822797 on Build GRCh38
rs1049335946 Chr.1: 207822597-207822597 on Build GRCh38
rs947440850 Chr.1: 207822834-207822834 on Build GRCh38
rs916276540 Chr.1: 207822396-207822396 on Build GRCh38
rs1042098307 Chr.1: 207822621-207822621 on Build GRCh38
rs990575631 Chr.1: 207822394-207822394 on Build GRCh38
rs989151571 Chr.1: 207822838-207822838 on Build GRCh38
rs1023394651 Chr.1: 207822687-207822687 on Build GRCh38
rs893187886 Chr.1: 207822603-207822603 on Build GRCh38
rs545983972 Chr.1: 207822612-207822612 on Build GRCh38
rs572257738 Chr.1: 207822520-207822520 on Build GRCh38
rs776149264 Chr.1: 207822704-207822704 on Build GRCh38
rs1040226477 Chr.1: 207822438-207822438 on Build GRCh38
rs1015238511 Chr.1: 207822720-207822720 on Build GRCh38
rs770633435 Chr.1: 207822533-207822533 on Build GRCh38
rs951871624 Chr.1: 207822715-207822715 on Build GRCh38
rs923102097 Chr.1: 207822472-207822472 on Build GRCh38
rs1005551247 Chr.1: 207822702-207822702 on Build GRCh38
rs969862946 Chr.1: 207822813-207822813 on Build GRCh38
rs887995106 Chr.1: 207822657-207822657 on Build GRCh38
rs562474921 Chr.1: 207822734-207822734 on Build GRCh38
rs957281933 Chr.1: 207822834-207822834 on Build GRCh38
rs996378093 Chr.1: 207822525-207822525 on Build GRCh38
rs940591603 Chr.1: 207822437-207822437 on Build GRCh38
rs760357264 Chr.1: 207822404-207822404 on Build GRCh38
rs1041755200 Chr.1: 207822837-207822837 on Build GRCh38
rs961971342 Chr.1: 207822728-207822728 on Build GRCh38
rs555600446 Chr.1: 207822429-207822429 on Build GRCh38
rs543786005 Chr.1: 207822732-207822732 on Build GRCh38

View Full Product Details