Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 43845200-43845705 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LINC00619
Entrez Gene ID
414260
Gene Name
long intergenic non-protein coding RNA 619
Gene Aliases
C10orf136, bA168P8.1
UniGene
Hs.351856
Gene Chromosome Location
Chr.10: 43845306-43850622 on Build GRCh38
Transcripts
NR_033923.1

SNP Detail

SNP ID Chromosome Location
rs930112374 Chr.10: 43845467-43845467 on Build GRCh38
rs996023273 Chr.10: 43845462-43845462 on Build GRCh38
rs990351496 Chr.10: 43845364-43845364 on Build GRCh38
rs1010016381 Chr.10: 43845602-43845602 on Build GRCh38
rs562761199 Chr.10: 43845619-43845619 on Build GRCh38
rs976996127 Chr.10: 43845637-43845637 on Build GRCh38
rs759246344 Chr.10: 43845485-43845485 on Build GRCh38
rs568572749 Chr.10: 43845362-43845362 on Build GRCh38
rs774042955 Chr.10: 43845424-43845424 on Build GRCh38
rs190774164 Chr.10: 43845667-43845667 on Build GRCh38
rs550417072 Chr.10: 43845337-43845337 on Build GRCh38
rs978211533 Chr.10: 43845334-43845334 on Build GRCh38
rs955889620 Chr.10: 43845575-43845575 on Build GRCh38
rs988963471 Chr.10: 43845339-43845340 on Build GRCh38
rs4948785 Chr.10: 43845417-43845417 on Build GRCh38
rs980720212 Chr.10: 43845603-43845603 on Build GRCh38
rs749148501 Chr.10: 43845393-43845393 on Build GRCh38
rs879019977 Chr.10: 43845472-43845472 on Build GRCh38
rs775515635 Chr.10: 43845503-43845503 on Build GRCh38
rs942097878 Chr.10: 43845670-43845670 on Build GRCh38
rs774241750 Chr.10: 43845455-43845455 on Build GRCh38
rs544193962 Chr.10: 43845596-43845596 on Build GRCh38
rs1021773159 Chr.10: 43845592-43845592 on Build GRCh38
rs754731488 Chr.10: 43845352-43845352 on Build GRCh38
rs185131580 Chr.10: 43845258-43845258 on Build GRCh38
rs957796017 Chr.10: 43845565-43845565 on Build GRCh38
rs762366914 Chr.10: 43845566-43845578 on Build GRCh38
rs1030389019 Chr.10: 43845314-43845314 on Build GRCh38
rs1031161698 Chr.10: 43845545-43845545 on Build GRCh38
rs536000682 Chr.10: 43845389-43845389 on Build GRCh38
rs897246663 Chr.10: 43845253-43845253 on Build GRCh38
rs1021773967 Chr.10: 43845611-43845611 on Build GRCh38
rs377158824 Chr.10: 43845628-43845634 on Build GRCh38
rs1016125364 Chr.10: 43845267-43845267 on Build GRCh38
rs914779366 Chr.10: 43845383-43845383 on Build GRCh38
rs956402681 Chr.10: 43845361-43845361 on Build GRCh38
rs764321717 Chr.10: 43845532-43845545 on Build GRCh38
rs900364577 Chr.10: 43845431-43845431 on Build GRCh38
rs201367173 Chr.10: 43845513-43845513 on Build GRCh38
rs947701366 Chr.10: 43845225-43845225 on Build GRCh38
rs981084024 Chr.10: 43845331-43845331 on Build GRCh38
rs141539358 Chr.10: 43845223-43845223 on Build GRCh38
rs750722131 Chr.10: 43845335-43845335 on Build GRCh38
rs74442777 Chr.10: 43845502-43845502 on Build GRCh38
rs928939975 Chr.10: 43845616-43845616 on Build GRCh38
rs779299234 Chr.10: 43845271-43845271 on Build GRCh38
rs10535963 Chr.10: 43845625-43845631 on Build GRCh38
rs538179264 Chr.10: 43845324-43845324 on Build GRCh38
rs907984911 Chr.10: 43845419-43845419 on Build GRCh38
rs914807943 Chr.10: 43845378-43845378 on Build GRCh38
rs772648836 Chr.10: 43845624-43845625 on Build GRCh38
rs765733125 Chr.10: 43845620-43845620 on Build GRCh38
rs1006124272 Chr.10: 43845282-43845282 on Build GRCh38
rs200839817 Chr.10: 43845340-43845341 on Build GRCh38
rs1010677231 Chr.10: 43845578-43845578 on Build GRCh38
rs965658056 Chr.10: 43845632-43845632 on Build GRCh38
rs1049996911 Chr.10: 43845226-43845226 on Build GRCh38
rs941857398 Chr.10: 43845423-43845423 on Build GRCh38
rs983499342 Chr.10: 43845666-43845666 on Build GRCh38
rs540889219 Chr.10: 43845642-43845642 on Build GRCh38
rs574549470 Chr.10: 43845641-43845641 on Build GRCh38
rs969923435 Chr.10: 43845317-43845317 on Build GRCh38
rs938696805 Chr.10: 43845408-43845408 on Build GRCh38
rs199663516 Chr.10: 43845564-43845564 on Build GRCh38
rs887642573 Chr.10: 43845509-43845509 on Build GRCh38
rs754304717 Chr.10: 43845265-43845265 on Build GRCh38
rs576878940 Chr.10: 43845514-43845514 on Build GRCh38
rs944237624 Chr.10: 43845416-43845416 on Build GRCh38
rs374217672 Chr.10: 43845589-43845589 on Build GRCh38
rs770966129 Chr.10: 43845437-43845437 on Build GRCh38
rs762348576 Chr.10: 43845618-43845618 on Build GRCh38
rs889818853 Chr.10: 43845231-43845231 on Build GRCh38
rs113953295 Chr.10: 43845335-43845336 on Build GRCh38
rs117506507 Chr.10: 43845235-43845235 on Build GRCh38
rs745821342 Chr.10: 43845234-43845234 on Build GRCh38
rs1048945671 Chr.10: 43845473-43845473 on Build GRCh38
rs147023490 Chr.10: 43845392-43845392 on Build GRCh38
rs995781358 Chr.10: 43845278-43845278 on Build GRCh38
rs757641590 Chr.10: 43845268-43845268 on Build GRCh38
rs928591764 Chr.10: 43845398-43845398 on Build GRCh38

View Full Product Details