Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 59958540-59959049 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LINC01553
Entrez Gene ID
283025
Gene Name
long intergenic non-protein coding RNA 1553
Gene Aliases
C10orf40
UniGene
Hs.522955
Gene Chromosome Location
Chr.10: 59958217-59960913 on Build GRCh38
Transcripts
NR_024340.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5503615 c.126C>T p.Y42Y Chr.10: 59958546-59958546 on Build GRCh38
3685980 c.104_105insT p.L35fs*7 Chr.10: 59958567-59958568 on Build GRCh38
3685981 c.104delT p.L35fs*1 Chr.10: 59958568-59958568 on Build GRCh38
3382941 c.112G>A p.V38I Chr.10: 59958560-59958560 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs373159918 Chr.10: 59958675-59958675 on Build GRCh38
rs567409491 Chr.10: 59958913-59958913 on Build GRCh38
rs1008607277 Chr.10: 59958903-59958903 on Build GRCh38
rs1042641701 Chr.10: 59958581-59958581 on Build GRCh38
rs546413355 Chr.10: 59958919-59958919 on Build GRCh38
rs958593017 Chr.10: 59958990-59958990 on Build GRCh38
rs1008721702 Chr.10: 59958599-59958599 on Build GRCh38
rs774773937 Chr.10: 59958662-59958662 on Build GRCh38
rs991475257 Chr.10: 59958707-59958707 on Build GRCh38
rs368392153 Chr.10: 59958820-59958820 on Build GRCh38
rs183392397 Chr.10: 59958598-59958598 on Build GRCh38
rs773941194 Chr.10: 59958621-59958621 on Build GRCh38
rs147492503 Chr.10: 59958632-59958632 on Build GRCh38
rs745978600 Chr.10: 59958674-59958674 on Build GRCh38
rs528427106 Chr.10: 59958698-59958698 on Build GRCh38
rs534129728 Chr.10: 59958655-59958655 on Build GRCh38
rs1051150526 Chr.10: 59958953-59958953 on Build GRCh38
rs76885798 Chr.10: 59958683-59958683 on Build GRCh38
rs762414867 Chr.10: 59959001-59959001 on Build GRCh38
rs756078177 Chr.10: 59958858-59958858 on Build GRCh38
rs564728879 Chr.10: 59958925-59958925 on Build GRCh38
rs936130661 Chr.10: 59958815-59958815 on Build GRCh38
rs889754987 Chr.10: 59958617-59958617 on Build GRCh38
rs1034740889 Chr.10: 59958797-59958797 on Build GRCh38
rs959006867 Chr.10: 59958845-59958845 on Build GRCh38
rs550235573 Chr.10: 59958584-59958584 on Build GRCh38
rs950495235 Chr.10: 59958783-59958783 on Build GRCh38
rs915149013 Chr.10: 59958859-59958859 on Build GRCh38
rs1027601461 Chr.10: 59958984-59958984 on Build GRCh38
rs759355644 Chr.10: 59958650-59958650 on Build GRCh38
rs946552224 Chr.10: 59958571-59958571 on Build GRCh38
rs7100839 Chr.10: 59958611-59958611 on Build GRCh38
rs889893786 Chr.10: 59958964-59958964 on Build GRCh38
rs991527231 Chr.10: 59959012-59959012 on Build GRCh38
rs781023343 Chr.10: 59958602-59958602 on Build GRCh38
rs931681064 Chr.10: 59958893-59958893 on Build GRCh38
rs866307740 Chr.10: 59958772-59958772 on Build GRCh38
rs900067271 Chr.10: 59958891-59958891 on Build GRCh38
rs566651515 Chr.10: 59958638-59958638 on Build GRCh38
rs764064272 Chr.10: 59958674-59958675 on Build GRCh38
rs890235271 Chr.10: 59958902-59958902 on Build GRCh38
rs1050045330 Chr.10: 59958899-59958899 on Build GRCh38
rs113678243 Chr.10: 59958668-59958668 on Build GRCh38
rs995049884 Chr.10: 59958967-59958967 on Build GRCh38
rs771439011 Chr.10: 59958658-59958658 on Build GRCh38
rs917362504 Chr.10: 59958769-59958769 on Build GRCh38
rs575846531 Chr.10: 59958917-59958917 on Build GRCh38
rs200484770 Chr.10: 59958689-59958689 on Build GRCh38
rs377177308 Chr.10: 59958796-59958796 on Build GRCh38
rs140968884 Chr.10: 59958569-59958569 on Build GRCh38
rs998714001 Chr.10: 59958604-59958604 on Build GRCh38
rs990468735 Chr.10: 59958847-59958847 on Build GRCh38
rs933845813 Chr.10: 59958930-59958930 on Build GRCh38
rs35215589 Chr.10: 59958844-59958844 on Build GRCh38
rs372310382 Chr.10: 59958878-59958878 on Build GRCh38
rs924812831 Chr.10: 59958801-59958801 on Build GRCh38
rs548808599 Chr.10: 59958758-59958758 on Build GRCh38
rs766046978 Chr.10: 59958574-59958574 on Build GRCh38
rs752638470 Chr.10: 59958821-59958821 on Build GRCh38

View Full Product Details