Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 101601305-101601806 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DPCD
Entrez Gene ID
25911
Gene Name
deleted in primary ciliary dyskinesia homolog (mouse)
Gene Aliases
-
UniGene
Hs.658128
Gene Chromosome Location
Chr.10: 101588332-101609653 on Build GRCh38
Transcripts
NM_015448.1 NR_036114.1 NR_036115.1

Gene Symbol
MIR3158-1
Entrez Gene ID
100422900
Gene Name
microRNA 3158-1
Gene Aliases
mir-3158-1
UniGene
Gene Chromosome Location
Chr.10: 101601417-101601497 on Build GRCh38
Transcripts
NM_015448.1 NR_036114.1 NR_036115.1

Gene Symbol
MIR3158-2
Entrez Gene ID
100423033
Gene Name
microRNA 3158-2
Gene Aliases
mir-3158-2
UniGene
Gene Chromosome Location
Chr.10: 101601417-101601497 on Build GRCh38
Transcripts
NM_015448.1 NR_036114.1 NR_036115.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6321573 c.389G>T p.R130I Chr.10: 101601321-101601321 on Build GRCh38
7258901 c.404+9C>T p.? Chr.10: 101601345-101601345 on Build GRCh38
7210021 c.374G>A p.R125H Chr.10: 101601306-101601306 on Build GRCh38
5999360 c.378C>T p.C126C Chr.10: 101601310-101601310 on Build GRCh38
6891610 c.402G>A p.K134K Chr.10: 101601334-101601334 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1038316849 Chr.10: 101601668-101601668 on Build GRCh38
rs762051949 Chr.10: 101601365-101601366 on Build GRCh38
rs763739283 Chr.10: 101601380-101601380 on Build GRCh38
rs183108669 Chr.10: 101601384-101601384 on Build GRCh38
rs1046772712 Chr.10: 101601564-101601564 on Build GRCh38
rs774007738 Chr.10: 101601492-101601492 on Build GRCh38
rs1019977571 Chr.10: 101601646-101601646 on Build GRCh38
rs780332966 Chr.10: 101601338-101601338 on Build GRCh38
rs774226423 Chr.10: 101601356-101601356 on Build GRCh38
rs368964908 Chr.10: 101601374-101601374 on Build GRCh38
rs768562986 Chr.10: 101601489-101601489 on Build GRCh38
rs191804222 Chr.10: 101601515-101601515 on Build GRCh38
rs975636031 Chr.10: 101601383-101601383 on Build GRCh38
rs7874 Chr.10: 101601331-101601331 on Build GRCh38
rs772856520 Chr.10: 101601519-101601519 on Build GRCh38
rs534587372 Chr.10: 101601426-101601426 on Build GRCh38
rs766643324 Chr.10: 101601404-101601404 on Build GRCh38
rs1029110551 Chr.10: 101601386-101601386 on Build GRCh38
rs923902784 Chr.10: 101601456-101601456 on Build GRCh38
rs548867129 Chr.10: 101601469-101601469 on Build GRCh38
rs112037539 Chr.10: 101601464-101601464 on Build GRCh38
rs574385107 Chr.10: 101601766-101601766 on Build GRCh38
rs776266924 Chr.10: 101601361-101601361 on Build GRCh38
rs954794391 Chr.10: 101601751-101601751 on Build GRCh38
rs766075869 Chr.10: 101601463-101601464 on Build GRCh38
rs767676479 Chr.10: 101601377-101601378 on Build GRCh38
rs899736333 Chr.10: 101601683-101601683 on Build GRCh38
rs1041729019 Chr.10: 101601476-101601476 on Build GRCh38
rs779424796 Chr.10: 101601405-101601405 on Build GRCh38
rs567025111 Chr.10: 101601488-101601488 on Build GRCh38
rs756370669 Chr.10: 101601402-101601402 on Build GRCh38
rs886809224 Chr.10: 101601654-101601654 on Build GRCh38
rs745401184 Chr.10: 101601453-101601453 on Build GRCh38
rs750621306 Chr.10: 101601383-101601384 on Build GRCh38
rs755370615 Chr.10: 101601417-101601417 on Build GRCh38
rs779487446 Chr.10: 101601346-101601346 on Build GRCh38
rs750482485 Chr.10: 101601400-101601400 on Build GRCh38
rs537939148 Chr.10: 101601660-101601660 on Build GRCh38
rs755246984 Chr.10: 101601345-101601345 on Build GRCh38
rs982839764 Chr.10: 101601458-101601458 on Build GRCh38
rs760531150 Chr.10: 101601453-101601453 on Build GRCh38
rs556238623 Chr.10: 101601662-101601662 on Build GRCh38
rs756317215 Chr.10: 101601333-101601333 on Build GRCh38
rs748532789 Chr.10: 101601350-101601350 on Build GRCh38
rs749635569 Chr.10: 101601340-101601340 on Build GRCh38
rs773750997 Chr.10: 101601690-101601690 on Build GRCh38
rs774350115 Chr.10: 101601381-101601381 on Build GRCh38
rs375618613 Chr.10: 101601365-101601365 on Build GRCh38
rs939045131 Chr.10: 101601558-101601558 on Build GRCh38
rs570591316 Chr.10: 101601591-101601591 on Build GRCh38
rs779136216 Chr.10: 101601423-101601423 on Build GRCh38
rs774967425 Chr.10: 101601377-101601377 on Build GRCh38
rs1011303911 Chr.10: 101601569-101601569 on Build GRCh38
rs1029887928 Chr.10: 101601740-101601740 on Build GRCh38
rs199517309 Chr.10: 101601378-101601378 on Build GRCh38
rs780888219 Chr.10: 101601446-101601446 on Build GRCh38
rs931397121 Chr.10: 101601451-101601451 on Build GRCh38
rs200909971 Chr.10: 101601352-101601352 on Build GRCh38
rs534410026 Chr.10: 101601548-101601548 on Build GRCh38
rs552789186 Chr.10: 101601556-101601556 on Build GRCh38
rs1002538615 Chr.10: 101601721-101601721 on Build GRCh38
rs761739077 Chr.10: 101601382-101601382 on Build GRCh38
rs908568638 Chr.10: 101601460-101601460 on Build GRCh38

View Full Product Details