Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 19507014-19507515 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NAV2
Entrez Gene ID
89797
Gene Name
neuron navigator 2
Gene Aliases
HELAD1, POMFIL2, RAINB1, STEERIN2, UNC53H2
UniGene
Hs.502116
Gene Chromosome Location
Chr.11: 19345200-20121601 on Build GRCh38
Transcripts
NM_001111018.1 XM_011520452.2 XM_017018520.1 XM_017018522.1 NR_049725.1

Gene Symbol
NAV2-AS5
Entrez Gene ID
100874012
Gene Name
NAV2 antisense RNA 5
Gene Aliases
-
UniGene
Hs.434602
Gene Chromosome Location
Chr.11: 19502672-19507229 on Build GRCh38
Transcripts
NM_001111018.1 XM_011520452.2 XM_017018520.1 XM_017018522.1 NR_049725.1

SNP Detail

SNP ID Chromosome Location
rs916764163 Chr.11: 19507364-19507364 on Build GRCh38
rs115853480 Chr.11: 19507295-19507295 on Build GRCh38
rs147928758 Chr.11: 19507267-19507267 on Build GRCh38
rs527638455 Chr.11: 19507465-19507465 on Build GRCh38
rs574201920 Chr.11: 19507331-19507331 on Build GRCh38
rs543797802 Chr.11: 19507195-19507195 on Build GRCh38
rs4423163 Chr.11: 19507275-19507275 on Build GRCh38
rs1052346377 Chr.11: 19507163-19507163 on Build GRCh38
rs996399435 Chr.11: 19507485-19507485 on Build GRCh38
rs960967109 Chr.11: 19507334-19507334 on Build GRCh38
rs144867698 Chr.11: 19507158-19507158 on Build GRCh38
rs561970407 Chr.11: 19507427-19507427 on Build GRCh38
rs1007078469 Chr.11: 19507274-19507274 on Build GRCh38
rs1043806380 Chr.11: 19507062-19507062 on Build GRCh38
rs979963066 Chr.11: 19507387-19507387 on Build GRCh38
rs539854024 Chr.11: 19507178-19507178 on Build GRCh38
rs948159819 Chr.11: 19507049-19507049 on Build GRCh38
rs941422406 Chr.11: 19507433-19507433 on Build GRCh38
rs998081669 Chr.11: 19507223-19507223 on Build GRCh38
rs545214774 Chr.11: 19507185-19507186 on Build GRCh38
rs978514666 Chr.11: 19507289-19507289 on Build GRCh38
rs75870897 Chr.11: 19507285-19507285 on Build GRCh38
rs139971716 Chr.11: 19507066-19507066 on Build GRCh38
rs969577342 Chr.11: 19507386-19507386 on Build GRCh38
rs993033224 Chr.11: 19507349-19507349 on Build GRCh38
rs1030645732 Chr.11: 19507225-19507225 on Build GRCh38
rs547808521 Chr.11: 19507473-19507473 on Build GRCh38
rs992404655 Chr.11: 19507359-19507359 on Build GRCh38
rs1025287412 Chr.11: 19507303-19507303 on Build GRCh38
rs958876901 Chr.11: 19507344-19507344 on Build GRCh38
rs904962950 Chr.11: 19507309-19507309 on Build GRCh38
rs752503313 Chr.11: 19507279-19507279 on Build GRCh38
rs1047467961 Chr.11: 19507371-19507371 on Build GRCh38
rs777554294 Chr.11: 19507450-19507450 on Build GRCh38
rs1023362193 Chr.11: 19507304-19507304 on Build GRCh38
rs1001666427 Chr.11: 19507314-19507314 on Build GRCh38
rs186237007 Chr.11: 19507202-19507202 on Build GRCh38
rs542109630 Chr.11: 19507419-19507419 on Build GRCh38
rs917475354 Chr.11: 19507363-19507363 on Build GRCh38
rs758637022 Chr.11: 19507219-19507219 on Build GRCh38
rs1019986729 Chr.11: 19507283-19507283 on Build GRCh38
rs2632051 Chr.11: 19507092-19507092 on Build GRCh38
rs1005597100 Chr.11: 19507134-19507134 on Build GRCh38
rs541865975 Chr.11: 19507475-19507475 on Build GRCh38
rs749309039 Chr.11: 19507467-19507467 on Build GRCh38
rs1051386162 Chr.11: 19507258-19507258 on Build GRCh38
rs555663825 Chr.11: 19507126-19507126 on Build GRCh38
rs761776634 Chr.11: 19507050-19507050 on Build GRCh38

View Full Product Details