Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 60660554-60661054 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LINC00301
Entrez Gene ID
283197
Gene Name
long intergenic non-protein coding RNA 301
Gene Aliases
C11orf64, NCRNA00301
UniGene
Hs.372650
Gene Chromosome Location
Chr.11: 60615751-60687149 on Build GRCh38
Transcripts
NR_026946.1

SNP Detail

SNP ID Chromosome Location
rs966655594 Chr.11: 60660746-60660746 on Build GRCh38
rs960122050 Chr.11: 60660979-60660979 on Build GRCh38
rs2306833 Chr.11: 60660609-60660609 on Build GRCh38
rs202067123 Chr.11: 60660871-60660872 on Build GRCh38
rs1040791290 Chr.11: 60660907-60660907 on Build GRCh38
rs943210614 Chr.11: 60660657-60660657 on Build GRCh38
rs1048130258 Chr.11: 60660935-60660935 on Build GRCh38
rs148799532 Chr.11: 60661006-60661006 on Build GRCh38
rs959908825 Chr.11: 60660715-60660715 on Build GRCh38
rs756260878 Chr.11: 60660856-60660856 on Build GRCh38
rs397840511 Chr.11: 60660907-60660907 on Build GRCh38
rs1001824820 Chr.11: 60660958-60660958 on Build GRCh38
rs1014056778 Chr.11: 60660989-60660989 on Build GRCh38
rs527350059 Chr.11: 60660729-60660729 on Build GRCh38
rs7943680 Chr.11: 60660713-60660713 on Build GRCh38
rs750656267 Chr.11: 60660851-60660851 on Build GRCh38
rs80012264 Chr.11: 60660792-60660792 on Build GRCh38
rs777591182 Chr.11: 60660951-60660951 on Build GRCh38
rs993851864 Chr.11: 60660659-60660659 on Build GRCh38
rs992165686 Chr.11: 60660716-60660716 on Build GRCh38
rs769349820 Chr.11: 60660852-60660853 on Build GRCh38
rs1002381283 Chr.11: 60660766-60660766 on Build GRCh38
rs896920942 Chr.11: 60660913-60660913 on Build GRCh38
rs78425093 Chr.11: 60660937-60660937 on Build GRCh38
rs3830318 Chr.11: 60660900-60660900 on Build GRCh38
rs556313213 Chr.11: 60660868-60660868 on Build GRCh38
rs1000699546 Chr.11: 60660872-60660872 on Build GRCh38
rs925115154 Chr.11: 60660621-60660621 on Build GRCh38
rs755431632 Chr.11: 60660760-60660760 on Build GRCh38
rs932958664 Chr.11: 60660698-60660698 on Build GRCh38
rs941679114 Chr.11: 60661007-60661007 on Build GRCh38
rs2306834 Chr.11: 60660692-60660692 on Build GRCh38
rs936956451 Chr.11: 60660848-60660848 on Build GRCh38
rs144811437 Chr.11: 60661011-60661011 on Build GRCh38
rs920187645 Chr.11: 60661009-60661009 on Build GRCh38
rs911343696 Chr.11: 60660878-60660878 on Build GRCh38
rs983637720 Chr.11: 60660994-60660994 on Build GRCh38
rs879257405 Chr.11: 60660653-60660653 on Build GRCh38
rs1026875410 Chr.11: 60660673-60660673 on Build GRCh38
rs974432390 Chr.11: 60660680-60660680 on Build GRCh38
rs145392653 Chr.11: 60660765-60660765 on Build GRCh38
rs918021453 Chr.11: 60660723-60660723 on Build GRCh38
rs780342408 Chr.11: 60660870-60660870 on Build GRCh38
rs2306835 Chr.11: 60660879-60660879 on Build GRCh38
rs966587660 Chr.11: 60660585-60660585 on Build GRCh38
rs993820792 Chr.11: 60660930-60660930 on Build GRCh38
rs1055589304 Chr.11: 60660776-60660776 on Build GRCh38
rs1001373607 Chr.11: 60660710-60660710 on Build GRCh38
rs1034273928 Chr.11: 60660972-60660972 on Build GRCh38

View Full Product Details