Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 61753917-61754422 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DKFZP434K028
Entrez Gene ID
26070
Gene Name
uncharacterized LOC26070
Gene Aliases
-
UniGene
Hs.564254
Gene Chromosome Location
Chr.11: 61754029-61757664 on Build GRCh38
Transcripts
NR_026882.1 NM_001127392.2 XM_005274222.1 XM_005274223.1 XM_005274224.1 XM_005274225.1 XM_005274226.1 XM_005274227.1 XM_005274228.1 XM_011545234.2

Gene Symbol
MYRF
Entrez Gene ID
745
Gene Name
myelin regulatory factor
Gene Aliases
C11orf9, MRF, Ndt80, pqn-47
UniGene
Hs.473109
Gene Chromosome Location
Chr.11: 61752617-61788518 on Build GRCh38
Transcripts
NR_026882.1 NM_001127392.2 XM_005274222.1 XM_005274223.1 XM_005274224.1 XM_005274225.1 XM_005274226.1 XM_005274227.1 XM_005274228.1 XM_011545234.2

SNP Detail

SNP ID Chromosome Location
rs905186804 Chr.11: 61754019-61754019 on Build GRCh38
rs926961834 Chr.11: 61754234-61754234 on Build GRCh38
rs198464 Chr.11: 61754149-61754149 on Build GRCh38
rs763759004 Chr.11: 61754376-61754376 on Build GRCh38
rs1032717329 Chr.11: 61754156-61754156 on Build GRCh38
rs572460729 Chr.11: 61754157-61754157 on Build GRCh38
rs1048839933 Chr.11: 61753952-61753952 on Build GRCh38
rs752188514 Chr.11: 61754257-61754257 on Build GRCh38
rs981038063 Chr.11: 61754233-61754233 on Build GRCh38
rs564295513 Chr.11: 61754335-61754335 on Build GRCh38
rs558309092 Chr.11: 61753983-61753983 on Build GRCh38
rs887614888 Chr.11: 61753970-61753970 on Build GRCh38
rs193234390 Chr.11: 61754277-61754277 on Build GRCh38
rs866375000 Chr.11: 61754206-61754206 on Build GRCh38
rs369197412 Chr.11: 61754271-61754271 on Build GRCh38
rs962804147 Chr.11: 61754394-61754394 on Build GRCh38
rs889119937 Chr.11: 61754366-61754366 on Build GRCh38
rs777104431 Chr.11: 61754021-61754021 on Build GRCh38
rs937097958 Chr.11: 61753946-61753946 on Build GRCh38
rs933804328 Chr.11: 61753955-61753955 on Build GRCh38
rs889170607 Chr.11: 61754032-61754032 on Build GRCh38
rs372727614 Chr.11: 61753966-61753966 on Build GRCh38
rs757999680 Chr.11: 61754275-61754275 on Build GRCh38
rs955265551 Chr.11: 61754249-61754249 on Build GRCh38
rs139967463 Chr.11: 61754263-61754263 on Build GRCh38
rs183750295 Chr.11: 61754363-61754363 on Build GRCh38
rs1036624197 Chr.11: 61754317-61754317 on Build GRCh38
rs901732822 Chr.11: 61754327-61754327 on Build GRCh38
rs1032469126 Chr.11: 61754089-61754089 on Build GRCh38
rs520987 Chr.11: 61753974-61753974 on Build GRCh38
rs901780754 Chr.11: 61754017-61754017 on Build GRCh38
rs113706608 Chr.11: 61754377-61754377 on Build GRCh38
rs541734386 Chr.11: 61754189-61754189 on Build GRCh38
rs996802969 Chr.11: 61754193-61754193 on Build GRCh38
rs902943152 Chr.11: 61754142-61754142 on Build GRCh38
rs368410455 Chr.11: 61754004-61754004 on Build GRCh38
rs576900227 Chr.11: 61753988-61753988 on Build GRCh38
rs1042556660 Chr.11: 61754094-61754094 on Build GRCh38
rs896681821 Chr.11: 61754048-61754048 on Build GRCh38
rs940553588 Chr.11: 61753971-61753971 on Build GRCh38
rs1000955626 Chr.11: 61754068-61754068 on Build GRCh38
rs764813872 Chr.11: 61754356-61754356 on Build GRCh38
rs117574068 Chr.11: 61754030-61754030 on Build GRCh38
rs941407528 Chr.11: 61754005-61754005 on Build GRCh38
rs1025200214 Chr.11: 61754192-61754192 on Build GRCh38
rs908982983 Chr.11: 61754272-61754272 on Build GRCh38

View Full Product Details